Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 30444587-30445077 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF536
Entrez Gene ID
9745
Gene Name
zinc finger protein 536
Gene Aliases
-
UniGene
Hs.378901
Gene Chromosome Location
Chr.19: 30224236-30713538 on Build GRCh38
Transcripts
NM_014717.2 XM_011527554.2 XM_011527555.2 XM_011527557.2 XM_011527558.2 XM_011527560.2 XM_017027527.1 XM_017027528.1 XM_017027529.1 XM_017027530.1 XM_017027531.1 XM_017027532.1 XM_017027533.1 XM_017027534.1 XM_017027535.1 XM_017027536.1 XM_017027537.1 XM_017027538.1 XM_017027539.1 XM_017027540.1 XM_017027541.1 XM_017027542.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
318331 c.1407G>T p.L469L Chr.19: 30444969-30444969 on Build GRCh38
710521 c.1214C>T p.S405F Chr.19: 30444776-30444776 on Build GRCh38
3892212 c.1215C>T p.S405S Chr.19: 30444777-30444777 on Build GRCh38
5493890 c.1246G>T p.G416C Chr.19: 30444808-30444808 on Build GRCh38
316841 c.1461_1462CC>AG p.S487>? Chr.19: 30445023-30445024 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764737514 Chr.19: 30444931-30444931 on Build GRCh38
rs771743074 Chr.19: 30444748-30444748 on Build GRCh38
rs370443078 Chr.19: 30444999-30444999 on Build GRCh38
rs759014086 Chr.19: 30444937-30444937 on Build GRCh38
rs974330651 Chr.19: 30444851-30444851 on Build GRCh38
rs932410747 Chr.19: 30444886-30444886 on Build GRCh38
rs757342308 Chr.19: 30444642-30444642 on Build GRCh38
rs1056417621 Chr.19: 30444920-30444920 on Build GRCh38
rs772800112 Chr.19: 30444753-30444753 on Build GRCh38
rs776624294 Chr.19: 30444981-30444981 on Build GRCh38
rs776358634 Chr.19: 30444861-30444861 on Build GRCh38
rs779579235 Chr.19: 30444736-30444736 on Build GRCh38
rs1052666191 Chr.19: 30444951-30444951 on Build GRCh38
rs764952724 Chr.19: 30444778-30444778 on Build GRCh38
rs376134887 Chr.19: 30444631-30444631 on Build GRCh38
rs565387236 Chr.19: 30445053-30445053 on Build GRCh38
rs375360729 Chr.19: 30444836-30444836 on Build GRCh38
rs770658949 Chr.19: 30444762-30444762 on Build GRCh38
rs766688042 Chr.19: 30444796-30444796 on Build GRCh38
rs200494382 Chr.19: 30444956-30444956 on Build GRCh38
rs756795445 Chr.19: 30444923-30444923 on Build GRCh38
rs776145243 Chr.19: 30444776-30444776 on Build GRCh38
rs770248403 Chr.19: 30445056-30445056 on Build GRCh38
rs750637190 Chr.19: 30444669-30444669 on Build GRCh38
rs758374956 Chr.19: 30445008-30445008 on Build GRCh38
rs534735196 Chr.19: 30444687-30444687 on Build GRCh38
rs577527616 Chr.19: 30444832-30444832 on Build GRCh38
rs759786682 Chr.19: 30444900-30444900 on Build GRCh38
rs749699610 Chr.19: 30444693-30444693 on Build GRCh38
rs957075769 Chr.19: 30444756-30444756 on Build GRCh38
rs755402618 Chr.19: 30444808-30444808 on Build GRCh38
rs763225279 Chr.19: 30445052-30445052 on Build GRCh38
rs779646468 Chr.19: 30444946-30444946 on Build GRCh38
rs765534247 Chr.19: 30444904-30444904 on Build GRCh38
rs756832106 Chr.19: 30444930-30444930 on Build GRCh38
rs767518850 Chr.19: 30444648-30444648 on Build GRCh38
rs779736621 Chr.19: 30445012-30445012 on Build GRCh38
rs775410184 Chr.19: 30444781-30444781 on Build GRCh38
rs759432182 Chr.19: 30444777-30444777 on Build GRCh38
rs768617605 Chr.19: 30444884-30444884 on Build GRCh38
rs573921467 Chr.19: 30444914-30444914 on Build GRCh38
rs759658668 Chr.19: 30444988-30444988 on Build GRCh38
rs201070042 Chr.19: 30444639-30444639 on Build GRCh38
rs184529144 Chr.19: 30445005-30445005 on Build GRCh38
rs200416337 Chr.19: 30444697-30444697 on Build GRCh38
rs750562223 Chr.19: 30444790-30444790 on Build GRCh38
rs61744158 Chr.19: 30444797-30444797 on Build GRCh38
rs769608026 Chr.19: 30444881-30444881 on Build GRCh38
rs917801085 Chr.19: 30444928-30444928 on Build GRCh38
rs61743191 Chr.19: 30445002-30445002 on Build GRCh38
rs779385772 Chr.19: 30444814-30444814 on Build GRCh38
rs761619375 Chr.19: 30444957-30444957 on Build GRCh38
rs1006523266 Chr.19: 30445044-30445044 on Build GRCh38
rs749144636 Chr.19: 30444947-30444947 on Build GRCh38
rs771069756 Chr.19: 30444966-30444966 on Build GRCh38
rs756307909 Chr.19: 30444672-30444672 on Build GRCh38
rs765623194 Chr.19: 30444995-30444995 on Build GRCh38
rs773279414 Chr.19: 30445038-30445038 on Build GRCh38
rs1002240451 Chr.19: 30444775-30444775 on Build GRCh38
rs745542323 Chr.19: 30444938-30444938 on Build GRCh38
rs150120798 Chr.19: 30444895-30444895 on Build GRCh38
rs769939047 Chr.19: 30445045-30445045 on Build GRCh38
rs752138319 Chr.19: 30445004-30445004 on Build GRCh38
rs141625611 Chr.19: 30445023-30445023 on Build GRCh38
rs746480413 Chr.19: 30444761-30444761 on Build GRCh38
rs758831432 Chr.19: 30444909-30444909 on Build GRCh38
rs965609448 Chr.19: 30444840-30444840 on Build GRCh38
rs140925588 Chr.19: 30444831-30444831 on Build GRCh38
rs543410703 Chr.19: 30444871-30444871 on Build GRCh38
rs780527084 Chr.19: 30444936-30444936 on Build GRCh38
rs770331654 Chr.19: 30444841-30444841 on Build GRCh38
rs905189576 Chr.19: 30444703-30444703 on Build GRCh38
rs755287534 Chr.19: 30444696-30444696 on Build GRCh38
rs7256870 Chr.19: 30444813-30444813 on Build GRCh38
rs755613678 Chr.19: 30445009-30445009 on Build GRCh38
rs747957400 Chr.19: 30445032-30445032 on Build GRCh38
rs563435645 Chr.19: 30444887-30444887 on Build GRCh38
rs771944197 Chr.19: 30445034-30445034 on Build GRCh38
rs1002294556 Chr.19: 30444987-30444987 on Build GRCh38
rs752956831 Chr.19: 30444996-30444996 on Build GRCh38
rs746003949 Chr.19: 30445043-30445043 on Build GRCh38
rs748602787 Chr.19: 30444744-30444744 on Build GRCh38
rs769482015 Chr.19: 30444941-30444941 on Build GRCh38
rs138204939 Chr.19: 30445020-30445020 on Build GRCh38
rs925638683 Chr.19: 30444903-30444903 on Build GRCh38
rs761077464 Chr.19: 30444793-30444793 on Build GRCh38
rs778404232 Chr.19: 30444912-30444912 on Build GRCh38
rs767602672 Chr.19: 30444789-30444789 on Build GRCh38
rs1046468051 Chr.19: 30444978-30444978 on Build GRCh38
rs762711372 Chr.19: 30444882-30444882 on Build GRCh38
rs753153162 Chr.19: 30444908-30444908 on Build GRCh38
rs778841122 Chr.19: 30444802-30444802 on Build GRCh38
rs148834715 Chr.19: 30444952-30444952 on Build GRCh38
rs146595841 Chr.19: 30444825-30444825 on Build GRCh38
rs867334103 Chr.19: 30444997-30444997 on Build GRCh38
rs775857517 Chr.19: 30445050-30445050 on Build GRCh38
rs753554537 Chr.19: 30444885-30444885 on Build GRCh38
rs768967733 Chr.19: 30444786-30444786 on Build GRCh38
rs1031148255 Chr.19: 30444705-30444705 on Build GRCh38
rs533533489 Chr.19: 30444783-30444783 on Build GRCh38
rs765223716 Chr.19: 30444633-30444633 on Build GRCh38
rs868755543 Chr.19: 30444812-30444812 on Build GRCh38
rs748875610 Chr.19: 30445017-30445017 on Build GRCh38

View Full Product Details