Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39658381-39658878 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LGALS16
Entrez Gene ID
148003
Gene Name
galectin 16
Gene Aliases
-
UniGene
Hs.667238
Gene Chromosome Location
Chr.19: 39655894-39660647 on Build GRCh38
Transcripts
NM_001190441.2

SNP Detail

SNP ID Chromosome Location
rs556222718 Chr.19: 39658614-39658614 on Build GRCh38
rs780869333 Chr.19: 39658533-39658533 on Build GRCh38
rs567987381 Chr.19: 39658582-39658582 on Build GRCh38
rs992381260 Chr.19: 39658402-39658402 on Build GRCh38
rs775802391 Chr.19: 39658474-39658474 on Build GRCh38
rs972445651 Chr.19: 39658773-39658773 on Build GRCh38
rs762301779 Chr.19: 39658583-39658583 on Build GRCh38
rs138874309 Chr.19: 39658502-39658502 on Build GRCh38
rs948227664 Chr.19: 39658425-39658425 on Build GRCh38
rs979215384 Chr.19: 39658783-39658783 on Build GRCh38
rs201930366 Chr.19: 39658462-39658462 on Build GRCh38
rs775701877 Chr.19: 39658479-39658479 on Build GRCh38
rs933657229 Chr.19: 39658791-39658791 on Build GRCh38
rs538753481 Chr.19: 39658642-39658642 on Build GRCh38
rs764476165 Chr.19: 39658482-39658482 on Build GRCh38
rs775823514 Chr.19: 39658565-39658565 on Build GRCh38
rs550603799 Chr.19: 39658523-39658523 on Build GRCh38
rs991490573 Chr.19: 39658740-39658740 on Build GRCh38
rs755812739 Chr.19: 39658619-39658619 on Build GRCh38
rs915929801 Chr.19: 39658767-39658767 on Build GRCh38
rs960716251 Chr.19: 39658402-39658403 on Build GRCh38
rs941071369 Chr.19: 39658443-39658443 on Build GRCh38
rs933688203 Chr.19: 39658512-39658512 on Build GRCh38
rs746005629 Chr.19: 39658553-39658553 on Build GRCh38
rs773245237 Chr.19: 39658460-39658460 on Build GRCh38
rs1860134 Chr.19: 39658657-39658657 on Build GRCh38
rs909622897 Chr.19: 39658441-39658441 on Build GRCh38
rs759839263 Chr.19: 39658466-39658466 on Build GRCh38
rs757965871 Chr.19: 39658417-39658417 on Build GRCh38
rs752412408 Chr.19: 39658616-39658616 on Build GRCh38
rs769600074 Chr.19: 39658455-39658455 on Build GRCh38
rs1050953567 Chr.19: 39658537-39658537 on Build GRCh38
rs753645682 Chr.19: 39658637-39658637 on Build GRCh38
rs774308933 Chr.19: 39658523-39658523 on Build GRCh38
rs757249480 Chr.19: 39658641-39658641 on Build GRCh38
rs934452622 Chr.19: 39658835-39658835 on Build GRCh38
rs761148943 Chr.19: 39658481-39658481 on Build GRCh38
rs770251530 Chr.19: 39658663-39658663 on Build GRCh38
rs776935637 Chr.19: 39658580-39658580 on Build GRCh38
rs773353226 Chr.19: 39658646-39658646 on Build GRCh38
rs918361556 Chr.19: 39658774-39658774 on Build GRCh38
rs772310905 Chr.19: 39658643-39658643 on Build GRCh38
rs780963327 Chr.19: 39658428-39658428 on Build GRCh38
rs751044406 Chr.19: 39658587-39658587 on Build GRCh38
rs960302198 Chr.19: 39658713-39658713 on Build GRCh38
rs779633728 Chr.19: 39658418-39658418 on Build GRCh38
rs754703401 Chr.19: 39658421-39658421 on Build GRCh38
rs777039011 Chr.19: 39658658-39658658 on Build GRCh38
rs758994018 Chr.19: 39658676-39658676 on Build GRCh38
rs924697340 Chr.19: 39658819-39658819 on Build GRCh38
rs747258971 Chr.19: 39658650-39658650 on Build GRCh38
rs73048204 Chr.19: 39658702-39658702 on Build GRCh38
rs772402294 Chr.19: 39658558-39658558 on Build GRCh38
rs549304633 Chr.19: 39658567-39658567 on Build GRCh38
rs771152415 Chr.19: 39658463-39658463 on Build GRCh38
rs748035302 Chr.19: 39658437-39658437 on Build GRCh38
rs752399690 Chr.19: 39658534-39658534 on Build GRCh38
rs562528341 Chr.19: 39658530-39658530 on Build GRCh38
rs184657849 Chr.19: 39658549-39658549 on Build GRCh38
rs566345929 Chr.19: 39658419-39658419 on Build GRCh38
rs748558919 Chr.19: 39658660-39658660 on Build GRCh38
rs746126008 Chr.19: 39658459-39658459 on Build GRCh38
rs534353150 Chr.19: 39658566-39658566 on Build GRCh38
rs35476378 Chr.19: 39658542-39658542 on Build GRCh38
rs952589082 Chr.19: 39658414-39658414 on Build GRCh38
rs761682840 Chr.19: 39658644-39658645 on Build GRCh38
rs187604453 Chr.19: 39658623-39658623 on Build GRCh38
rs767063992 Chr.19: 39658689-39658689 on Build GRCh38
rs749877785 Chr.19: 39658413-39658413 on Build GRCh38
rs147471254 Chr.19: 39658585-39658585 on Build GRCh38
rs772487352 Chr.19: 39658473-39658473 on Build GRCh38
rs552315094 Chr.19: 39658649-39658649 on Build GRCh38
rs774653270 Chr.19: 39658464-39658464 on Build GRCh38
rs1040949496 Chr.19: 39658595-39658595 on Build GRCh38
rs147244273 Chr.19: 39658776-39658776 on Build GRCh38
rs997104 Chr.19: 39658690-39658690 on Build GRCh38
rs116331309 Chr.19: 39658531-39658531 on Build GRCh38
rs1050754882 Chr.19: 39658824-39658824 on Build GRCh38
rs889466806 Chr.19: 39658828-39658828 on Build GRCh38
rs149409914 Chr.19: 39658546-39658546 on Build GRCh38
rs928895592 Chr.19: 39658446-39658446 on Build GRCh38
rs145491433 Chr.19: 39658771-39658771 on Build GRCh38
rs759083932 Chr.19: 39658605-39658605 on Build GRCh38
rs776923011 Chr.19: 39658452-39658452 on Build GRCh38
rs773736751 Chr.19: 39658665-39658665 on Build GRCh38

View Full Product Details