Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39828195-39828692 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYRK1B
Entrez Gene ID
9149
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 1B
Gene Aliases
AOMS3, MIRK
UniGene
Hs.130988
Gene Chromosome Location
Chr.19: 39825347-39834233 on Build GRCh38
Transcripts
NM_004714.2 NM_006483.2 NM_006484.2 XM_005259398.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
291996 c.749C>G p.P250R Chr.19: 39828355-39828355 on Build GRCh38
5389754 c.643C>T p.R215W Chr.19: 39828461-39828461 on Build GRCh38
7103813 c.780C>G p.F260L Chr.19: 39828324-39828324 on Build GRCh38
1590096 c.624C>T p.G208G Chr.19: 39828480-39828480 on Build GRCh38
3533844 c.765C>T p.I255I Chr.19: 39828339-39828339 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs923009178 Chr.19: 39828611-39828611 on Build GRCh38
rs889000417 Chr.19: 39828474-39828474 on Build GRCh38
rs765975316 Chr.19: 39828548-39828548 on Build GRCh38
rs755481777 Chr.19: 39828319-39828319 on Build GRCh38
rs775510898 Chr.19: 39828260-39828260 on Build GRCh38
rs761239809 Chr.19: 39828614-39828614 on Build GRCh38
rs761998506 Chr.19: 39828270-39828270 on Build GRCh38
rs748256087 Chr.19: 39828389-39828389 on Build GRCh38
rs746160700 Chr.19: 39828591-39828591 on Build GRCh38
rs1037161500 Chr.19: 39828594-39828594 on Build GRCh38
rs750085108 Chr.19: 39828462-39828462 on Build GRCh38
rs567231327 Chr.19: 39828435-39828435 on Build GRCh38
rs763155290 Chr.19: 39828399-39828399 on Build GRCh38
rs996451047 Chr.19: 39828265-39828265 on Build GRCh38
rs766716069 Chr.19: 39828312-39828312 on Build GRCh38
rs150608731 Chr.19: 39828542-39828542 on Build GRCh38
rs1035754740 Chr.19: 39828498-39828498 on Build GRCh38
rs771083795 Chr.19: 39828501-39828501 on Build GRCh38
rs372504975 Chr.19: 39828510-39828510 on Build GRCh38
rs756096765 Chr.19: 39828579-39828579 on Build GRCh38
rs764808279 Chr.19: 39828527-39828527 on Build GRCh38
rs922520369 Chr.19: 39828556-39828556 on Build GRCh38
rs779017023 Chr.19: 39828588-39828588 on Build GRCh38
rs776640518 Chr.19: 39828378-39828378 on Build GRCh38
rs376341893 Chr.19: 39828414-39828414 on Build GRCh38
rs992170060 Chr.19: 39828287-39828287 on Build GRCh38
rs768522719 Chr.19: 39828363-39828363 on Build GRCh38
rs900876850 Chr.19: 39828239-39828239 on Build GRCh38
rs765464654 Chr.19: 39828283-39828283 on Build GRCh38
rs375195429 Chr.19: 39828271-39828271 on Build GRCh38
rs771887480 Chr.19: 39828344-39828344 on Build GRCh38
rs147484701 Chr.19: 39828436-39828436 on Build GRCh38
rs112350522 Chr.19: 39828620-39828620 on Build GRCh38
rs924400394 Chr.19: 39828236-39828236 on Build GRCh38
rs1042903412 Chr.19: 39828230-39828230 on Build GRCh38
rs758024803 Chr.19: 39828471-39828471 on Build GRCh38
rs1041331765 Chr.19: 39828238-39828238 on Build GRCh38
rs762371793 Chr.19: 39828632-39828632 on Build GRCh38
rs972244887 Chr.19: 39828594-39828595 on Build GRCh38
rs141849649 Chr.19: 39828485-39828485 on Build GRCh38
rs779778950 Chr.19: 39828345-39828345 on Build GRCh38
rs750105462 Chr.19: 39828293-39828293 on Build GRCh38
rs771838065 Chr.19: 39828257-39828257 on Build GRCh38
rs773323978 Chr.19: 39828492-39828492 on Build GRCh38
rs760041088 Chr.19: 39828432-39828432 on Build GRCh38
rs1022989703 Chr.19: 39828659-39828659 on Build GRCh38
rs779874553 Chr.19: 39828477-39828477 on Build GRCh38
rs756931266 Chr.19: 39828327-39828327 on Build GRCh38
rs745689275 Chr.19: 39828256-39828256 on Build GRCh38
rs746346346 Chr.19: 39828480-39828480 on Build GRCh38
rs545713001 Chr.19: 39828638-39828638 on Build GRCh38
rs888076509 Chr.19: 39828590-39828590 on Build GRCh38
rs898014915 Chr.19: 39828605-39828605 on Build GRCh38
rs772735330 Chr.19: 39828621-39828621 on Build GRCh38
rs371539604 Chr.19: 39828483-39828483 on Build GRCh38
rs201053732 Chr.19: 39828534-39828534 on Build GRCh38
rs751298437 Chr.19: 39828554-39828554 on Build GRCh38
rs538827167 Chr.19: 39828578-39828578 on Build GRCh38
rs369511200 Chr.19: 39828288-39828288 on Build GRCh38
rs776037379 Chr.19: 39828608-39828608 on Build GRCh38
rs35858874 Chr.19: 39828404-39828404 on Build GRCh38
rs745601238 Chr.19: 39828335-39828335 on Build GRCh38
rs148106934 Chr.19: 39828448-39828448 on Build GRCh38
rs759826670 Chr.19: 39828504-39828504 on Build GRCh38
rs778713560 Chr.19: 39828334-39828334 on Build GRCh38
rs563701615 Chr.19: 39828649-39828649 on Build GRCh38
rs76318664 Chr.19: 39828592-39828592 on Build GRCh38
rs143588223 Chr.19: 39828323-39828323 on Build GRCh38
rs758018879 Chr.19: 39828547-39828547 on Build GRCh38
rs931476895 Chr.19: 39828300-39828300 on Build GRCh38
rs749506295 Chr.19: 39828500-39828500 on Build GRCh38
rs993791983 Chr.19: 39828434-39828434 on Build GRCh38
rs538308215 Chr.19: 39828450-39828450 on Build GRCh38
rs774739741 Chr.19: 39828503-39828503 on Build GRCh38
rs997170423 Chr.19: 39828461-39828461 on Build GRCh38
rs754630649 Chr.19: 39828274-39828280 on Build GRCh38
rs1050309264 Chr.19: 39828465-39828465 on Build GRCh38
rs1049395189 Chr.19: 39828589-39828589 on Build GRCh38
rs56077108 Chr.19: 39828587-39828587 on Build GRCh38
rs763375868 Chr.19: 39828289-39828289 on Build GRCh38
rs776730186 Chr.19: 39828269-39828269 on Build GRCh38
rs1013135804 Chr.19: 39828643-39828643 on Build GRCh38
rs377036516 Chr.19: 39828513-39828513 on Build GRCh38
rs754777135 Chr.19: 39828564-39828564 on Build GRCh38
rs768079960 Chr.19: 39828507-39828507 on Build GRCh38
rs373276735 Chr.19: 39828324-39828324 on Build GRCh38
rs746933234 Chr.19: 39828349-39828349 on Build GRCh38
rs781018755 Chr.19: 39828569-39828569 on Build GRCh38
rs773432280 Chr.19: 39828398-39828398 on Build GRCh38
rs769864310 Chr.19: 39828489-39828489 on Build GRCh38
rs556237495 Chr.19: 39828479-39828479 on Build GRCh38
rs777863705 Chr.19: 39828580-39828580 on Build GRCh38
rs934612811 Chr.19: 39828237-39828237 on Build GRCh38
rs928608143 Chr.19: 39828219-39828219 on Build GRCh38
rs769933803 Chr.19: 39828390-39828390 on Build GRCh38
rs760585920 Chr.19: 39828268-39828268 on Build GRCh38
rs947234411 Chr.19: 39828223-39828223 on Build GRCh38

View Full Product Details