Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40729071-40729486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITPKC
Entrez Gene ID
80271
Gene Name
inositol-trisphosphate 3-kinase C
Gene Aliases
IP3-3KC, IP3KC
UniGene
Hs.515415
Gene Chromosome Location
Chr.19: 40717103-40740860 on Build GRCh38
Transcripts
NM_025194.2 XM_017027324.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1681051 c.1373G>T p.G458V Chr.19: 40729319-40729319 on Build GRCh38
5718468 c.1442C>T p.S481F Chr.19: 40729388-40729388 on Build GRCh38
6446381 c.1310A>G p.E437G Chr.19: 40729256-40729256 on Build GRCh38
5748065 c.1313A>G p.Q438R Chr.19: 40729259-40729259 on Build GRCh38
115980 c.1345C>G p.L449V Chr.19: 40729291-40729291 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs558083572 Chr.19: 40729164-40729164 on Build GRCh38
rs773751212 Chr.19: 40729274-40729274 on Build GRCh38
rs780285372 Chr.19: 40729337-40729337 on Build GRCh38
rs182503287 Chr.19: 40729196-40729196 on Build GRCh38
rs35158136 Chr.19: 40729143-40729144 on Build GRCh38
rs957439554 Chr.19: 40729227-40729227 on Build GRCh38
rs151041227 Chr.19: 40729175-40729175 on Build GRCh38
rs1040287848 Chr.19: 40729099-40729099 on Build GRCh38
rs142308355 Chr.19: 40729229-40729229 on Build GRCh38
rs775892434 Chr.19: 40729165-40729165 on Build GRCh38
rs774980784 Chr.19: 40729289-40729289 on Build GRCh38
rs187084067 Chr.19: 40729451-40729451 on Build GRCh38
rs747028866 Chr.19: 40729344-40729344 on Build GRCh38
rs994997521 Chr.19: 40729121-40729121 on Build GRCh38
rs369015336 Chr.19: 40729457-40729457 on Build GRCh38
rs10420685 Chr.19: 40729215-40729215 on Build GRCh38
rs751093488 Chr.19: 40729187-40729187 on Build GRCh38
rs753695795 Chr.19: 40729295-40729295 on Build GRCh38
rs900601396 Chr.19: 40729318-40729318 on Build GRCh38
rs749864734 Chr.19: 40729413-40729413 on Build GRCh38
rs569548945 Chr.19: 40729130-40729130 on Build GRCh38
rs757237709 Chr.19: 40729216-40729216 on Build GRCh38
rs867628325 Chr.19: 40729200-40729200 on Build GRCh38
rs112628847 Chr.19: 40729262-40729262 on Build GRCh38
rs369305104 Chr.19: 40729222-40729222 on Build GRCh38
rs752398741 Chr.19: 40729205-40729205 on Build GRCh38
rs10420651 Chr.19: 40729133-40729133 on Build GRCh38
rs758404959 Chr.19: 40729321-40729321 on Build GRCh38
rs763601962 Chr.19: 40729294-40729294 on Build GRCh38
rs765606668 Chr.19: 40729184-40729184 on Build GRCh38
rs913354890 Chr.19: 40729240-40729240 on Build GRCh38
rs1020544429 Chr.19: 40729453-40729453 on Build GRCh38
rs773556820 Chr.19: 40729399-40729399 on Build GRCh38
rs765100469 Chr.19: 40729314-40729314 on Build GRCh38
rs750442375 Chr.19: 40729315-40729315 on Build GRCh38
rs769122852 Chr.19: 40729173-40729173 on Build GRCh38
rs776194505 Chr.19: 40729436-40729436 on Build GRCh38
rs763431871 Chr.19: 40729276-40729276 on Build GRCh38
rs778673488 Chr.19: 40729219-40729219 on Build GRCh38
rs745891358 Chr.19: 40729155-40729155 on Build GRCh38
rs757083194 Chr.19: 40729422-40729432 on Build GRCh38
rs900213683 Chr.19: 40729110-40729110 on Build GRCh38
rs1004580424 Chr.19: 40729144-40729144 on Build GRCh38
rs757039960 Chr.19: 40729302-40729302 on Build GRCh38
rs201169443 Chr.19: 40729368-40729368 on Build GRCh38
rs867402774 Chr.19: 40729127-40729127 on Build GRCh38
rs760957070 Chr.19: 40729171-40729171 on Build GRCh38
rs1034303277 Chr.19: 40729460-40729460 on Build GRCh38
rs760060444 Chr.19: 40729423-40729423 on Build GRCh38
rs368662548 Chr.19: 40729419-40729419 on Build GRCh38
rs767075589 Chr.19: 40729279-40729279 on Build GRCh38
rs770703385 Chr.19: 40729153-40729153 on Build GRCh38
rs777624525 Chr.19: 40729212-40729212 on Build GRCh38
rs907079660 Chr.19: 40729456-40729456 on Build GRCh38
rs1049269721 Chr.19: 40729412-40729412 on Build GRCh38
rs755754696 Chr.19: 40729206-40729206 on Build GRCh38
rs1027444508 Chr.19: 40729131-40729131 on Build GRCh38
rs373104401 Chr.19: 40729247-40729247 on Build GRCh38
rs779060278 Chr.19: 40729154-40729154 on Build GRCh38
rs774999187 Chr.19: 40729422-40729422 on Build GRCh38
rs907858196 Chr.19: 40729316-40729316 on Build GRCh38
rs370620477 Chr.19: 40729266-40729266 on Build GRCh38
rs755251942 Chr.19: 40729353-40729353 on Build GRCh38
rs1020368982 Chr.19: 40729143-40729143 on Build GRCh38
rs562478696 Chr.19: 40729290-40729290 on Build GRCh38
rs763626955 Chr.19: 40729431-40729431 on Build GRCh38
rs1004523071 Chr.19: 40729425-40729425 on Build GRCh38
rs770366804 Chr.19: 40729267-40729267 on Build GRCh38
rs770108570 Chr.19: 40729389-40729389 on Build GRCh38
rs780267002 Chr.19: 40729241-40729241 on Build GRCh38
rs781232808 Chr.19: 40729355-40729355 on Build GRCh38
rs376093815 Chr.19: 40729382-40729382 on Build GRCh38
rs944730339 Chr.19: 40729317-40729317 on Build GRCh38
rs79940110 Chr.19: 40729199-40729199 on Build GRCh38

View Full Product Details