Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41386418-41386927 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC5
Entrez Gene ID
56915
Gene Name
exosome component 5
Gene Aliases
RRP41B, RRP46, Rrp46p, hRrp46p, p12B
UniGene
Hs.283741
Gene Chromosome Location
Chr.19: 41386370-41397351 on Build GRCh38
Transcripts
NM_020158.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5504551 c.639C>G p.A213A Chr.19: 41386702-41386702 on Build GRCh38
5447323 c.691C>T p.R231C Chr.19: 41386650-41386650 on Build GRCh38
6196546 c.656_657insCGTCTTC p.F223fs*>16 Chr.19: 41386684-41386685 on Build GRCh38
4942180 c.617T>C p.L206P Chr.19: 41386724-41386724 on Build GRCh38
1205844 c.650C>T p.S217L Chr.19: 41386691-41386691 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1010116411 Chr.19: 41386442-41386442 on Build GRCh38
rs945600962 Chr.19: 41386842-41386842 on Build GRCh38
rs759597734 Chr.19: 41386758-41386758 on Build GRCh38
rs1010602119 Chr.19: 41386587-41386587 on Build GRCh38
rs939236283 Chr.19: 41386838-41386838 on Build GRCh38
rs777754423 Chr.19: 41386730-41386730 on Build GRCh38
rs749337722 Chr.19: 41386845-41386845 on Build GRCh38
rs147964002 Chr.19: 41386677-41386677 on Build GRCh38
rs772562372 Chr.19: 41386741-41386741 on Build GRCh38
rs116100212 Chr.19: 41386734-41386734 on Build GRCh38
rs888278792 Chr.19: 41386443-41386443 on Build GRCh38
rs1033744873 Chr.19: 41386561-41386561 on Build GRCh38
rs768634351 Chr.19: 41386637-41386637 on Build GRCh38
rs549559334 Chr.19: 41386584-41386584 on Build GRCh38
rs35219501 Chr.19: 41386668-41386668 on Build GRCh38
rs779589894 Chr.19: 41386616-41386616 on Build GRCh38
rs781207479 Chr.19: 41386721-41386721 on Build GRCh38
rs756427806 Chr.19: 41386688-41386688 on Build GRCh38
rs970445078 Chr.19: 41386452-41386452 on Build GRCh38
rs978672225 Chr.19: 41386614-41386614 on Build GRCh38
rs66763950 Chr.19: 41386447-41386447 on Build GRCh38
rs9642 Chr.19: 41386514-41386514 on Build GRCh38
rs150745081 Chr.19: 41386802-41386802 on Build GRCh38
rs984804073 Chr.19: 41386812-41386812 on Build GRCh38
rs748120442 Chr.19: 41386728-41386728 on Build GRCh38
rs753050560 Chr.19: 41386565-41386565 on Build GRCh38
rs540292584 Chr.19: 41386690-41386690 on Build GRCh38
rs751654580 Chr.19: 41386676-41386676 on Build GRCh38
rs564403948 Chr.19: 41386507-41386507 on Build GRCh38
rs141286660 Chr.19: 41386729-41386729 on Build GRCh38
rs771175954 Chr.19: 41386745-41386745 on Build GRCh38
rs1002338686 Chr.19: 41386855-41386855 on Build GRCh38
rs867734815 Chr.19: 41386822-41386822 on Build GRCh38
rs572790157 Chr.19: 41386517-41386517 on Build GRCh38
rs768771795 Chr.19: 41386849-41386849 on Build GRCh38
rs372991427 Chr.19: 41386755-41386755 on Build GRCh38
rs756487101 Chr.19: 41386580-41386580 on Build GRCh38
rs79234312 Chr.19: 41386759-41386759 on Build GRCh38
rs767958737 Chr.19: 41386683-41386683 on Build GRCh38
rs1046638441 Chr.19: 41386844-41386844 on Build GRCh38
rs186043263 Chr.19: 41386790-41386790 on Build GRCh38
rs563286016 Chr.19: 41386612-41386612 on Build GRCh38
rs180811354 Chr.19: 41386851-41386851 on Build GRCh38
rs186386564 Chr.19: 41386897-41386897 on Build GRCh38
rs927778915 Chr.19: 41386831-41386831 on Build GRCh38
rs773520721 Chr.19: 41386665-41386665 on Build GRCh38
rs758033402 Chr.19: 41386695-41386695 on Build GRCh38
rs973346311 Chr.19: 41386544-41386544 on Build GRCh38
rs944113775 Chr.19: 41386608-41386608 on Build GRCh38
rs189332626 Chr.19: 41386809-41386809 on Build GRCh38
rs1015477609 Chr.19: 41386470-41386470 on Build GRCh38
rs1045843715 Chr.19: 41386829-41386829 on Build GRCh38
rs73931470 Chr.19: 41386887-41386887 on Build GRCh38
rs771787458 Chr.19: 41386734-41386735 on Build GRCh38
rs746514345 Chr.19: 41386705-41386705 on Build GRCh38
rs878868792 Chr.19: 41386778-41386778 on Build GRCh38
rs780026452 Chr.19: 41386817-41386817 on Build GRCh38
rs769714615 Chr.19: 41386661-41386661 on Build GRCh38
rs956899067 Chr.19: 41386441-41386441 on Build GRCh38
rs761642243 Chr.19: 41386659-41386659 on Build GRCh38
rs1055082877 Chr.19: 41386696-41386696 on Build GRCh38
rs531084639 Chr.19: 41386684-41386684 on Build GRCh38
rs36035938 Chr.19: 41386702-41386702 on Build GRCh38
rs550225229 Chr.19: 41386667-41386667 on Build GRCh38
rs746537581 Chr.19: 41386617-41386617 on Build GRCh38
rs781656020 Chr.19: 41386563-41386563 on Build GRCh38
rs148660919 Chr.19: 41386611-41386611 on Build GRCh38
rs912125278 Chr.19: 41386650-41386650 on Build GRCh38
rs768062768 Chr.19: 41386706-41386706 on Build GRCh38
rs906455579 Chr.19: 41386493-41386493 on Build GRCh38
rs754247072 Chr.19: 41386691-41386691 on Build GRCh38
rs775925286 Chr.19: 41386777-41386777 on Build GRCh38
rs774453258 Chr.19: 41386746-41386746 on Build GRCh38
rs963662428 Chr.19: 41386526-41386526 on Build GRCh38
rs766504163 Chr.19: 41386501-41386501 on Build GRCh38
rs1004052586 Chr.19: 41386461-41386461 on Build GRCh38
rs879227847 Chr.19: 41386731-41386731 on Build GRCh38
rs975471770 Chr.19: 41386786-41386786 on Build GRCh38
rs747706254 Chr.19: 41386715-41386716 on Build GRCh38
rs144175057 Chr.19: 41386600-41386600 on Build GRCh38
rs554838531 Chr.19: 41386649-41386649 on Build GRCh38

View Full Product Details