Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41410701-41411204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCKDHA
Entrez Gene ID
593
Gene Name
branched chain keto acid dehydrogenase E1, alpha polypeptide
Gene Aliases
BCKDE1A, MSU, MSUD1, OVD1A
UniGene
Hs.433307
Gene Chromosome Location
Chr.19: 41397789-41425005 on Build GRCh38
Transcripts
NM_000709.3 NM_001164783.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
712786 c.252G>A p.R84R Chr.19: 41410780-41410780 on Build GRCh38
4665371 c.350G>A p.R117H Chr.19: 41410984-41410984 on Build GRCh38
5484306 c.174G>A p.S58S Chr.19: 41410702-41410702 on Build GRCh38
6443135 c.345_346GG>AA p.M115_D116>IN Chr.19: 41410979-41410980 on Build GRCh38
4665370 c.317A>G p.Y106C Chr.19: 41410951-41410951 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755338376 Chr.19: 41410800-41410800 on Build GRCh38
rs772698478 Chr.19: 41410895-41410895 on Build GRCh38
rs398123497 Chr.19: 41410825-41410825 on Build GRCh38
rs147510183 Chr.19: 41410774-41410774 on Build GRCh38
rs775424394 Chr.19: 41410745-41410745 on Build GRCh38
rs201366184 Chr.19: 41410964-41410964 on Build GRCh38
rs769086498 Chr.19: 41411035-41411035 on Build GRCh38
rs200646708 Chr.19: 41410918-41410918 on Build GRCh38
rs199704138 Chr.19: 41410899-41410899 on Build GRCh38
rs373555514 Chr.19: 41410742-41410742 on Build GRCh38
rs552517766 Chr.19: 41410773-41410773 on Build GRCh38
rs938963707 Chr.19: 41411132-41411133 on Build GRCh38
rs398123496 Chr.19: 41410817-41410817 on Build GRCh38
rs201219694 Chr.19: 41410863-41410863 on Build GRCh38
rs979507391 Chr.19: 41411137-41411137 on Build GRCh38
rs754135764 Chr.19: 41411033-41411034 on Build GRCh38
rs780103142 Chr.19: 41410856-41410856 on Build GRCh38
rs746001370 Chr.19: 41410960-41410960 on Build GRCh38
rs757507972 Chr.19: 41410956-41410956 on Build GRCh38
rs201500256 Chr.19: 41410843-41410843 on Build GRCh38
rs192832793 Chr.19: 41411021-41411021 on Build GRCh38
rs934456348 Chr.19: 41411167-41411167 on Build GRCh38
rs373438406 Chr.19: 41410851-41410851 on Build GRCh38
rs777132193 Chr.19: 41411036-41411036 on Build GRCh38
rs759842379 Chr.19: 41410842-41410842 on Build GRCh38
rs775387563 Chr.19: 41411027-41411027 on Build GRCh38
rs952949299 Chr.19: 41410963-41410963 on Build GRCh38
rs775709563 Chr.19: 41410898-41410898 on Build GRCh38
rs398123495 Chr.19: 41410777-41410777 on Build GRCh38
rs778250707 Chr.19: 41410813-41410813 on Build GRCh38
rs147370725 Chr.19: 41410740-41410740 on Build GRCh38
rs142967869 Chr.19: 41410928-41410928 on Build GRCh38
rs774803865 Chr.19: 41411113-41411113 on Build GRCh38
rs561186353 Chr.19: 41410835-41410835 on Build GRCh38
rs781428377 Chr.19: 41410827-41410827 on Build GRCh38
rs1017097372 Chr.19: 41411087-41411087 on Build GRCh38
rs73931479 Chr.19: 41411083-41411083 on Build GRCh38
rs752700457 Chr.19: 41411005-41411005 on Build GRCh38
rs762043084 Chr.19: 41411094-41411094 on Build GRCh38
rs755966459 Chr.19: 41410940-41410940 on Build GRCh38
rs368375544 Chr.19: 41410837-41410837 on Build GRCh38
rs933606851 Chr.19: 41411019-41411019 on Build GRCh38
rs968089679 Chr.19: 41411123-41411123 on Build GRCh38
rs779124818 Chr.19: 41411032-41411032 on Build GRCh38
rs368479345 Chr.19: 41411089-41411089 on Build GRCh38
rs749324349 Chr.19: 41410882-41410882 on Build GRCh38
rs765966197 Chr.19: 41410919-41410919 on Build GRCh38
rs532803527 Chr.19: 41410927-41410927 on Build GRCh38
rs756747400 Chr.19: 41410736-41410736 on Build GRCh38
rs769090259 Chr.19: 41410975-41410975 on Build GRCh38
rs1027441046 Chr.19: 41411118-41411118 on Build GRCh38
rs368891152 Chr.19: 41410811-41410811 on Build GRCh38
rs188135164 Chr.19: 41410983-41410983 on Build GRCh38
rs774899842 Chr.19: 41411058-41411058 on Build GRCh38
rs552552525 Chr.19: 41411111-41411111 on Build GRCh38
rs761915209 Chr.19: 41410769-41410769 on Build GRCh38
rs35521052 Chr.19: 41411026-41411026 on Build GRCh38
rs916624011 Chr.19: 41410938-41410938 on Build GRCh38
rs749487379 Chr.19: 41410953-41410953 on Build GRCh38
rs990776009 Chr.19: 41411114-41411114 on Build GRCh38
rs753696309 Chr.19: 41411014-41411014 on Build GRCh38
rs200242482 Chr.19: 41410878-41410878 on Build GRCh38
rs748297608 Chr.19: 41410831-41410831 on Build GRCh38
rs768944277 Chr.19: 41410767-41410767 on Build GRCh38
rs552323788 Chr.19: 41411074-41411074 on Build GRCh38
rs746861446 Chr.19: 41410766-41410766 on Build GRCh38
rs761363859 Chr.19: 41410850-41410850 on Build GRCh38
rs760001681 Chr.19: 41410892-41410892 on Build GRCh38
rs764284154 Chr.19: 41410993-41410993 on Build GRCh38
rs573976272 Chr.19: 41410985-41410985 on Build GRCh38
rs778956980 Chr.19: 41410958-41410958 on Build GRCh38
rs745873859 Chr.19: 41411033-41411033 on Build GRCh38
rs774178264 Chr.19: 41411047-41411047 on Build GRCh38
rs369995436 Chr.19: 41410853-41410853 on Build GRCh38
rs750133814 Chr.19: 41410908-41410908 on Build GRCh38
rs922714716 Chr.19: 41410810-41410810 on Build GRCh38
rs369236118 Chr.19: 41410880-41410880 on Build GRCh38
rs398123498 Chr.19: 41410981-41410981 on Build GRCh38
rs748582297 Chr.19: 41411039-41411039 on Build GRCh38
rs753452195 Chr.19: 41410805-41410805 on Build GRCh38
rs747299996 Chr.19: 41410973-41410973 on Build GRCh38
rs777920549 Chr.19: 41410947-41410947 on Build GRCh38
rs577534319 Chr.19: 41410906-41410906 on Build GRCh38
rs904434121 Chr.19: 41410785-41410785 on Build GRCh38
rs748405960 Chr.19: 41410989-41410989 on Build GRCh38
rs1003232442 Chr.19: 41410911-41410911 on Build GRCh38
rs376204997 Chr.19: 41410922-41410922 on Build GRCh38
rs534878617 Chr.19: 41411096-41411096 on Build GRCh38
rs756052603 Chr.19: 41411011-41411011 on Build GRCh38
rs371286159 Chr.19: 41410854-41410854 on Build GRCh38
rs372195607 Chr.19: 41410804-41410804 on Build GRCh38
rs1005700237 Chr.19: 41411119-41411119 on Build GRCh38
rs766928512 Chr.19: 41410779-41410779 on Build GRCh38
rs780156666 Chr.19: 41410823-41410823 on Build GRCh38
rs770410589 Chr.19: 41411163-41411163 on Build GRCh38
rs1056149241 Chr.19: 41411113-41411113 on Build GRCh38
rs758919046 Chr.19: 41411054-41411054 on Build GRCh38
rs143167070 Chr.19: 41410768-41410768 on Build GRCh38
rs753142032 Chr.19: 41410849-41410849 on Build GRCh38
rs755016319 Chr.19: 41410861-41410861 on Build GRCh38
rs777414203 Chr.19: 41410730-41410730 on Build GRCh38
rs148571328 Chr.19: 41410816-41410816 on Build GRCh38
rs751571713 Chr.19: 41410859-41410859 on Build GRCh38
rs908547972 Chr.19: 41410751-41410751 on Build GRCh38
rs746587969 Chr.19: 41411106-41411106 on Build GRCh38
rs760654267 Chr.19: 41411060-41411060 on Build GRCh38
rs11549938 Chr.19: 41410772-41410772 on Build GRCh38
rs780819372 Chr.19: 41410862-41410862 on Build GRCh38
rs924064162 Chr.19: 41411152-41411152 on Build GRCh38
rs751862507 Chr.19: 41410789-41410789 on Build GRCh38
rs769625040 Chr.19: 41410778-41410778 on Build GRCh38
rs150700696 Chr.19: 41410970-41410970 on Build GRCh38
rs200757652 Chr.19: 41411034-41411034 on Build GRCh38
rs138025447 Chr.19: 41410741-41410741 on Build GRCh38
rs780656085 Chr.19: 41410887-41410888 on Build GRCh38
rs1023622940 Chr.19: 41411117-41411117 on Build GRCh38
rs377496852 Chr.19: 41411013-41411013 on Build GRCh38
rs398123499 Chr.19: 41411004-41411004 on Build GRCh38
rs771017948 Chr.19: 41410889-41410889 on Build GRCh38
rs748762939 Chr.19: 41410731-41410731 on Build GRCh38
rs767029689 Chr.19: 41411057-41411057 on Build GRCh38
rs11549937 Chr.19: 41410925-41410925 on Build GRCh38
rs376894084 Chr.19: 41410826-41410826 on Build GRCh38
rs770640951 Chr.19: 41411042-41411042 on Build GRCh38

View Full Product Details