Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41843844-41844387 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LYPD4
Entrez Gene ID
147719
Gene Name
LY6/PLAUR domain containing 4
Gene Aliases
SMR
UniGene
Hs.382361
Gene Chromosome Location
Chr.19: 41833686-41844771 on Build GRCh38
Transcripts
NM_173506.6 XM_005258543.3 XM_005278383.4 XM_005278385.4 XM_005278387.4 XM_005278388.3 XM_006723035.2

SNP Detail

SNP ID Chromosome Location
rs530401661 Chr.19: 41844267-41844267 on Build GRCh38
rs970870589 Chr.19: 41844164-41844164 on Build GRCh38
rs201372576 Chr.19: 41843906-41843906 on Build GRCh38
rs932307134 Chr.19: 41844308-41844308 on Build GRCh38
rs1003742123 Chr.19: 41844359-41844359 on Build GRCh38
rs914488614 Chr.19: 41844237-41844237 on Build GRCh38
rs538906368 Chr.19: 41844015-41844015 on Build GRCh38
rs777462411 Chr.19: 41844334-41844334 on Build GRCh38
rs782519227 Chr.19: 41844265-41844265 on Build GRCh38
rs564814572 Chr.19: 41844360-41844360 on Build GRCh38
rs989518740 Chr.19: 41844213-41844213 on Build GRCh38
rs959621259 Chr.19: 41844211-41844211 on Build GRCh38
rs899469409 Chr.19: 41844026-41844026 on Build GRCh38
rs977343903 Chr.19: 41844268-41844268 on Build GRCh38
rs1033780381 Chr.19: 41844196-41844196 on Build GRCh38
rs552627122 Chr.19: 41844336-41844336 on Build GRCh38
rs1015631042 Chr.19: 41843929-41843929 on Build GRCh38
rs924348287 Chr.19: 41843991-41843991 on Build GRCh38
rs188039263 Chr.19: 41844168-41844168 on Build GRCh38
rs947183660 Chr.19: 41843986-41843986 on Build GRCh38
rs996555520 Chr.19: 41844048-41844048 on Build GRCh38
rs914521064 Chr.19: 41843984-41843984 on Build GRCh38
rs575180261 Chr.19: 41843877-41843877 on Build GRCh38
rs906663642 Chr.19: 41844348-41844348 on Build GRCh38
rs1004380739 Chr.19: 41844317-41844317 on Build GRCh38
rs982938617 Chr.19: 41843910-41843910 on Build GRCh38
rs991784297 Chr.19: 41843971-41843971 on Build GRCh38
rs982224100 Chr.19: 41843935-41843935 on Build GRCh38
rs968656136 Chr.19: 41844246-41844246 on Build GRCh38
rs952870201 Chr.19: 41843907-41843907 on Build GRCh38
rs561584157 Chr.19: 41844298-41844298 on Build GRCh38
rs959417628 Chr.19: 41843957-41843957 on Build GRCh38
rs1015683163 Chr.19: 41844122-41844122 on Build GRCh38
rs556710402 Chr.19: 41844326-41844326 on Build GRCh38
rs139101061 Chr.19: 41843987-41843987 on Build GRCh38
rs1004332060 Chr.19: 41844100-41844100 on Build GRCh38
rs193246921 Chr.19: 41844309-41844309 on Build GRCh38
rs924416476 Chr.19: 41844269-41844269 on Build GRCh38
rs73549145 Chr.19: 41843876-41843876 on Build GRCh38
rs188477414 Chr.19: 41844327-41844327 on Build GRCh38
rs557920013 Chr.19: 41844105-41844105 on Build GRCh38
rs926574913 Chr.19: 41843951-41843951 on Build GRCh38
rs531205968 Chr.19: 41844331-41844331 on Build GRCh38
rs921047310 Chr.19: 41844290-41844290 on Build GRCh38
rs943746471 Chr.19: 41844010-41844010 on Build GRCh38
rs183626723 Chr.19: 41844310-41844310 on Build GRCh38
rs76812595 Chr.19: 41844282-41844282 on Build GRCh38
rs970821828 Chr.19: 41843932-41843932 on Build GRCh38
rs943899952 Chr.19: 41844272-41844272 on Build GRCh38
rs888740091 Chr.19: 41844067-41844067 on Build GRCh38
rs1048611287 Chr.19: 41844066-41844066 on Build GRCh38

View Full Product Details