Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41848840-41849333 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMRTC2
Entrez Gene ID
63946
Gene Name
DMRT like family C2
Gene Aliases
-
UniGene
Hs.350507
Gene Chromosome Location
Chr.19: 41844743-41852330 on Build GRCh38
Transcripts
NM_001040283.2 XM_011527214.1 XM_011527215.1 XM_011527216.2 XM_017027123.1 XM_017027124.1 XM_017027125.1 XM_017027126.1 XM_017027127.1 XM_017027128.1 XM_017027129.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4903081 c.694C>T p.H232Y Chr.19: 41849195-41849195 on Build GRCh38
5577406 c.712C>T p.P238S Chr.19: 41849213-41849213 on Build GRCh38
3534530 c.688G>A p.G230R Chr.19: 41849189-41849189 on Build GRCh38
3534529 c.654G>A p.Q218Q Chr.19: 41849155-41849155 on Build GRCh38
5529855 c.647C>T p.S216F Chr.19: 41849148-41849148 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs17853910 Chr.19: 41848898-41848898 on Build GRCh38
rs375271526 Chr.19: 41849145-41849145 on Build GRCh38
rs558894286 Chr.19: 41848920-41848920 on Build GRCh38
rs782270178 Chr.19: 41849237-41849237 on Build GRCh38
rs782314612 Chr.19: 41849233-41849233 on Build GRCh38
rs1055427732 Chr.19: 41849078-41849078 on Build GRCh38
rs782119789 Chr.19: 41849246-41849246 on Build GRCh38
rs782800557 Chr.19: 41849255-41849255 on Build GRCh38
rs782531854 Chr.19: 41849202-41849202 on Build GRCh38
rs183622708 Chr.19: 41849080-41849080 on Build GRCh38
rs782052285 Chr.19: 41848938-41848938 on Build GRCh38
rs782134710 Chr.19: 41849099-41849099 on Build GRCh38
rs782533304 Chr.19: 41849303-41849303 on Build GRCh38
rs371216175 Chr.19: 41849115-41849115 on Build GRCh38
rs782496035 Chr.19: 41848972-41848972 on Build GRCh38
rs781823672 Chr.19: 41849279-41849279 on Build GRCh38
rs189285681 Chr.19: 41849082-41849082 on Build GRCh38
rs782349470 Chr.19: 41849114-41849114 on Build GRCh38
rs781812164 Chr.19: 41849194-41849194 on Build GRCh38
rs371621401 Chr.19: 41848930-41848930 on Build GRCh38
rs1008438100 Chr.19: 41849152-41849152 on Build GRCh38
rs782741230 Chr.19: 41849191-41849191 on Build GRCh38
rs371867212 Chr.19: 41849122-41849122 on Build GRCh38
rs782531999 Chr.19: 41849215-41849215 on Build GRCh38
rs782747662 Chr.19: 41849170-41849170 on Build GRCh38
rs782482650 Chr.19: 41849284-41849284 on Build GRCh38
rs782022689 Chr.19: 41848949-41848949 on Build GRCh38
rs374220011 Chr.19: 41848887-41848887 on Build GRCh38
rs782698481 Chr.19: 41849203-41849203 on Build GRCh38
rs782237730 Chr.19: 41849044-41849044 on Build GRCh38
rs782740475 Chr.19: 41849251-41849251 on Build GRCh38
rs782248330 Chr.19: 41849205-41849205 on Build GRCh38
rs201302617 Chr.19: 41848990-41848990 on Build GRCh38
rs782724452 Chr.19: 41849138-41849138 on Build GRCh38
rs919904494 Chr.19: 41849282-41849282 on Build GRCh38
rs200990664 Chr.19: 41848989-41848989 on Build GRCh38
rs782772098 Chr.19: 41849273-41849273 on Build GRCh38
rs940559212 Chr.19: 41848961-41848961 on Build GRCh38
rs782519303 Chr.19: 41849263-41849263 on Build GRCh38
rs782208407 Chr.19: 41848984-41848984 on Build GRCh38
rs368632681 Chr.19: 41849236-41849236 on Build GRCh38
rs782282221 Chr.19: 41849021-41849021 on Build GRCh38
rs782464529 Chr.19: 41849187-41849187 on Build GRCh38
rs374622487 Chr.19: 41848951-41848951 on Build GRCh38
rs148724244 Chr.19: 41848931-41848931 on Build GRCh38
rs782146252 Chr.19: 41849213-41849213 on Build GRCh38
rs782561862 Chr.19: 41849301-41849302 on Build GRCh38
rs781933990 Chr.19: 41849234-41849234 on Build GRCh38
rs151277709 Chr.19: 41848954-41848954 on Build GRCh38
rs782682284 Chr.19: 41849307-41849307 on Build GRCh38
rs906983346 Chr.19: 41848997-41848997 on Build GRCh38
rs907686497 Chr.19: 41848900-41848900 on Build GRCh38
rs200152667 Chr.19: 41848980-41848980 on Build GRCh38
rs145179597 Chr.19: 41848883-41848883 on Build GRCh38
rs781959531 Chr.19: 41849252-41849252 on Build GRCh38
rs367552578 Chr.19: 41848968-41848968 on Build GRCh38
rs782580801 Chr.19: 41849018-41849018 on Build GRCh38
rs782049925 Chr.19: 41849121-41849121 on Build GRCh38
rs140255599 Chr.19: 41849230-41849230 on Build GRCh38
rs782197429 Chr.19: 41849300-41849300 on Build GRCh38
rs781913731 Chr.19: 41849256-41849256 on Build GRCh38
rs782602456 Chr.19: 41849227-41849227 on Build GRCh38
rs782495210 Chr.19: 41848878-41848878 on Build GRCh38
rs781866567 Chr.19: 41849179-41849179 on Build GRCh38
rs374714026 Chr.19: 41849188-41849188 on Build GRCh38
rs377724016 Chr.19: 41848923-41848923 on Build GRCh38
rs782329023 Chr.19: 41848948-41848948 on Build GRCh38
rs782571567 Chr.19: 41849294-41849294 on Build GRCh38
rs782677269 Chr.19: 41848896-41848896 on Build GRCh38
rs782073409 Chr.19: 41849253-41849253 on Build GRCh38
rs535043320 Chr.19: 41849003-41849003 on Build GRCh38
rs782159590 Chr.19: 41849166-41849166 on Build GRCh38
rs782405247 Chr.19: 41848894-41848894 on Build GRCh38
rs782414628 Chr.19: 41849242-41849242 on Build GRCh38
rs782088416 Chr.19: 41848958-41848958 on Build GRCh38
rs1019650435 Chr.19: 41849243-41849243 on Build GRCh38

View Full Product Details