Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 43197907-43198420 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSG4
Entrez Gene ID
5672
Gene Name
pregnancy specific beta-1-glycoprotein 4
Gene Aliases
PSBG-4, PSBG-9, PSG9
UniGene
Hs.711363
Gene Chromosome Location
Chr.19: 43192702-43205774 on Build GRCh38
Transcripts
NM_001276495.1 NM_001316339.1 NM_002780.4 NM_213633.2 XM_017026995.1 XM_017026996.1 XM_017026997.1 XM_017026998.1 XM_017026999.1 XM_017027000.1 XM_017027001.1 XM_017027002.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5032970 c.551G>T p.G184V Chr.19: 43198155-43198155 on Build GRCh38
2149769 c.437C>G p.T146S Chr.19: 43198269-43198269 on Build GRCh38
4901651 c.637G>A p.G213R Chr.19: 43198069-43198069 on Build GRCh38
3736632 c.455C>T p.S152F Chr.19: 43198251-43198251 on Build GRCh38
6151638 c.671G>A p.S224N Chr.19: 43198035-43198035 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774435026 Chr.19: 43198193-43198193 on Build GRCh38
rs748671467 Chr.19: 43198296-43198296 on Build GRCh38
rs748411143 Chr.19: 43198339-43198339 on Build GRCh38
rs376900449 Chr.19: 43198003-43198003 on Build GRCh38
rs139754497 Chr.19: 43198345-43198345 on Build GRCh38
rs371257082 Chr.19: 43198311-43198311 on Build GRCh38
rs369022798 Chr.19: 43197982-43197982 on Build GRCh38
rs751386330 Chr.19: 43198174-43198174 on Build GRCh38
rs759940767 Chr.19: 43198186-43198186 on Build GRCh38
rs766489546 Chr.19: 43198314-43198314 on Build GRCh38
rs753211051 Chr.19: 43198139-43198139 on Build GRCh38
rs772269350 Chr.19: 43198346-43198346 on Build GRCh38
rs200436261 Chr.19: 43197931-43197931 on Build GRCh38
rs145207742 Chr.19: 43198105-43198105 on Build GRCh38
rs531158313 Chr.19: 43198365-43198365 on Build GRCh38
rs561170850 Chr.19: 43198018-43198018 on Build GRCh38
rs769167488 Chr.19: 43198233-43198233 on Build GRCh38
rs770631123 Chr.19: 43198216-43198216 on Build GRCh38
rs375824745 Chr.19: 43197969-43197969 on Build GRCh38
rs769045052 Chr.19: 43198276-43198276 on Build GRCh38
rs774859357 Chr.19: 43198120-43198120 on Build GRCh38
rs186019748 Chr.19: 43198224-43198224 on Build GRCh38
rs775375742 Chr.19: 43198015-43198015 on Build GRCh38
rs73548062 Chr.19: 43198226-43198226 on Build GRCh38
rs748880731 Chr.19: 43198181-43198181 on Build GRCh38
rs775950740 Chr.19: 43198103-43198103 on Build GRCh38
rs766807245 Chr.19: 43198119-43198119 on Build GRCh38
rs80312463 Chr.19: 43198335-43198335 on Build GRCh38
rs758168710 Chr.19: 43198086-43198086 on Build GRCh38
rs200687668 Chr.19: 43198245-43198245 on Build GRCh38
rs568862956 Chr.19: 43198342-43198342 on Build GRCh38
rs562024727 Chr.19: 43198349-43198349 on Build GRCh38
rs775544544 Chr.19: 43198268-43198281 on Build GRCh38
rs750605503 Chr.19: 43198244-43198244 on Build GRCh38
rs11428975 Chr.19: 43197946-43197947 on Build GRCh38
rs546817701 Chr.19: 43197965-43197965 on Build GRCh38
rs146166080 Chr.19: 43198011-43198011 on Build GRCh38
rs559284599 Chr.19: 43198189-43198189 on Build GRCh38
rs532667133 Chr.19: 43198074-43198074 on Build GRCh38
rs576794839 Chr.19: 43198351-43198351 on Build GRCh38
rs199766455 Chr.19: 43198184-43198184 on Build GRCh38
rs761551644 Chr.19: 43197973-43197973 on Build GRCh38
rs762878954 Chr.19: 43198302-43198302 on Build GRCh38
rs779647600 Chr.19: 43198254-43198254 on Build GRCh38
rs776044309 Chr.19: 43198187-43198187 on Build GRCh38
rs73548061 Chr.19: 43198175-43198175 on Build GRCh38
rs377671105 Chr.19: 43198304-43198304 on Build GRCh38
rs571960356 Chr.19: 43198210-43198210 on Build GRCh38
rs773060457 Chr.19: 43198034-43198034 on Build GRCh38
rs546236237 Chr.19: 43198261-43198261 on Build GRCh38
rs758921542 Chr.19: 43197987-43197987 on Build GRCh38
rs138406653 Chr.19: 43198257-43198257 on Build GRCh38
rs1038286761 Chr.19: 43198370-43198370 on Build GRCh38
rs749552851 Chr.19: 43198084-43198084 on Build GRCh38
rs752835228 Chr.19: 43198066-43198066 on Build GRCh38
rs78731124 Chr.19: 43198256-43198256 on Build GRCh38
rs759080487 Chr.19: 43197974-43197981 on Build GRCh38
rs140068216 Chr.19: 43198183-43198183 on Build GRCh38
rs781352640 Chr.19: 43198000-43198000 on Build GRCh38
rs764208733 Chr.19: 43198160-43198160 on Build GRCh38
rs757503957 Chr.19: 43198168-43198168 on Build GRCh38
rs754012240 Chr.19: 43198165-43198165 on Build GRCh38
rs576123327 Chr.19: 43197948-43197948 on Build GRCh38
rs137963702 Chr.19: 43198235-43198235 on Build GRCh38
rs925112378 Chr.19: 43198322-43198322 on Build GRCh38
rs777453397 Chr.19: 43198180-43198180 on Build GRCh38
rs145753670 Chr.19: 43198047-43198047 on Build GRCh38
rs777114922 Chr.19: 43198286-43198286 on Build GRCh38
rs143008288 Chr.19: 43198268-43198268 on Build GRCh38
rs756814499 Chr.19: 43198262-43198262 on Build GRCh38
rs146130390 Chr.19: 43198170-43198170 on Build GRCh38
rs772009289 Chr.19: 43197950-43197950 on Build GRCh38
rs373225896 Chr.19: 43197990-43197990 on Build GRCh38
rs189261972 Chr.19: 43198374-43198374 on Build GRCh38
rs778299521 Chr.19: 43198265-43198265 on Build GRCh38
rs528483691 Chr.19: 43198129-43198129 on Build GRCh38
rs373053254 Chr.19: 43198196-43198196 on Build GRCh38
rs756187783 Chr.19: 43198141-43198141 on Build GRCh38
rs150736166 Chr.19: 43198026-43198026 on Build GRCh38
rs754776543 Chr.19: 43198194-43198194 on Build GRCh38
rs199534289 Chr.19: 43198048-43198048 on Build GRCh38
rs765482477 Chr.19: 43197977-43197977 on Build GRCh38
rs755537243 Chr.19: 43198054-43198054 on Build GRCh38
rs775465333 Chr.19: 43197957-43197957 on Build GRCh38
rs879699450 Chr.19: 43198247-43198247 on Build GRCh38
rs570985384 Chr.19: 43198382-43198382 on Build GRCh38
rs776238651 Chr.19: 43198027-43198027 on Build GRCh38
rs758016420 Chr.19: 43198271-43198271 on Build GRCh38
rs769849561 Chr.19: 43198298-43198298 on Build GRCh38
rs563613270 Chr.19: 43198062-43198062 on Build GRCh38
rs761880287 Chr.19: 43198241-43198241 on Build GRCh38
rs374110942 Chr.19: 43198191-43198191 on Build GRCh38
rs748277035 Chr.19: 43198001-43198001 on Build GRCh38
rs9789333 Chr.19: 43198190-43198190 on Build GRCh38
rs778620146 Chr.19: 43198004-43198004 on Build GRCh38
rs763374737 Chr.19: 43198043-43198043 on Build GRCh38
rs373538412 Chr.19: 43198320-43198320 on Build GRCh38
rs765321336 Chr.19: 43198248-43198248 on Build GRCh38
rs543619125 Chr.19: 43198243-43198243 on Build GRCh38
rs993998473 Chr.19: 43198377-43198377 on Build GRCh38
rs747350141 Chr.19: 43198273-43198273 on Build GRCh38
rs768646719 Chr.19: 43198096-43198096 on Build GRCh38
rs555809394 Chr.19: 43197947-43197947 on Build GRCh38
rs767272807 Chr.19: 43198251-43198251 on Build GRCh38
rs746883332 Chr.19: 43198092-43198092 on Build GRCh38
rs769377274 Chr.19: 43198179-43198179 on Build GRCh38
rs768054043 Chr.19: 43198130-43198130 on Build GRCh38
rs764039363 Chr.19: 43198324-43198324 on Build GRCh38
rs754987366 Chr.19: 43197997-43197997 on Build GRCh38
rs761105504 Chr.19: 43198232-43198232 on Build GRCh38
rs899294242 Chr.19: 43198371-43198371 on Build GRCh38
rs139058318 Chr.19: 43198032-43198032 on Build GRCh38
rs752149380 Chr.19: 43197996-43197996 on Build GRCh38
rs552283703 Chr.19: 43198104-43198104 on Build GRCh38
rs538767331 Chr.19: 43198202-43198202 on Build GRCh38
rs147615358 Chr.19: 43198115-43198115 on Build GRCh38
rs772974266 Chr.19: 43198107-43198107 on Build GRCh38
rs748081092 Chr.19: 43198176-43198176 on Build GRCh38
rs756206359 Chr.19: 43198067-43198067 on Build GRCh38
rs777358008 Chr.19: 43198208-43198208 on Build GRCh38
rs761308655 Chr.19: 43198028-43198028 on Build GRCh38
rs541286633 Chr.19: 43198016-43198016 on Build GRCh38
rs143978380 Chr.19: 43198203-43198203 on Build GRCh38
rs761193521 Chr.19: 43198188-43198188 on Build GRCh38
rs148538453 Chr.19: 43198089-43198089 on Build GRCh38
rs763701035 Chr.19: 43197959-43197959 on Build GRCh38
rs770581007 Chr.19: 43197946-43197946 on Build GRCh38
rs1052150463 Chr.19: 43198331-43198331 on Build GRCh38

View Full Product Details