Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 43533704-43534211 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF575
Entrez Gene ID
284346
Gene Name
zinc finger protein 575
Gene Aliases
-
UniGene
Hs.213534
Gene Chromosome Location
Chr.19: 43531805-43536138 on Build GRCh38
Transcripts
NM_174945.2 XM_005258783.3 XM_011526793.2

SNP Detail

SNP ID Chromosome Location
rs1033366566 Chr.19: 43533809-43533809 on Build GRCh38
rs2256507 Chr.19: 43534154-43534154 on Build GRCh38
rs1034201061 Chr.19: 43534136-43534136 on Build GRCh38
rs998409590 Chr.19: 43533835-43533835 on Build GRCh38
rs950909178 Chr.19: 43533942-43533942 on Build GRCh38
rs901036049 Chr.19: 43533816-43533816 on Build GRCh38
rs935846411 Chr.19: 43534049-43534049 on Build GRCh38
rs770887333 Chr.19: 43534181-43534181 on Build GRCh38
rs911598495 Chr.19: 43534007-43534007 on Build GRCh38
rs916045052 Chr.19: 43534063-43534063 on Build GRCh38
rs915657224 Chr.19: 43533757-43533757 on Build GRCh38
rs570287534 Chr.19: 43533989-43533989 on Build GRCh38
rs573931468 Chr.19: 43533925-43533925 on Build GRCh38
rs1007616944 Chr.19: 43533944-43533944 on Build GRCh38
rs963686951 Chr.19: 43533988-43533988 on Build GRCh38
rs1003529461 Chr.19: 43534133-43534133 on Build GRCh38
rs529034630 Chr.19: 43533753-43533753 on Build GRCh38
rs557924228 Chr.19: 43534040-43534040 on Build GRCh38
rs1033758915 Chr.19: 43533874-43533874 on Build GRCh38
rs146940698 Chr.19: 43534054-43534054 on Build GRCh38
rs539192650 Chr.19: 43534011-43534011 on Build GRCh38
rs567978983 Chr.19: 43533921-43533921 on Build GRCh38
rs536531959 Chr.19: 43533937-43533937 on Build GRCh38
rs553471761 Chr.19: 43534142-43534142 on Build GRCh38
rs959453532 Chr.19: 43534141-43534142 on Build GRCh38
rs554154733 Chr.19: 43534106-43534106 on Build GRCh38
rs368817980 Chr.19: 43533815-43533815 on Build GRCh38
rs950322231 Chr.19: 43534085-43534106 on Build GRCh38
rs1045617252 Chr.19: 43534101-43534101 on Build GRCh38
rs948424261 Chr.19: 43533769-43533769 on Build GRCh38
rs921683669 Chr.19: 43534044-43534044 on Build GRCh38
rs960272315 Chr.19: 43534045-43534045 on Build GRCh38
rs912134294 Chr.19: 43534184-43534184 on Build GRCh38
rs944378789 Chr.19: 43534012-43534012 on Build GRCh38
rs948474486 Chr.19: 43534068-43534068 on Build GRCh38
rs904156889 Chr.19: 43533789-43533789 on Build GRCh38
rs934612846 Chr.19: 43533797-43533797 on Build GRCh38
rs376402949 Chr.19: 43534128-43534128 on Build GRCh38
rs1030460301 Chr.19: 43533858-43533858 on Build GRCh38
rs757557391 Chr.19: 43533932-43533933 on Build GRCh38
rs1027783230 Chr.19: 43534146-43534146 on Build GRCh38
rs1004071666 Chr.19: 43533855-43533855 on Build GRCh38
rs572012058 Chr.19: 43533941-43533941 on Build GRCh38
rs1010149134 Chr.19: 43534092-43534092 on Build GRCh38
rs147756835 Chr.19: 43533825-43533825 on Build GRCh38
rs918942256 Chr.19: 43533926-43533927 on Build GRCh38
rs550464019 Chr.19: 43533964-43533964 on Build GRCh38
rs983736781 Chr.19: 43534172-43534172 on Build GRCh38
rs374445473 Chr.19: 43533772-43533772 on Build GRCh38
rs1026962930 Chr.19: 43534010-43534010 on Build GRCh38
rs774450798 Chr.19: 43533969-43533969 on Build GRCh38
rs978457627 Chr.19: 43534100-43534100 on Build GRCh38
rs1039223926 Chr.19: 43534032-43534032 on Build GRCh38
rs970702073 Chr.19: 43533838-43533838 on Build GRCh38

View Full Product Details