Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 43719132-43719661 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRGC
Entrez Gene ID
56269
Gene Name
immunity related GTPase cinema
Gene Aliases
CINEMA, IFGGE, IRGC1, Iigp5, R30953_1
UniGene
Hs.515444
Gene Chromosome Location
Chr.19: 43716062-43720021 on Build GRCh38
Transcripts
NM_019612.3 XM_005259058.3 XM_006723283.3 XM_011527118.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5532813 c.692C>T p.T231I Chr.19: 43719250-43719250 on Build GRCh38
1394310 c.1053C>T p.S351S Chr.19: 43719611-43719611 on Build GRCh38
4693056 c.908G>A p.R303H Chr.19: 43719466-43719466 on Build GRCh38
6151600 c.1076G>T p.R359L Chr.19: 43719634-43719634 on Build GRCh38
4693054 c.693C>T p.T231T Chr.19: 43719251-43719251 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780295900 Chr.19: 43719490-43719490 on Build GRCh38
rs757989962 Chr.19: 43719537-43719537 on Build GRCh38
rs754820557 Chr.19: 43719351-43719351 on Build GRCh38
rs780678651 Chr.19: 43719413-43719413 on Build GRCh38
rs565345774 Chr.19: 43719482-43719482 on Build GRCh38
rs780870515 Chr.19: 43719230-43719230 on Build GRCh38
rs773511810 Chr.19: 43719214-43719214 on Build GRCh38
rs142253899 Chr.19: 43719533-43719533 on Build GRCh38
rs754495869 Chr.19: 43719189-43719189 on Build GRCh38
rs768121065 Chr.19: 43719615-43719615 on Build GRCh38
rs778782722 Chr.19: 43719170-43719170 on Build GRCh38
rs769872744 Chr.19: 43719506-43719506 on Build GRCh38
rs749644485 Chr.19: 43719277-43719277 on Build GRCh38
rs199678200 Chr.19: 43719173-43719173 on Build GRCh38
rs771083825 Chr.19: 43719573-43719573 on Build GRCh38
rs773546328 Chr.19: 43719507-43719507 on Build GRCh38
rs777050741 Chr.19: 43719306-43719306 on Build GRCh38
rs755683247 Chr.19: 43719383-43719383 on Build GRCh38
rs926507114 Chr.19: 43719368-43719368 on Build GRCh38
rs187509627 Chr.19: 43719491-43719491 on Build GRCh38
rs760614681 Chr.19: 43719464-43719464 on Build GRCh38
rs141857999 Chr.19: 43719632-43719632 on Build GRCh38
rs766020551 Chr.19: 43719344-43719344 on Build GRCh38
rs758855988 Chr.19: 43719486-43719486 on Build GRCh38
rs371517810 Chr.19: 43719513-43719513 on Build GRCh38
rs761670484 Chr.19: 43719588-43719588 on Build GRCh38
rs939202107 Chr.19: 43719636-43719636 on Build GRCh38
rs376749873 Chr.19: 43719575-43719575 on Build GRCh38
rs773838678 Chr.19: 43719341-43719341 on Build GRCh38
rs747072003 Chr.19: 43719416-43719416 on Build GRCh38
rs762085853 Chr.19: 43719420-43719420 on Build GRCh38
rs375528390 Chr.19: 43719538-43719538 on Build GRCh38
rs1041595614 Chr.19: 43719451-43719451 on Build GRCh38
rs746497854 Chr.19: 43719182-43719182 on Build GRCh38
rs369246997 Chr.19: 43719567-43719567 on Build GRCh38
rs202106053 Chr.19: 43719179-43719179 on Build GRCh38
rs766182531 Chr.19: 43719609-43719609 on Build GRCh38
rs543218904 Chr.19: 43719611-43719611 on Build GRCh38
rs199599515 Chr.19: 43719485-43719485 on Build GRCh38
rs777863585 Chr.19: 43719625-43719625 on Build GRCh38
rs957717217 Chr.19: 43719279-43719279 on Build GRCh38
rs752113357 Chr.19: 43719357-43719357 on Build GRCh38
rs756134216 Chr.19: 43719546-43719546 on Build GRCh38
rs766889301 Chr.19: 43719158-43719158 on Build GRCh38
rs748285751 Chr.19: 43719503-43719503 on Build GRCh38
rs747713451 Chr.19: 43719261-43719261 on Build GRCh38
rs199882680 Chr.19: 43719607-43719607 on Build GRCh38
rs369487767 Chr.19: 43719218-43719218 on Build GRCh38
rs750213262 Chr.19: 43719229-43719229 on Build GRCh38
rs370990305 Chr.19: 43719587-43719587 on Build GRCh38
rs761525743 Chr.19: 43719618-43719618 on Build GRCh38
rs775641874 Chr.19: 43719226-43719226 on Build GRCh38
rs546481895 Chr.19: 43719550-43719550 on Build GRCh38
rs376496268 Chr.19: 43719617-43719617 on Build GRCh38
rs557407959 Chr.19: 43719243-43719243 on Build GRCh38
rs201350493 Chr.19: 43719263-43719263 on Build GRCh38
rs184064504 Chr.19: 43719453-43719453 on Build GRCh38
rs201293395 Chr.19: 43719401-43719401 on Build GRCh38
rs758334834 Chr.19: 43719238-43719238 on Build GRCh38
rs752810297 Chr.19: 43719165-43719165 on Build GRCh38
rs760760691 Chr.19: 43719227-43719227 on Build GRCh38
rs141330617 Chr.19: 43719454-43719454 on Build GRCh38
rs34471331 Chr.19: 43719260-43719260 on Build GRCh38
rs553364902 Chr.19: 43719566-43719566 on Build GRCh38
rs781370552 Chr.19: 43719502-43719502 on Build GRCh38
rs772428142 Chr.19: 43719412-43719412 on Build GRCh38
rs747716884 Chr.19: 43719208-43719208 on Build GRCh38
rs778557453 Chr.19: 43719624-43719624 on Build GRCh38
rs374269776 Chr.19: 43719608-43719608 on Build GRCh38
rs138990135 Chr.19: 43719529-43719529 on Build GRCh38
rs372886723 Chr.19: 43719209-43719209 on Build GRCh38
rs759571667 Chr.19: 43719160-43719160 on Build GRCh38
rs763558549 Chr.19: 43719466-43719466 on Build GRCh38
rs145563243 Chr.19: 43719570-43719570 on Build GRCh38
rs73935064 Chr.19: 43719569-43719569 on Build GRCh38
rs751400292 Chr.19: 43719350-43719350 on Build GRCh38
rs751629551 Chr.19: 43719159-43719159 on Build GRCh38
rs774786720 Chr.19: 43719285-43719285 on Build GRCh38
rs201718846 Chr.19: 43719302-43719302 on Build GRCh38
rs766405490 Chr.19: 43719239-43719239 on Build GRCh38
rs367855155 Chr.19: 43719400-43719400 on Build GRCh38
rs906432646 Chr.19: 43719518-43719518 on Build GRCh38
rs767536356 Chr.19: 43719353-43719353 on Build GRCh38
rs768797167 Chr.19: 43719418-43719418 on Build GRCh38
rs756387844 Chr.19: 43719166-43719166 on Build GRCh38
rs767453078 Chr.19: 43719223-43719223 on Build GRCh38
rs772657761 Chr.19: 43719299-43719299 on Build GRCh38
rs529910557 Chr.19: 43719426-43719426 on Build GRCh38
rs549975409 Chr.19: 43719434-43719434 on Build GRCh38
rs542350356 Chr.19: 43719395-43719395 on Build GRCh38
rs764784153 Chr.19: 43719532-43719532 on Build GRCh38
rs201085444 Chr.19: 43719469-43719469 on Build GRCh38
rs1003443833 Chr.19: 43719375-43719375 on Build GRCh38
rs760877732 Chr.19: 43719300-43719300 on Build GRCh38
rs924799320 Chr.19: 43719212-43719212 on Build GRCh38
rs369104782 Chr.19: 43719155-43719155 on Build GRCh38
rs762139049 Chr.19: 43719342-43719342 on Build GRCh38
rs906394054 Chr.19: 43719310-43719310 on Build GRCh38
rs376050824 Chr.19: 43719210-43719210 on Build GRCh38
rs560626765 Chr.19: 43719614-43719614 on Build GRCh38
rs34432569 Chr.19: 43719542-43719542 on Build GRCh38
rs776854343 Chr.19: 43719419-43719419 on Build GRCh38
rs757440735 Chr.19: 43719635-43719635 on Build GRCh38
rs201214577 Chr.19: 43719574-43719574 on Build GRCh38
rs544043530 Chr.19: 43719174-43719174 on Build GRCh38
rs753316685 Chr.19: 43719526-43719526 on Build GRCh38
rs746848166 Chr.19: 43719583-43719583 on Build GRCh38
rs200430483 Chr.19: 43719465-43719465 on Build GRCh38
rs574684743 Chr.19: 43719172-43719172 on Build GRCh38
rs749380064 Chr.19: 43719629-43719629 on Build GRCh38
rs866180953 Chr.19: 43719153-43719153 on Build GRCh38
rs147576016 Chr.19: 43719384-43719384 on Build GRCh38
rs977049152 Chr.19: 43719604-43719604 on Build GRCh38
rs756086638 Chr.19: 43719303-43719303 on Build GRCh38
rs562203488 Chr.19: 43719409-43719409 on Build GRCh38
rs749714322 Chr.19: 43719217-43719217 on Build GRCh38
rs199780198 Chr.19: 43719298-43719298 on Build GRCh38
rs771836583 Chr.19: 43719511-43719511 on Build GRCh38
rs1041254422 Chr.19: 43719234-43719234 on Build GRCh38
rs764738059 Chr.19: 43719601-43719601 on Build GRCh38
rs368626038 Chr.19: 43719425-43719425 on Build GRCh38
rs760108301 Chr.19: 43719222-43719222 on Build GRCh38
rs34051422 Chr.19: 43719515-43719515 on Build GRCh38
rs371254678 Chr.19: 43719509-43719509 on Build GRCh38
rs771230618 Chr.19: 43719280-43719280 on Build GRCh38
rs764460673 Chr.19: 43719228-43719228 on Build GRCh38
rs778669792 Chr.19: 43719483-43719483 on Build GRCh38
rs757018591 Chr.19: 43719190-43719190 on Build GRCh38
rs754945473 Chr.19: 43719257-43719257 on Build GRCh38
rs777437733 Chr.19: 43719267-43719267 on Build GRCh38
rs764069377 Chr.19: 43719525-43719525 on Build GRCh38
rs1053479385 Chr.19: 43719496-43719496 on Build GRCh38

View Full Product Details