Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 43820458-43820902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LYPD5
Entrez Gene ID
284348
Gene Name
LY6/PLAUR domain containing 5
Gene Aliases
PRO4356
UniGene
Hs.44289
Gene Chromosome Location
Chr.19: 43794835-43827199 on Build GRCh38
Transcripts
NM_182573.2 XM_011526794.2

SNP Detail

SNP ID Chromosome Location
rs970870812 Chr.19: 43820602-43820602 on Build GRCh38
rs574176009 Chr.19: 43820572-43820572 on Build GRCh38
rs893419026 Chr.19: 43820636-43820636 on Build GRCh38
rs1050657204 Chr.19: 43820632-43820632 on Build GRCh38
rs945467308 Chr.19: 43820491-43820491 on Build GRCh38
rs940759076 Chr.19: 43820581-43820581 on Build GRCh38
rs561092224 Chr.19: 43820791-43820791 on Build GRCh38
rs911956764 Chr.19: 43820689-43820689 on Build GRCh38
rs953343656 Chr.19: 43820698-43820698 on Build GRCh38
rs958618055 Chr.19: 43820525-43820525 on Build GRCh38
rs560570860 Chr.19: 43820508-43820508 on Build GRCh38
rs924801490 Chr.19: 43820749-43820749 on Build GRCh38
rs10421636 Chr.19: 43820665-43820665 on Build GRCh38
rs908391723 Chr.19: 43820571-43820571 on Build GRCh38
rs73558515 Chr.19: 43820654-43820654 on Build GRCh38
rs1002592739 Chr.19: 43820647-43820647 on Build GRCh38
rs535361403 Chr.19: 43820788-43820788 on Build GRCh38
rs868361327 Chr.19: 43820591-43820591 on Build GRCh38
rs947517453 Chr.19: 43820637-43820637 on Build GRCh38
rs1004467609 Chr.19: 43820785-43820785 on Build GRCh38
rs888123797 Chr.19: 43820796-43820796 on Build GRCh38
rs774648418 Chr.19: 43820703-43820703 on Build GRCh38
rs192898864 Chr.19: 43820574-43820574 on Build GRCh38
rs976188081 Chr.19: 43820670-43820670 on Build GRCh38
rs1034284657 Chr.19: 43820821-43820821 on Build GRCh38
rs950386541 Chr.19: 43820575-43820575 on Build GRCh38
rs533858494 Chr.19: 43820555-43820555 on Build GRCh38
rs1004096895 Chr.19: 43820808-43820808 on Build GRCh38
rs773315065 Chr.19: 43820795-43820795 on Build GRCh38
rs991363316 Chr.19: 43820533-43820533 on Build GRCh38
rs79007440 Chr.19: 43820663-43820663 on Build GRCh38
rs116858228 Chr.19: 43820869-43820869 on Build GRCh38
rs537852315 Chr.19: 43820850-43820850 on Build GRCh38
rs796109694 Chr.19: 43820544-43820544 on Build GRCh38
rs923383668 Chr.19: 43820697-43820697 on Build GRCh38
rs569085834 Chr.19: 43820565-43820565 on Build GRCh38
rs1041865650 Chr.19: 43820638-43820638 on Build GRCh38
rs545355172 Chr.19: 43820656-43820656 on Build GRCh38
rs1042492335 Chr.19: 43820492-43820492 on Build GRCh38
rs552288088 Chr.19: 43820717-43820717 on Build GRCh38
rs756787139 Chr.19: 43820652-43820652 on Build GRCh38
rs1023976099 Chr.19: 43820595-43820595 on Build GRCh38
rs1039308567 Chr.19: 43820746-43820746 on Build GRCh38
rs926906396 Chr.19: 43820651-43820651 on Build GRCh38
rs995203586 Chr.19: 43820753-43820753 on Build GRCh38
rs571292733 Chr.19: 43820550-43820550 on Build GRCh38
rs903362580 Chr.19: 43820640-43820640 on Build GRCh38
rs914593584 Chr.19: 43820705-43820705 on Build GRCh38
rs938883219 Chr.19: 43820752-43820752 on Build GRCh38
rs1048966733 Chr.19: 43820756-43820756 on Build GRCh38
rs932007153 Chr.19: 43820627-43820627 on Build GRCh38
rs890326047 Chr.19: 43820515-43820515 on Build GRCh38
rs1022519445 Chr.19: 43820876-43820876 on Build GRCh38
rs557808317 Chr.19: 43820487-43820487 on Build GRCh38
rs994751937 Chr.19: 43820573-43820573 on Build GRCh38
rs944536915 Chr.19: 43820733-43820733 on Build GRCh38
rs982219736 Chr.19: 43820619-43820619 on Build GRCh38
rs553952332 Chr.19: 43820563-43820563 on Build GRCh38
rs894253256 Chr.19: 43820769-43820769 on Build GRCh38
rs1056871223 Chr.19: 43820768-43820768 on Build GRCh38
rs748677098 Chr.19: 43820741-43820741 on Build GRCh38
rs976583261 Chr.19: 43820597-43820597 on Build GRCh38
rs575823389 Chr.19: 43820700-43820710 on Build GRCh38
rs886115091 Chr.19: 43820778-43820778 on Build GRCh38
rs900315132 Chr.19: 43820748-43820748 on Build GRCh38
rs556778829 Chr.19: 43820604-43820604 on Build GRCh38

View Full Product Details