Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 43951061-43951559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF221
Entrez Gene ID
7638
Gene Name
zinc finger protein 221
Gene Aliases
-
UniGene
Hs.631598
Gene Chromosome Location
Chr.19: 43951196-43970207 on Build GRCh38
Transcripts
NM_001297588.1 NM_001297589.1 NM_013359.2 XM_017027232.1 XM_017027233.1

SNP Detail

SNP ID Chromosome Location
rs542616295 Chr.19: 43951404-43951404 on Build GRCh38
rs746859686 Chr.19: 43951217-43951217 on Build GRCh38
rs372595801 Chr.19: 43951375-43951375 on Build GRCh38
rs758069473 Chr.19: 43951103-43951103 on Build GRCh38
rs967312188 Chr.19: 43951144-43951144 on Build GRCh38
rs907224360 Chr.19: 43951327-43951327 on Build GRCh38
rs573664518 Chr.19: 43951360-43951360 on Build GRCh38
rs144522324 Chr.19: 43951132-43951132 on Build GRCh38
rs879668578 Chr.19: 43951145-43951145 on Build GRCh38
rs1004982496 Chr.19: 43951439-43951439 on Build GRCh38
rs73038529 Chr.19: 43951110-43951110 on Build GRCh38
rs1046379190 Chr.19: 43951291-43951291 on Build GRCh38
rs902974865 Chr.19: 43951481-43951481 on Build GRCh38
rs569787653 Chr.19: 43951167-43951167 on Build GRCh38
rs146174739 Chr.19: 43951445-43951445 on Build GRCh38
rs375680420 Chr.19: 43951427-43951427 on Build GRCh38
rs144598041 Chr.19: 43951531-43951531 on Build GRCh38
rs577809977 Chr.19: 43951240-43951240 on Build GRCh38
rs563726957 Chr.19: 43951215-43951215 on Build GRCh38
rs184806962 Chr.19: 43951239-43951239 on Build GRCh38
rs906879752 Chr.19: 43951288-43951288 on Build GRCh38
rs770750348 Chr.19: 43951227-43951227 on Build GRCh38
rs1019279098 Chr.19: 43951177-43951177 on Build GRCh38
rs983925561 Chr.19: 43951202-43951202 on Build GRCh38
rs527911839 Chr.19: 43951514-43951514 on Build GRCh38
rs913400789 Chr.19: 43951276-43951276 on Build GRCh38
rs549167749 Chr.19: 43951513-43951513 on Build GRCh38
rs975042507 Chr.19: 43951225-43951225 on Build GRCh38
rs549780456 Chr.19: 43951158-43951158 on Build GRCh38
rs1045439882 Chr.19: 43951104-43951104 on Build GRCh38
rs371143154 Chr.19: 43951433-43951433 on Build GRCh38
rs1025457744 Chr.19: 43951422-43951422 on Build GRCh38
rs772336070 Chr.19: 43951119-43951119 on Build GRCh38
rs1034366760 Chr.19: 43951116-43951116 on Build GRCh38
rs1020122659 Chr.19: 43951450-43951450 on Build GRCh38
rs16976902 Chr.19: 43951244-43951244 on Build GRCh38
rs545129434 Chr.19: 43951499-43951499 on Build GRCh38
rs554138022 Chr.19: 43951248-43951248 on Build GRCh38
rs995258794 Chr.19: 43951134-43951134 on Build GRCh38
rs975057685 Chr.19: 43951527-43951527 on Build GRCh38
rs1045877143 Chr.19: 43951286-43951286 on Build GRCh38
rs907180112 Chr.19: 43951099-43951099 on Build GRCh38
rs116639567 Chr.19: 43951509-43951509 on Build GRCh38
rs751536229 Chr.19: 43951411-43951411 on Build GRCh38
rs1055827843 Chr.19: 43951382-43951382 on Build GRCh38
rs777422734 Chr.19: 43951133-43951133 on Build GRCh38
rs919662412 Chr.19: 43951226-43951226 on Build GRCh38
rs377172739 Chr.19: 43951511-43951511 on Build GRCh38
rs1054208884 Chr.19: 43951092-43951092 on Build GRCh38
rs906843449 Chr.19: 43951106-43951106 on Build GRCh38
rs940961936 Chr.19: 43951249-43951249 on Build GRCh38
rs950872095 Chr.19: 43951174-43951174 on Build GRCh38
rs1034199889 Chr.19: 43951120-43951120 on Build GRCh38
rs1055639161 Chr.19: 43951348-43951348 on Build GRCh38
rs911928117 Chr.19: 43951203-43951203 on Build GRCh38
rs937327049 Chr.19: 43951372-43951372 on Build GRCh38
rs1011388063 Chr.19: 43951141-43951141 on Build GRCh38
rs975174274 Chr.19: 43951222-43951222 on Build GRCh38
rs539128528 Chr.19: 43951179-43951179 on Build GRCh38
rs1020660187 Chr.19: 43951142-43951142 on Build GRCh38
rs1001581088 Chr.19: 43951299-43951299 on Build GRCh38
rs1045725830 Chr.19: 43951095-43951095 on Build GRCh38
rs930822741 Chr.19: 43951241-43951241 on Build GRCh38

View Full Product Details