Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44259842-44260380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF233
Entrez Gene ID
353355
Gene Name
zinc finger protein 233
Gene Aliases
-
UniGene
Hs.466891
Gene Chromosome Location
Chr.19: 44259880-44275317 on Build GRCh38
Transcripts
NM_001207005.1 NM_181756.2 XM_017026758.1 XM_017026759.1 XM_017026760.1 XM_017026761.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7190279 c.178G>A p.V60I Chr.19: 44259918-44259918 on Build GRCh38
5768600 c.214A>G p.T72A Chr.19: 44259954-44259954 on Build GRCh38
5422380 c.234C>T p.S78S Chr.19: 44259974-44259974 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs146867371 Chr.19: 44260273-44260273 on Build GRCh38
rs527279715 Chr.19: 44259950-44259950 on Build GRCh38
rs567330553 Chr.19: 44259999-44259999 on Build GRCh38
rs752727851 Chr.19: 44259918-44259918 on Build GRCh38
rs750821246 Chr.19: 44259970-44259970 on Build GRCh38
rs940272216 Chr.19: 44260121-44260121 on Build GRCh38
rs960409533 Chr.19: 44260035-44260035 on Build GRCh38
rs959875605 Chr.19: 44260280-44260280 on Build GRCh38
rs934087299 Chr.19: 44259930-44259930 on Build GRCh38
rs946541117 Chr.19: 44260209-44260209 on Build GRCh38
rs186334053 Chr.19: 44260319-44260319 on Build GRCh38
rs8100437 Chr.19: 44260197-44260197 on Build GRCh38
rs554018291 Chr.19: 44260135-44260136 on Build GRCh38
rs761710664 Chr.19: 44259873-44259873 on Build GRCh38
rs948975935 Chr.19: 44259897-44259897 on Build GRCh38
rs760627428 Chr.19: 44259932-44259932 on Build GRCh38
rs767369974 Chr.19: 44260044-44260044 on Build GRCh38
rs953414130 Chr.19: 44259921-44259921 on Build GRCh38
rs1007573211 Chr.19: 44260158-44260158 on Build GRCh38
rs767580826 Chr.19: 44259917-44259917 on Build GRCh38
rs992573512 Chr.19: 44260009-44260009 on Build GRCh38
rs972676774 Chr.19: 44260123-44260123 on Build GRCh38
rs936934845 Chr.19: 44260080-44260080 on Build GRCh38
rs1052668471 Chr.19: 44260092-44260092 on Build GRCh38
rs774291447 Chr.19: 44259908-44259908 on Build GRCh38
rs566111104 Chr.19: 44260178-44260178 on Build GRCh38
rs190756513 Chr.19: 44260138-44260138 on Build GRCh38
rs562456061 Chr.19: 44259885-44259885 on Build GRCh38
rs1049543509 Chr.19: 44259936-44259936 on Build GRCh38
rs8100414 Chr.19: 44260132-44260132 on Build GRCh38
rs1016708148 Chr.19: 44260159-44260159 on Build GRCh38
rs993416515 Chr.19: 44260227-44260227 on Build GRCh38
rs1043969160 Chr.19: 44260210-44260210 on Build GRCh38
rs1005681492 Chr.19: 44260354-44260354 on Build GRCh38
rs760408934 Chr.19: 44260155-44260155 on Build GRCh38
rs1056132212 Chr.19: 44260274-44260274 on Build GRCh38
rs574555994 Chr.19: 44260267-44260267 on Build GRCh38
rs8101360 Chr.19: 44260186-44260186 on Build GRCh38
rs758713799 Chr.19: 44260023-44260023 on Build GRCh38
rs909853049 Chr.19: 44259899-44259899 on Build GRCh38
rs550370731 Chr.19: 44260116-44260116 on Build GRCh38
rs940010776 Chr.19: 44259903-44259903 on Build GRCh38
rs753119676 Chr.19: 44260328-44260328 on Build GRCh38
rs191072831 Chr.19: 44260351-44260351 on Build GRCh38
rs925569265 Chr.19: 44260059-44260059 on Build GRCh38
rs963598267 Chr.19: 44259989-44259989 on Build GRCh38
rs889606346 Chr.19: 44260192-44260192 on Build GRCh38
rs8100554 Chr.19: 44260266-44260266 on Build GRCh38
rs769999004 Chr.19: 44259898-44259898 on Build GRCh38
rs901228199 Chr.19: 44259925-44259925 on Build GRCh38
rs115638388 Chr.19: 44260254-44260254 on Build GRCh38
rs780374965 Chr.19: 44260079-44260079 on Build GRCh38
rs978809657 Chr.19: 44260047-44260047 on Build GRCh38
rs570243322 Chr.19: 44260137-44260137 on Build GRCh38
rs544952437 Chr.19: 44259939-44259939 on Build GRCh38
rs756876415 Chr.19: 44259951-44259951 on Build GRCh38
rs763576607 Chr.19: 44259946-44259946 on Build GRCh38
rs984763789 Chr.19: 44260068-44260069 on Build GRCh38
rs547473178 Chr.19: 44259959-44259959 on Build GRCh38
rs183118948 Chr.19: 44260216-44260216 on Build GRCh38
rs558979674 Chr.19: 44260165-44260165 on Build GRCh38
rs919116382 Chr.19: 44260001-44260001 on Build GRCh38
rs761730641 Chr.19: 44259934-44259934 on Build GRCh38
rs567186108 Chr.19: 44260288-44260288 on Build GRCh38
rs896190921 Chr.19: 44260275-44260275 on Build GRCh38
rs765881910 Chr.19: 44260302-44260302 on Build GRCh38
rs948413908 Chr.19: 44260015-44260015 on Build GRCh38
rs915609311 Chr.19: 44260012-44260012 on Build GRCh38
rs530333705 Chr.19: 44260109-44260109 on Build GRCh38
rs564927549 Chr.19: 44259947-44259947 on Build GRCh38
rs781144052 Chr.19: 44260190-44260190 on Build GRCh38
rs548264131 Chr.19: 44260310-44260310 on Build GRCh38
rs1233444 Chr.19: 44259911-44259911 on Build GRCh38
rs569029770 Chr.19: 44260107-44260107 on Build GRCh38
rs1023135236 Chr.19: 44260053-44260053 on Build GRCh38
rs749855556 Chr.19: 44260002-44260002 on Build GRCh38

View Full Product Details