Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45939425-45939926 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOVA2
Entrez Gene ID
4858
Gene Name
NOVA alternative splicing regulator 2
Gene Aliases
ANOVA, NOVA3
UniGene
Hs.375439
Gene Chromosome Location
Chr.19: 45933734-45973399 on Build GRCh38
Transcripts
NM_002516.3 XM_006723230.3 XM_017026838.1 XM_017026839.1 XM_017026840.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7168226 c.1443G>C p.Q481H Chr.19: 45939899-45939899 on Build GRCh38
3536034 c.1431C>T p.V477V Chr.19: 45939911-45939911 on Build GRCh38
5856200 c.1433C>T p.T478I Chr.19: 45939909-45939909 on Build GRCh38
6728311 c.1427G>A p.R476Q Chr.19: 45939915-45939915 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs897265806 Chr.19: 45939460-45939460 on Build GRCh38
rs568257652 Chr.19: 45939454-45939454 on Build GRCh38
rs750854209 Chr.19: 45939814-45939816 on Build GRCh38
rs1007336755 Chr.19: 45939811-45939811 on Build GRCh38
rs907782024 Chr.19: 45939861-45939861 on Build GRCh38
rs975911666 Chr.19: 45939641-45939641 on Build GRCh38
rs1018359446 Chr.19: 45939502-45939502 on Build GRCh38
rs996161469 Chr.19: 45939513-45939513 on Build GRCh38
rs986985657 Chr.19: 45939595-45939623 on Build GRCh38
rs995481843 Chr.19: 45939468-45939468 on Build GRCh38
rs754138677 Chr.19: 45939905-45939905 on Build GRCh38
rs561120388 Chr.19: 45939688-45939688 on Build GRCh38
rs771032556 Chr.19: 45939792-45939793 on Build GRCh38
rs1020506925 Chr.19: 45939790-45939790 on Build GRCh38
rs537383279 Chr.19: 45939464-45939464 on Build GRCh38
rs573915980 Chr.19: 45939794-45939794 on Build GRCh38
rs746954601 Chr.19: 45939817-45939817 on Build GRCh38
rs914242762 Chr.19: 45939687-45939687 on Build GRCh38
rs748000755 Chr.19: 45939825-45939825 on Build GRCh38
rs762542956 Chr.19: 45939834-45939834 on Build GRCh38
rs889790269 Chr.19: 45939805-45939805 on Build GRCh38
rs558664673 Chr.19: 45939769-45939769 on Build GRCh38
rs933243327 Chr.19: 45939630-45939630 on Build GRCh38
rs907717362 Chr.19: 45939657-45939657 on Build GRCh38
rs1018410528 Chr.19: 45939799-45939799 on Build GRCh38
rs764499606 Chr.19: 45939875-45939875 on Build GRCh38
rs769508580 Chr.19: 45939828-45939828 on Build GRCh38
rs765903387 Chr.19: 45939842-45939842 on Build GRCh38
rs1044646806 Chr.19: 45939724-45939724 on Build GRCh38
rs1026999451 Chr.19: 45939578-45939578 on Build GRCh38
rs537146196 Chr.19: 45939503-45939503 on Build GRCh38
rs550902744 Chr.19: 45939573-45939573 on Build GRCh38
rs923248212 Chr.19: 45939620-45939620 on Build GRCh38
rs754790207 Chr.19: 45939619-45939619 on Build GRCh38
rs374293055 Chr.19: 45939845-45939845 on Build GRCh38
rs771735604 Chr.19: 45939483-45939484 on Build GRCh38
rs965897508 Chr.19: 45939637-45939637 on Build GRCh38
rs895449072 Chr.19: 45939759-45939759 on Build GRCh38
rs937047200 Chr.19: 45939715-45939715 on Build GRCh38
rs554785195 Chr.19: 45939804-45939804 on Build GRCh38
rs759742084 Chr.19: 45939482-45939484 on Build GRCh38
rs543564984 Chr.19: 45939854-45939854 on Build GRCh38
rs1028077254 Chr.19: 45939524-45939524 on Build GRCh38
rs772864620 Chr.19: 45939830-45939830 on Build GRCh38
rs539040461 Chr.19: 45939677-45939677 on Build GRCh38
rs369270770 Chr.19: 45939552-45939552 on Build GRCh38
rs907480187 Chr.19: 45939732-45939732 on Build GRCh38
rs983784912 Chr.19: 45939857-45939857 on Build GRCh38
rs941837696 Chr.19: 45939661-45939661 on Build GRCh38
rs756409134 Chr.19: 45939816-45939816 on Build GRCh38
rs768129569 Chr.19: 45939850-45939850 on Build GRCh38
rs911499712 Chr.19: 45939799-45939813 on Build GRCh38
rs776416570 Chr.19: 45939824-45939824 on Build GRCh38
rs1017977491 Chr.19: 45939633-45939633 on Build GRCh38
rs572530689 Chr.19: 45939773-45939773 on Build GRCh38
rs964063962 Chr.19: 45939601-45939601 on Build GRCh38
rs372964567 Chr.19: 45939712-45939712 on Build GRCh38
rs574703482 Chr.19: 45939846-45939846 on Build GRCh38
rs149556287 Chr.19: 45939685-45939686 on Build GRCh38
rs1057193289 Chr.19: 45939757-45939757 on Build GRCh38
rs370671584 Chr.19: 45939818-45939818 on Build GRCh38
rs753390329 Chr.19: 45939481-45939484 on Build GRCh38
rs905600037 Chr.19: 45939682-45939682 on Build GRCh38
rs77788223 Chr.19: 45939496-45939503 on Build GRCh38
rs869262058 Chr.19: 45939497-45939501 on Build GRCh38
rs963204123 Chr.19: 45939897-45939897 on Build GRCh38
rs570628347 Chr.19: 45939650-45939650 on Build GRCh38
rs1000348891 Chr.19: 45939795-45939795 on Build GRCh38
rs777677337 Chr.19: 45939695-45939695 on Build GRCh38
rs924341199 Chr.19: 45939647-45939647 on Build GRCh38
rs370545814 Chr.19: 45939860-45939860 on Build GRCh38

View Full Product Details