Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46678242-46678749 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRKD2
Entrez Gene ID
25865
Gene Name
protein kinase D2
Gene Aliases
HSPC187, PKD2, nPKC-D2
UniGene
Hs.466987
Gene Chromosome Location
Chr.19: 46674316-46717127 on Build GRCh38
Transcripts
NM_001079880.1 NM_001079881.1 NM_001079882.1 NM_016457.4 XM_005258716.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4079593 c.2161G>A p.E721K Chr.19: 46678573-46678573 on Build GRCh38
712233 c.2126G>A p.R709H Chr.19: 46678608-46678608 on Build GRCh38
439840 c.2277C>G p.D759E Chr.19: 46678457-46678457 on Build GRCh38
6755998 c.2130A>G p.S710S Chr.19: 46678604-46678604 on Build GRCh38
6151432 c.2213G>T p.G738V Chr.19: 46678521-46678521 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371434874 Chr.19: 46678695-46678695 on Build GRCh38
rs754648498 Chr.19: 46678386-46678386 on Build GRCh38
rs752280760 Chr.19: 46678461-46678461 on Build GRCh38
rs775542092 Chr.19: 46678428-46678428 on Build GRCh38
rs926442685 Chr.19: 46678631-46678631 on Build GRCh38
rs780298147 Chr.19: 46678668-46678668 on Build GRCh38
rs753514675 Chr.19: 46678711-46678711 on Build GRCh38
rs201854545 Chr.19: 46678541-46678541 on Build GRCh38
rs370149673 Chr.19: 46678435-46678435 on Build GRCh38
rs750316895 Chr.19: 46678374-46678374 on Build GRCh38
rs866307765 Chr.19: 46678477-46678477 on Build GRCh38
rs779832744 Chr.19: 46678382-46678382 on Build GRCh38
rs543781700 Chr.19: 46678333-46678333 on Build GRCh38
rs1036127381 Chr.19: 46678289-46678289 on Build GRCh38
rs112260573 Chr.19: 46678496-46678496 on Build GRCh38
rs766259352 Chr.19: 46678445-46678445 on Build GRCh38
rs905995871 Chr.19: 46678267-46678267 on Build GRCh38
rs755714612 Chr.19: 46678465-46678465 on Build GRCh38
rs760478401 Chr.19: 46678701-46678701 on Build GRCh38
rs761841448 Chr.19: 46678577-46678577 on Build GRCh38
rs756792184 Chr.19: 46678624-46678624 on Build GRCh38
rs767250424 Chr.19: 46678454-46678454 on Build GRCh38
rs761463286 Chr.19: 46678712-46678712 on Build GRCh38
rs775169586 Chr.19: 46678698-46678698 on Build GRCh38
rs777150439 Chr.19: 46678475-46678475 on Build GRCh38
rs199910856 Chr.19: 46678466-46678466 on Build GRCh38
rs752547523 Chr.19: 46678386-46678386 on Build GRCh38
rs757860667 Chr.19: 46678484-46678484 on Build GRCh38
rs780669827 Chr.19: 46678398-46678398 on Build GRCh38
rs748749845 Chr.19: 46678478-46678478 on Build GRCh38
rs775850563 Chr.19: 46678517-46678517 on Build GRCh38
rs751301703 Chr.19: 46678383-46678383 on Build GRCh38
rs752841343 Chr.19: 46678364-46678364 on Build GRCh38
rs55933311 Chr.19: 46678417-46678417 on Build GRCh38
rs771407663 Chr.19: 46678326-46678326 on Build GRCh38
rs368181665 Chr.19: 46678368-46678368 on Build GRCh38
rs928344124 Chr.19: 46678325-46678325 on Build GRCh38
rs982819869 Chr.19: 46678520-46678520 on Build GRCh38
rs867339595 Chr.19: 46678288-46678288 on Build GRCh38
rs376566966 Chr.19: 46678616-46678616 on Build GRCh38
rs748276771 Chr.19: 46678681-46678681 on Build GRCh38
rs772365850 Chr.19: 46678427-46678427 on Build GRCh38
rs980297233 Chr.19: 46678309-46678309 on Build GRCh38
rs34376220 Chr.19: 46678380-46678380 on Build GRCh38
rs747736186 Chr.19: 46678407-46678407 on Build GRCh38
rs956064837 Chr.19: 46678725-46678725 on Build GRCh38
rs746360678 Chr.19: 46678628-46678628 on Build GRCh38
rs754211889 Chr.19: 46678542-46678542 on Build GRCh38
rs371552622 Chr.19: 46678670-46678670 on Build GRCh38
rs1006660174 Chr.19: 46678589-46678589 on Build GRCh38
rs978930540 Chr.19: 46678389-46678389 on Build GRCh38
rs769166457 Chr.19: 46678409-46678409 on Build GRCh38
rs779264263 Chr.19: 46678508-46678508 on Build GRCh38
rs754736557 Chr.19: 46678451-46678451 on Build GRCh38
rs774251458 Chr.19: 46678592-46678592 on Build GRCh38
rs139718534 Chr.19: 46678706-46678706 on Build GRCh38
rs765155895 Chr.19: 46678347-46678348 on Build GRCh38
rs753426610 Chr.19: 46678619-46678619 on Build GRCh38
rs767789374 Chr.19: 46678354-46678354 on Build GRCh38
rs201032879 Chr.19: 46678574-46678574 on Build GRCh38
rs901895501 Chr.19: 46678682-46678682 on Build GRCh38
rs763752080 Chr.19: 46678617-46678617 on Build GRCh38
rs768869479 Chr.19: 46678561-46678561 on Build GRCh38
rs768796295 Chr.19: 46678671-46678671 on Build GRCh38
rs753891237 Chr.19: 46678442-46678442 on Build GRCh38
rs201225333 Chr.19: 46678410-46678410 on Build GRCh38
rs764079163 Chr.19: 46678441-46678441 on Build GRCh38
rs763425050 Chr.19: 46678318-46678318 on Build GRCh38
rs776803892 Chr.19: 46678676-46678676 on Build GRCh38
rs779519593 Chr.19: 46678495-46678495 on Build GRCh38
rs970207778 Chr.19: 46678273-46678273 on Build GRCh38
rs758876482 Chr.19: 46678636-46678636 on Build GRCh38
rs113283031 Chr.19: 46678707-46678707 on Build GRCh38
rs368256973 Chr.19: 46678686-46678686 on Build GRCh38
rs1010628622 Chr.19: 46678361-46678361 on Build GRCh38
rs771497654 Chr.19: 46678418-46678418 on Build GRCh38
rs564075946 Chr.19: 46678334-46678334 on Build GRCh38
rs774867853 Chr.19: 46678421-46678421 on Build GRCh38
rs35036065 Chr.19: 46678276-46678277 on Build GRCh38
rs774341208 Chr.19: 46678683-46678683 on Build GRCh38
rs767210044 Chr.19: 46678608-46678608 on Build GRCh38
rs766343388 Chr.19: 46678590-46678590 on Build GRCh38
rs759559374 Chr.19: 46678321-46678322 on Build GRCh38
rs757498782 Chr.19: 46678265-46678265 on Build GRCh38
rs779609817 Chr.19: 46678627-46678627 on Build GRCh38

View Full Product Details