Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48639167-48639663 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA11
Entrez Gene ID
770
Gene Name
carbonic anhydrase 11
Gene Aliases
CARPX1
UniGene
Hs.428446
Gene Chromosome Location
Chr.19: 48637942-48646312 on Build GRCh38
Transcripts
NM_001217.4 NR_136241.1 NR_004401.2

Gene Symbol
SEC1P
Entrez Gene ID
653677
Gene Name
secretory blood group 1, pseudogene
Gene Aliases
SEC1
UniGene
Hs.443160
Gene Chromosome Location
Chr.19: 48638039-48682245 on Build GRCh38
Transcripts
NM_001217.4 NR_136241.1 NR_004401.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4792682 c.651C>A p.Y217* Chr.19: 48639449-48639449 on Build GRCh38
1728748 c.760C>T p.L254F Chr.19: 48639340-48639340 on Build GRCh38
3823554 c.762C>G p.L254L Chr.19: 48639338-48639338 on Build GRCh38
1712561 c.716G>A p.G239D Chr.19: 48639384-48639384 on Build GRCh38
4668235 c.652T>C p.F218L Chr.19: 48639448-48639448 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770524116 Chr.19: 48639257-48639257 on Build GRCh38
rs761868132 Chr.19: 48639442-48639442 on Build GRCh38
rs376085341 Chr.19: 48639384-48639384 on Build GRCh38
rs775227660 Chr.19: 48639285-48639285 on Build GRCh38
rs1045227937 Chr.19: 48639297-48639297 on Build GRCh38
rs773703326 Chr.19: 48639265-48639265 on Build GRCh38
rs754442009 Chr.19: 48639628-48639628 on Build GRCh38
rs371736063 Chr.19: 48639552-48639552 on Build GRCh38
rs776676371 Chr.19: 48639299-48639299 on Build GRCh38
rs780838422 Chr.19: 48639517-48639517 on Build GRCh38
rs758966880 Chr.19: 48639611-48639611 on Build GRCh38
rs769917037 Chr.19: 48639298-48639298 on Build GRCh38
rs563153316 Chr.19: 48639341-48639341 on Build GRCh38
rs929600766 Chr.19: 48639202-48639202 on Build GRCh38
rs969173439 Chr.19: 48639632-48639632 on Build GRCh38
rs752968557 Chr.19: 48639464-48639464 on Build GRCh38
rs145539923 Chr.19: 48639576-48639576 on Build GRCh38
rs768532247 Chr.19: 48639423-48639423 on Build GRCh38
rs142656783 Chr.19: 48639589-48639589 on Build GRCh38
rs909615279 Chr.19: 48639266-48639266 on Build GRCh38
rs199878745 Chr.19: 48639336-48639336 on Build GRCh38
rs984005936 Chr.19: 48639334-48639334 on Build GRCh38
rs532709332 Chr.19: 48639291-48639291 on Build GRCh38
rs776282558 Chr.19: 48639437-48639437 on Build GRCh38
rs760574976 Chr.19: 48639286-48639286 on Build GRCh38
rs779570734 Chr.19: 48639489-48639489 on Build GRCh38
rs779871913 Chr.19: 48639395-48639395 on Build GRCh38
rs751625242 Chr.19: 48639326-48639326 on Build GRCh38
rs370565241 Chr.19: 48639331-48639331 on Build GRCh38
rs146524258 Chr.19: 48639598-48639598 on Build GRCh38
rs892495253 Chr.19: 48639507-48639507 on Build GRCh38
rs779675680 Chr.19: 48639505-48639505 on Build GRCh38
rs766546963 Chr.19: 48639323-48639323 on Build GRCh38
rs140803568 Chr.19: 48639347-48639347 on Build GRCh38
rs1022373361 Chr.19: 48639533-48639533 on Build GRCh38
rs370921025 Chr.19: 48639511-48639511 on Build GRCh38
rs918277261 Chr.19: 48639200-48639200 on Build GRCh38
rs775869463 Chr.19: 48639377-48639378 on Build GRCh38
rs761621442 Chr.19: 48639314-48639314 on Build GRCh38
rs772181062 Chr.19: 48639563-48639563 on Build GRCh38
rs200825270 Chr.19: 48639290-48639290 on Build GRCh38
rs375501601 Chr.19: 48639499-48639499 on Build GRCh38
rs771839146 Chr.19: 48639269-48639269 on Build GRCh38
rs761018211 Chr.19: 48639570-48639570 on Build GRCh38
rs746636245 Chr.19: 48639403-48639403 on Build GRCh38
rs945249085 Chr.19: 48639523-48639523 on Build GRCh38
rs759552086 Chr.19: 48639330-48639330 on Build GRCh38
rs921968093 Chr.19: 48639471-48639472 on Build GRCh38
rs141145201 Chr.19: 48639601-48639601 on Build GRCh38
rs950494853 Chr.19: 48639618-48639618 on Build GRCh38
rs757703781 Chr.19: 48639371-48639371 on Build GRCh38
rs368624160 Chr.19: 48639470-48639470 on Build GRCh38
rs192724985 Chr.19: 48639454-48639454 on Build GRCh38
rs758636829 Chr.19: 48639191-48639191 on Build GRCh38
rs766042493 Chr.19: 48639617-48639617 on Build GRCh38
rs750908345 Chr.19: 48639494-48639494 on Build GRCh38
rs747900497 Chr.19: 48639522-48639522 on Build GRCh38
rs769632800 Chr.19: 48639446-48639446 on Build GRCh38
rs754982944 Chr.19: 48639620-48639620 on Build GRCh38
rs369114898 Chr.19: 48639390-48639390 on Build GRCh38
rs148357487 Chr.19: 48639267-48639267 on Build GRCh38
rs1008020295 Chr.19: 48639536-48639536 on Build GRCh38
rs754409395 Chr.19: 48639356-48639356 on Build GRCh38
rs150549053 Chr.19: 48639586-48639586 on Build GRCh38
rs758028258 Chr.19: 48639496-48639496 on Build GRCh38
rs1033271237 Chr.19: 48639263-48639263 on Build GRCh38
rs775545136 Chr.19: 48639566-48639566 on Build GRCh38
rs760777101 Chr.19: 48639469-48639469 on Build GRCh38
rs112078432 Chr.19: 48639548-48639548 on Build GRCh38
rs780447021 Chr.19: 48639527-48639527 on Build GRCh38
rs757585813 Chr.19: 48639479-48639479 on Build GRCh38
rs754143954 Chr.19: 48639473-48639473 on Build GRCh38
rs141672508 Chr.19: 48639534-48639534 on Build GRCh38
rs537702330 Chr.19: 48639386-48639386 on Build GRCh38
rs773065782 Chr.19: 48639449-48639449 on Build GRCh38

View Full Product Details