Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48661317-48661817 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NTN5
Entrez Gene ID
126147
Gene Name
netrin 5
Gene Aliases
-
UniGene
Hs.326217
Gene Chromosome Location
Chr.19: 48661407-48673119 on Build GRCh38
Transcripts
NM_145807.2 XM_017026274.1 NR_004401.2

Gene Symbol
SEC1P
Entrez Gene ID
653677
Gene Name
secretory blood group 1, pseudogene
Gene Aliases
SEC1
UniGene
Hs.443160
Gene Chromosome Location
Chr.19: 48638039-48682245 on Build GRCh38
Transcripts
NM_145807.2 XM_017026274.1 NR_004401.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5833806 c.1408_1410delGAG p.E470delE Chr.19: 48661737-48661739 on Build GRCh38
7270768 c.1348G>T p.G450C Chr.19: 48661799-48661799 on Build GRCh38
5741670 c.1417G>A p.G473R Chr.19: 48661730-48661730 on Build GRCh38
5763129 c.1411C>T p.R471C Chr.19: 48661736-48661736 on Build GRCh38
6730707 c.1432G>T p.V478L Chr.19: 48661715-48661715 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs281392 Chr.19: 48661695-48661695 on Build GRCh38
rs769814142 Chr.19: 48661624-48661624 on Build GRCh38
rs998161267 Chr.19: 48661527-48661527 on Build GRCh38
rs930028500 Chr.19: 48661364-48661364 on Build GRCh38
rs199848394 Chr.19: 48661696-48661696 on Build GRCh38
rs767982577 Chr.19: 48661663-48661663 on Build GRCh38
rs759748406 Chr.19: 48661659-48661659 on Build GRCh38
rs766422739 Chr.19: 48661656-48661656 on Build GRCh38
rs945857365 Chr.19: 48661665-48661665 on Build GRCh38
rs983328962 Chr.19: 48661372-48661372 on Build GRCh38
rs918693005 Chr.19: 48661363-48661363 on Build GRCh38
rs559734915 Chr.19: 48661706-48661706 on Build GRCh38
rs570381533 Chr.19: 48661730-48661730 on Build GRCh38
rs768446257 Chr.19: 48661709-48661709 on Build GRCh38
rs1006974354 Chr.19: 48661507-48661507 on Build GRCh38
rs963208102 Chr.19: 48661605-48661605 on Build GRCh38
rs764499894 Chr.19: 48661728-48661728 on Build GRCh38
rs1021829318 Chr.19: 48661467-48661467 on Build GRCh38
rs760971006 Chr.19: 48661725-48661725 on Build GRCh38
rs35405892 Chr.19: 48661621-48661621 on Build GRCh38
rs1018491891 Chr.19: 48661514-48661514 on Build GRCh38
rs545653841 Chr.19: 48661538-48661538 on Build GRCh38
rs748045623 Chr.19: 48661711-48661711 on Build GRCh38
rs532776696 Chr.19: 48661439-48661439 on Build GRCh38
rs552076829 Chr.19: 48661729-48661729 on Build GRCh38
rs780790400 Chr.19: 48661557-48661557 on Build GRCh38
rs887448635 Chr.19: 48661619-48661619 on Build GRCh38
rs746645117 Chr.19: 48661763-48661763 on Build GRCh38
rs965574411 Chr.19: 48661526-48661526 on Build GRCh38
rs764902141 Chr.19: 48661735-48661735 on Build GRCh38
rs773076052 Chr.19: 48661780-48661780 on Build GRCh38
rs993839853 Chr.19: 48661563-48661563 on Build GRCh38
rs776588151 Chr.19: 48661651-48661651 on Build GRCh38
rs979897461 Chr.19: 48661615-48661615 on Build GRCh38
rs576000113 Chr.19: 48661685-48661685 on Build GRCh38
rs780953303 Chr.19: 48661768-48661768 on Build GRCh38
rs767638933 Chr.19: 48661718-48661718 on Build GRCh38
rs748151966 Chr.19: 48661769-48661769 on Build GRCh38
rs754654912 Chr.19: 48661534-48661534 on Build GRCh38
rs759554123 Chr.19: 48661717-48661717 on Build GRCh38
rs764611256 Chr.19: 48661355-48661355 on Build GRCh38
rs771064689 Chr.19: 48661715-48661715 on Build GRCh38
rs557632600 Chr.19: 48661566-48661566 on Build GRCh38
rs763040934 Chr.19: 48661654-48661654 on Build GRCh38
rs1025676409 Chr.19: 48661753-48661753 on Build GRCh38
rs111408451 Chr.19: 48661625-48661625 on Build GRCh38
rs570196205 Chr.19: 48661737-48661739 on Build GRCh38
rs1044332267 Chr.19: 48661433-48661433 on Build GRCh38
rs779686399 Chr.19: 48661702-48661702 on Build GRCh38
rs770526726 Chr.19: 48661631-48661631 on Build GRCh38
rs745956709 Chr.19: 48661788-48661788 on Build GRCh38
rs745496710 Chr.19: 48661635-48661635 on Build GRCh38
rs941959579 Chr.19: 48661397-48661397 on Build GRCh38
rs879900003 Chr.19: 48661346-48661346 on Build GRCh38
rs951522710 Chr.19: 48661537-48661537 on Build GRCh38
rs923452961 Chr.19: 48661410-48661410 on Build GRCh38
rs927420431 Chr.19: 48661523-48661523 on Build GRCh38
rs891560435 Chr.19: 48661453-48661453 on Build GRCh38
rs754163722 Chr.19: 48661429-48661429 on Build GRCh38
rs531320794 Chr.19: 48661762-48661762 on Build GRCh38
rs757612285 Chr.19: 48661435-48661435 on Build GRCh38
rs1044365676 Chr.19: 48661492-48661492 on Build GRCh38
rs998833186 Chr.19: 48661742-48661742 on Build GRCh38
rs752595529 Chr.19: 48661743-48661745 on Build GRCh38
rs757838150 Chr.19: 48661731-48661731 on Build GRCh38
rs909023991 Chr.19: 48661376-48661376 on Build GRCh38
rs776479410 Chr.19: 48661710-48661710 on Build GRCh38
rs115919347 Chr.19: 48661687-48661687 on Build GRCh38
rs1047600071 Chr.19: 48661591-48661591 on Build GRCh38
rs533570940 Chr.19: 48661721-48661721 on Build GRCh38
rs774400591 Chr.19: 48661716-48661716 on Build GRCh38
rs769728097 Chr.19: 48661777-48661777 on Build GRCh38
rs770743005 Chr.19: 48661765-48661765 on Build GRCh38
rs1010389849 Chr.19: 48661454-48661454 on Build GRCh38
rs756434952 Chr.19: 48661682-48661682 on Build GRCh38
rs748813199 Chr.19: 48661627-48661627 on Build GRCh38
rs747682529 Chr.19: 48661561-48661561 on Build GRCh38
rs989054436 Chr.19: 48661452-48661452 on Build GRCh38
rs111816376 Chr.19: 48661342-48661343 on Build GRCh38
rs757881929 Chr.19: 48661755-48661755 on Build GRCh38
rs1057172056 Chr.19: 48661440-48661440 on Build GRCh38
rs202110682 Chr.19: 48661698-48661698 on Build GRCh38
rs281391 Chr.19: 48661483-48661483 on Build GRCh38
rs768477843 Chr.19: 48661646-48661646 on Build GRCh38
rs758972559 Chr.19: 48661702-48661705 on Build GRCh38
rs773250607 Chr.19: 48661670-48661670 on Build GRCh38
rs772035423 Chr.19: 48661638-48661638 on Build GRCh38
rs760728575 Chr.19: 48661645-48661645 on Build GRCh38
rs553730471 Chr.19: 48661378-48661378 on Build GRCh38
rs202223346 Chr.19: 48661744-48661744 on Build GRCh38
rs779592292 Chr.19: 48661760-48661760 on Build GRCh38

View Full Product Details