Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48664290-48664786 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NTN5
Entrez Gene ID
126147
Gene Name
netrin 5
Gene Aliases
-
UniGene
Hs.326217
Gene Chromosome Location
Chr.19: 48661407-48673119 on Build GRCh38
Transcripts
NM_145807.2 XM_011526443.2 XM_017026274.1 NR_004401.2

Gene Symbol
SEC1P
Entrez Gene ID
653677
Gene Name
secretory blood group 1, pseudogene
Gene Aliases
SEC1
UniGene
Hs.443160
Gene Chromosome Location
Chr.19: 48638039-48682245 on Build GRCh38
Transcripts
NM_145807.2 XM_011526443.2 XM_017026274.1 NR_004401.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
302779 c.820G>T p.A274S Chr.19: 48664579-48664579 on Build GRCh38
4998955 c.635G>A p.C212Y Chr.19: 48664764-48664764 on Build GRCh38
3107755 c.721C>T p.R241W Chr.19: 48664678-48664678 on Build GRCh38
4877957 c.777G>T p.W259C Chr.19: 48664622-48664622 on Build GRCh38
6386501 c.730C>T p.H244Y Chr.19: 48664669-48664669 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111606022 Chr.19: 48664422-48664422 on Build GRCh38
rs770975532 Chr.19: 48664634-48664634 on Build GRCh38
rs962450569 Chr.19: 48664408-48664408 on Build GRCh38
rs939994621 Chr.19: 48664389-48664389 on Build GRCh38
rs1036954225 Chr.19: 48664391-48664391 on Build GRCh38
rs765599298 Chr.19: 48664673-48664673 on Build GRCh38
rs754816283 Chr.19: 48664698-48664698 on Build GRCh38
rs376906018 Chr.19: 48664672-48664672 on Build GRCh38
rs769197839 Chr.19: 48664711-48664711 on Build GRCh38
rs767288192 Chr.19: 48664699-48664699 on Build GRCh38
rs762324837 Chr.19: 48664682-48664682 on Build GRCh38
rs772351756 Chr.19: 48664646-48664646 on Build GRCh38
rs755814946 Chr.19: 48664742-48664742 on Build GRCh38
rs764626373 Chr.19: 48664340-48664340 on Build GRCh38
rs780265011 Chr.19: 48664747-48664747 on Build GRCh38
rs781061581 Chr.19: 48664558-48664558 on Build GRCh38
rs750056666 Chr.19: 48664549-48664549 on Build GRCh38
rs762568687 Chr.19: 48664735-48664735 on Build GRCh38
rs141446313 Chr.19: 48664623-48664623 on Build GRCh38
rs114909456 Chr.19: 48664594-48664594 on Build GRCh38
rs759791666 Chr.19: 48664576-48664576 on Build GRCh38
rs148717503 Chr.19: 48664603-48664603 on Build GRCh38
rs772384209 Chr.19: 48664582-48664582 on Build GRCh38
rs367658447 Chr.19: 48664705-48664705 on Build GRCh38
rs376771984 Chr.19: 48664740-48664740 on Build GRCh38
rs769328411 Chr.19: 48664554-48664555 on Build GRCh38
rs544641083 Chr.19: 48664739-48664739 on Build GRCh38
rs868658601 Chr.19: 48664469-48664469 on Build GRCh38
rs761061833 Chr.19: 48664590-48664590 on Build GRCh38
rs751183242 Chr.19: 48664695-48664695 on Build GRCh38
rs987032734 Chr.19: 48664496-48664496 on Build GRCh38
rs770358549 Chr.19: 48664728-48664728 on Build GRCh38
rs1015383573 Chr.19: 48664395-48664395 on Build GRCh38
rs925261323 Chr.19: 48664692-48664692 on Build GRCh38
rs546925909 Chr.19: 48664510-48664510 on Build GRCh38
rs748055945 Chr.19: 48664632-48664632 on Build GRCh38
rs540482894 Chr.19: 48664677-48664677 on Build GRCh38
rs796789099 Chr.19: 48664487-48664487 on Build GRCh38
rs530605400 Chr.19: 48664385-48664385 on Build GRCh38
rs781149146 Chr.19: 48664628-48664628 on Build GRCh38
rs958261088 Chr.19: 48664629-48664629 on Build GRCh38
rs528427395 Chr.19: 48664449-48664449 on Build GRCh38
rs368780671 Chr.19: 48664670-48664670 on Build GRCh38
rs1015250762 Chr.19: 48664614-48664614 on Build GRCh38
rs764778785 Chr.19: 48664598-48664598 on Build GRCh38
rs777128239 Chr.19: 48664726-48664726 on Build GRCh38
rs774978236 Chr.19: 48664704-48664712 on Build GRCh38
rs779047949 Chr.19: 48664641-48664641 on Build GRCh38
rs775806148 Chr.19: 48664654-48664654 on Build GRCh38
rs184969096 Chr.19: 48664521-48664521 on Build GRCh38
rs371752057 Chr.19: 48664727-48664727 on Build GRCh38
rs144755455 Chr.19: 48664529-48664529 on Build GRCh38
rs930978428 Chr.19: 48664481-48664481 on Build GRCh38
rs549190722 Chr.19: 48664426-48664426 on Build GRCh38
rs371501834 Chr.19: 48664700-48664700 on Build GRCh38
rs758078058 Chr.19: 48664602-48664602 on Build GRCh38
rs751381720 Chr.19: 48664620-48664620 on Build GRCh38
rs373772020 Chr.19: 48664703-48664703 on Build GRCh38
rs945430732 Chr.19: 48664501-48664501 on Build GRCh38
rs745867762 Chr.19: 48664645-48664645 on Build GRCh38
rs756775632 Chr.19: 48664537-48664537 on Build GRCh38
rs750098003 Chr.19: 48664601-48664601 on Build GRCh38
rs761446704 Chr.19: 48664596-48664596 on Build GRCh38
rs912563542 Chr.19: 48664498-48664499 on Build GRCh38
rs770968795 Chr.19: 48664569-48664569 on Build GRCh38
rs999659722 Chr.19: 48664609-48664609 on Build GRCh38
rs567448100 Chr.19: 48664448-48664448 on Build GRCh38
rs372577584 Chr.19: 48664744-48664744 on Build GRCh38
rs756683918 Chr.19: 48664337-48664337 on Build GRCh38
rs201443263 Chr.19: 48664736-48664736 on Build GRCh38
rs563414669 Chr.19: 48664317-48664317 on Build GRCh38
rs17851884 Chr.19: 48664748-48664748 on Build GRCh38
rs745982299 Chr.19: 48664702-48664702 on Build GRCh38
rs1013738348 Chr.19: 48664536-48664536 on Build GRCh38
rs766090776 Chr.19: 48664689-48664689 on Build GRCh38
rs777324588 Chr.19: 48664671-48664671 on Build GRCh38
rs746743820 Chr.19: 48664552-48664552 on Build GRCh38
rs758173584 Chr.19: 48664551-48664551 on Build GRCh38
rs1053863850 Chr.19: 48664752-48664752 on Build GRCh38
rs199710487 Chr.19: 48664743-48664743 on Build GRCh38
rs773556123 Chr.19: 48664734-48664734 on Build GRCh38
rs780459269 Chr.19: 48664706-48664706 on Build GRCh38
rs974262419 Chr.19: 48664419-48664419 on Build GRCh38
rs768597125 Chr.19: 48664553-48664553 on Build GRCh38
rs778458958 Chr.19: 48664548-48664548 on Build GRCh38
rs775927885 Chr.19: 48664583-48664583 on Build GRCh38
rs774486814 Chr.19: 48664571-48664571 on Build GRCh38
rs192509971 Chr.19: 48664701-48664701 on Build GRCh38
rs749403281 Chr.19: 48664563-48664563 on Build GRCh38
rs189737503 Chr.19: 48664522-48664522 on Build GRCh38
rs370652949 Chr.19: 48664320-48664320 on Build GRCh38
rs577386582 Chr.19: 48664452-48664453 on Build GRCh38
rs749849106 Chr.19: 48664707-48664707 on Build GRCh38

View Full Product Details