Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48713768-48714261 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAMSTR
Entrez Gene ID
284358
Gene Name
MEF2 activating motif and SAP domain containing transcriptional regulator
Gene Aliases
MASTR
UniGene
Hs.191815
Gene Chromosome Location
Chr.19: 48705718-48719898 on Build GRCh38
Transcripts
NM_001130915.1 NM_001297753.1 NM_182574.2 XM_011526807.2 XM_011526808.2 XM_011526809.2 XM_017026640.1 XM_017026641.1 XM_017026642.1 XM_017026643.1 XM_017026644.1 XM_017026645.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6228380 c.742T>G p.S248A Chr.19: 48714027-48714027 on Build GRCh38
6706574 c.726C>T p.V242V Chr.19: 48714043-48714043 on Build GRCh38
6198603 c.851C>A p.P284Q Chr.19: 48713918-48713918 on Build GRCh38
3422977 c.815C>G p.P272R Chr.19: 48713954-48713954 on Build GRCh38
1214318 c.767C>T p.A256V Chr.19: 48714002-48714002 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758980028 Chr.19: 48714040-48714040 on Build GRCh38
rs748885723 Chr.19: 48713807-48713808 on Build GRCh38
rs1046360685 Chr.19: 48714193-48714193 on Build GRCh38
rs765136248 Chr.19: 48713870-48713870 on Build GRCh38
rs752775222 Chr.19: 48713910-48713910 on Build GRCh38
rs372319706 Chr.19: 48713976-48713976 on Build GRCh38
rs181475230 Chr.19: 48713974-48713974 on Build GRCh38
rs761870992 Chr.19: 48714034-48714034 on Build GRCh38
rs775751284 Chr.19: 48714022-48714022 on Build GRCh38
rs768181577 Chr.19: 48713926-48713926 on Build GRCh38
rs1034170139 Chr.19: 48713927-48713927 on Build GRCh38
rs1031465470 Chr.19: 48714015-48714015 on Build GRCh38
rs1031895356 Chr.19: 48713885-48713885 on Build GRCh38
rs560663961 Chr.19: 48714241-48714241 on Build GRCh38
rs374338052 Chr.19: 48713924-48713924 on Build GRCh38
rs281386 Chr.19: 48714048-48714048 on Build GRCh38
rs772096251 Chr.19: 48713959-48713960 on Build GRCh38
rs768858262 Chr.19: 48714093-48714093 on Build GRCh38
rs919682510 Chr.19: 48714195-48714195 on Build GRCh38
rs990098043 Chr.19: 48713911-48713911 on Build GRCh38
rs757740623 Chr.19: 48713829-48713829 on Build GRCh38
rs953799788 Chr.19: 48714064-48714064 on Build GRCh38
rs1037696087 Chr.19: 48714127-48714127 on Build GRCh38
rs920906564 Chr.19: 48713835-48713835 on Build GRCh38
rs371967263 Chr.19: 48713842-48713842 on Build GRCh38
rs572750501 Chr.19: 48714060-48714060 on Build GRCh38
rs957350051 Chr.19: 48714223-48714223 on Build GRCh38
rs549511294 Chr.19: 48713992-48713992 on Build GRCh38
rs780511534 Chr.19: 48714046-48714046 on Build GRCh38
rs749004868 Chr.19: 48714079-48714079 on Build GRCh38
rs1037177193 Chr.19: 48713920-48713920 on Build GRCh38
rs540042962 Chr.19: 48714065-48714065 on Build GRCh38
rs762968050 Chr.19: 48714014-48714014 on Build GRCh38
rs769628040 Chr.19: 48713986-48713986 on Build GRCh38
rs779448848 Chr.19: 48713830-48713830 on Build GRCh38
rs754472029 Chr.19: 48714053-48714053 on Build GRCh38
rs763056541 Chr.19: 48714094-48714094 on Build GRCh38
rs111820143 Chr.19: 48714195-48714199 on Build GRCh38
rs747954141 Chr.19: 48713982-48713982 on Build GRCh38
rs148076324 Chr.19: 48713871-48713871 on Build GRCh38
rs774917272 Chr.19: 48713845-48713845 on Build GRCh38
rs759602557 Chr.19: 48713902-48713902 on Build GRCh38
rs116626237 Chr.19: 48713836-48713836 on Build GRCh38
rs780801701 Chr.19: 48713979-48713979 on Build GRCh38
rs137857137 Chr.19: 48714160-48714160 on Build GRCh38
rs568905558 Chr.19: 48713989-48713989 on Build GRCh38
rs375183994 Chr.19: 48714031-48714031 on Build GRCh38
rs780891751 Chr.19: 48714059-48714059 on Build GRCh38
rs1050004472 Chr.19: 48713998-48713998 on Build GRCh38
rs774413745 Chr.19: 48714083-48714083 on Build GRCh38
rs746680336 Chr.19: 48713848-48713848 on Build GRCh38
rs940215604 Chr.19: 48714114-48714114 on Build GRCh38
rs758776680 Chr.19: 48713955-48713955 on Build GRCh38
rs750707501 Chr.19: 48714035-48714035 on Build GRCh38
rs552948616 Chr.19: 48713794-48713794 on Build GRCh38
rs150770743 Chr.19: 48714032-48714032 on Build GRCh38
rs78556799 Chr.19: 48714183-48714183 on Build GRCh38
rs138857721 Chr.19: 48713939-48713939 on Build GRCh38
rs371082453 Chr.19: 48713882-48713882 on Build GRCh38
rs1013639753 Chr.19: 48714191-48714191 on Build GRCh38
rs760812028 Chr.19: 48714027-48714027 on Build GRCh38
rs1000039168 Chr.19: 48714038-48714038 on Build GRCh38
rs767646790 Chr.19: 48713909-48713909 on Build GRCh38
rs1022892973 Chr.19: 48714103-48714103 on Build GRCh38
rs773011701 Chr.19: 48713971-48713972 on Build GRCh38
rs375155475 Chr.19: 48714208-48714208 on Build GRCh38
rs201114031 Chr.19: 48713879-48713879 on Build GRCh38
rs558390287 Chr.19: 48714073-48714073 on Build GRCh38
rs1011806793 Chr.19: 48714232-48714232 on Build GRCh38
rs746393869 Chr.19: 48713946-48713946 on Build GRCh38
rs372156923 Chr.19: 48714019-48714019 on Build GRCh38
rs141234591 Chr.19: 48713916-48713916 on Build GRCh38
rs768253167 Chr.19: 48713851-48713851 on Build GRCh38
rs892996664 Chr.19: 48714050-48714050 on Build GRCh38
rs985558348 Chr.19: 48714222-48714222 on Build GRCh38
rs760902003 Chr.19: 48714090-48714090 on Build GRCh38
rs757626386 Chr.19: 48713935-48713935 on Build GRCh38
rs145040442 Chr.19: 48713926-48713926 on Build GRCh38
rs185765112 Chr.19: 48714216-48714216 on Build GRCh38
rs768339055 Chr.19: 48713978-48713978 on Build GRCh38
rs281385 Chr.19: 48714004-48714004 on Build GRCh38
rs577078584 Chr.19: 48714142-48714142 on Build GRCh38
rs940268132 Chr.19: 48713880-48713880 on Build GRCh38
rs1002553283 Chr.19: 48714158-48714158 on Build GRCh38
rs377039948 Chr.19: 48713838-48713838 on Build GRCh38
rs761698803 Chr.19: 48714020-48714020 on Build GRCh38
rs201674816 Chr.19: 48713818-48713818 on Build GRCh38
rs746486281 Chr.19: 48713967-48713967 on Build GRCh38
rs761565363 Chr.19: 48713866-48713866 on Build GRCh38
rs372243558 Chr.19: 48714070-48714070 on Build GRCh38
rs779566328 Chr.19: 48713960-48713960 on Build GRCh38
rs998750493 Chr.19: 48714212-48714212 on Build GRCh38
rs911485343 Chr.19: 48714227-48714227 on Build GRCh38
rs779979772 Chr.19: 48714148-48714148 on Build GRCh38
rs781621130 Chr.19: 48713863-48713864 on Build GRCh38
rs773810533 Chr.19: 48713942-48713959 on Build GRCh38
rs969929604 Chr.19: 48714150-48714150 on Build GRCh38
rs895223115 Chr.19: 48714188-48714188 on Build GRCh38
rs771037686 Chr.19: 48714082-48714082 on Build GRCh38
rs749628996 Chr.19: 48713800-48713800 on Build GRCh38
rs761198388 Chr.19: 48713954-48713977 on Build GRCh38
rs776444780 Chr.19: 48713862-48713862 on Build GRCh38

View Full Product Details