Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48799583-48800071 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAT2
Entrez Gene ID
587
Gene Name
branched chain amino acid transaminase 2
Gene Aliases
BCAM, BCATM, BCT2, PP18
UniGene
Hs.512670
Gene Chromosome Location
Chr.19: 48795062-48811063 on Build GRCh38
Transcripts
NM_001164773.1 NM_001190.3 NM_001284325.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
439926 c.656G>A p.R219Q Chr.19: 48799714-48799714 on Build GRCh38
999084 c.615C>T p.S205S Chr.19: 48799755-48799755 on Build GRCh38
439927 c.616G>A p.V206M Chr.19: 48799754-48799754 on Build GRCh38
999085 c.490G>A p.G164S Chr.19: 48800022-48800022 on Build GRCh38
999086 c.461T>G p.V154G Chr.19: 48800051-48800051 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111322841 Chr.19: 48799612-48799612 on Build GRCh38
rs747075655 Chr.19: 48799701-48799701 on Build GRCh38
rs753566808 Chr.19: 48799946-48799946 on Build GRCh38
rs752745354 Chr.19: 48799671-48799671 on Build GRCh38
rs772311876 Chr.19: 48799695-48799695 on Build GRCh38
rs138273695 Chr.19: 48799923-48799924 on Build GRCh38
rs1033304218 Chr.19: 48799913-48799913 on Build GRCh38
rs754885254 Chr.19: 48799657-48799657 on Build GRCh38
rs79924510 Chr.19: 48799881-48799881 on Build GRCh38
rs1045285691 Chr.19: 48799838-48799838 on Build GRCh38
rs773739531 Chr.19: 48799728-48799728 on Build GRCh38
rs749866079 Chr.19: 48800003-48800003 on Build GRCh38
rs749674051 Chr.19: 48799845-48799845 on Build GRCh38
rs1022339040 Chr.19: 48799968-48799968 on Build GRCh38
rs773653450 Chr.19: 48799840-48799840 on Build GRCh38
rs377190765 Chr.19: 48800023-48800023 on Build GRCh38
rs1010922800 Chr.19: 48799967-48799967 on Build GRCh38
rs199930503 Chr.19: 48799748-48799748 on Build GRCh38
rs911136797 Chr.19: 48799999-48799999 on Build GRCh38
rs528950005 Chr.19: 48799755-48799755 on Build GRCh38
rs377427799 Chr.19: 48799861-48799861 on Build GRCh38
rs983831500 Chr.19: 48800048-48800048 on Build GRCh38
rs777002963 Chr.19: 48799715-48799715 on Build GRCh38
rs756622271 Chr.19: 48799988-48799988 on Build GRCh38
rs767115092 Chr.19: 48799750-48799750 on Build GRCh38
rs762731350 Chr.19: 48799641-48799641 on Build GRCh38
rs202130376 Chr.19: 48799765-48799765 on Build GRCh38
rs765140805 Chr.19: 48799948-48799948 on Build GRCh38
rs1048183673 Chr.19: 48799809-48799809 on Build GRCh38
rs1025165159 Chr.19: 48799966-48799966 on Build GRCh38
rs147045522 Chr.19: 48799712-48799713 on Build GRCh38
rs751697837 Chr.19: 48799955-48799955 on Build GRCh38
rs11548193 Chr.19: 48799813-48799813 on Build GRCh38
rs755257830 Chr.19: 48799957-48799957 on Build GRCh38
rs986571893 Chr.19: 48799984-48799984 on Build GRCh38
rs200989265 Chr.19: 48800019-48800019 on Build GRCh38
rs755892212 Chr.19: 48799678-48799678 on Build GRCh38
rs377598825 Chr.19: 48800013-48800013 on Build GRCh38
rs139804733 Chr.19: 48799731-48799731 on Build GRCh38
rs777505048 Chr.19: 48799767-48799767 on Build GRCh38
rs745470533 Chr.19: 48800014-48800014 on Build GRCh38
rs141873048 Chr.19: 48799683-48799683 on Build GRCh38
rs762276014 Chr.19: 48799622-48799622 on Build GRCh38
rs1012327340 Chr.19: 48799822-48799822 on Build GRCh38
rs747904091 Chr.19: 48799649-48799653 on Build GRCh38
rs771179659 Chr.19: 48800006-48800006 on Build GRCh38
rs768593940 Chr.19: 48799808-48799808 on Build GRCh38
rs780147581 Chr.19: 48799796-48799796 on Build GRCh38
rs753659619 Chr.19: 48799770-48799770 on Build GRCh38
rs764898598 Chr.19: 48800030-48800030 on Build GRCh38
rs753034699 Chr.19: 48800046-48800046 on Build GRCh38
rs201078371 Chr.19: 48799977-48799977 on Build GRCh38
rs11548192 Chr.19: 48799740-48799740 on Build GRCh38
rs765752062 Chr.19: 48799628-48799628 on Build GRCh38
rs975271762 Chr.19: 48799716-48799716 on Build GRCh38
rs751627246 Chr.19: 48799834-48799834 on Build GRCh38
rs762456218 Chr.19: 48799720-48799720 on Build GRCh38
rs769143474 Chr.19: 48799711-48799711 on Build GRCh38
rs751053520 Chr.19: 48799655-48799655 on Build GRCh38
rs757000896 Chr.19: 48799947-48799947 on Build GRCh38
rs780995085 Chr.19: 48799665-48799665 on Build GRCh38
rs750250807 Chr.19: 48800034-48800034 on Build GRCh38
rs866011824 Chr.19: 48799732-48799732 on Build GRCh38
rs781079074 Chr.19: 48799976-48799976 on Build GRCh38
rs778419323 Chr.19: 48799993-48799993 on Build GRCh38
rs760187465 Chr.19: 48800021-48800021 on Build GRCh38
rs765961034 Chr.19: 48799739-48799739 on Build GRCh38
rs11548194 Chr.19: 48799714-48799714 on Build GRCh38
rs755095307 Chr.19: 48800040-48800040 on Build GRCh38
rs1042293172 Chr.19: 48799879-48799879 on Build GRCh38
rs894905096 Chr.19: 48799943-48799943 on Build GRCh38
rs771392617 Chr.19: 48799853-48799853 on Build GRCh38
rs193099355 Chr.19: 48799983-48799983 on Build GRCh38
rs758561917 Chr.19: 48799786-48799786 on Build GRCh38
rs369994559 Chr.19: 48799746-48799746 on Build GRCh38
rs748621319 Chr.19: 48799814-48799814 on Build GRCh38
rs373806842 Chr.19: 48799882-48799882 on Build GRCh38
rs929811487 Chr.19: 48800035-48800035 on Build GRCh38
rs773860470 Chr.19: 48799859-48799859 on Build GRCh38
rs369918856 Chr.19: 48800041-48800041 on Build GRCh38
rs369209966 Chr.19: 48799632-48799632 on Build GRCh38
rs780137162 Chr.19: 48799700-48799700 on Build GRCh38
rs758664912 Chr.19: 48799950-48799950 on Build GRCh38
rs989429849 Chr.19: 48799607-48799607 on Build GRCh38
rs533276867 Chr.19: 48799877-48799877 on Build GRCh38
rs761847809 Chr.19: 48800028-48800028 on Build GRCh38
rs376389720 Chr.19: 48799652-48799652 on Build GRCh38
rs766907460 Chr.19: 48799868-48799868 on Build GRCh38
rs747326261 Chr.19: 48799806-48799806 on Build GRCh38
rs149715085 Chr.19: 48800026-48800026 on Build GRCh38
rs150646107 Chr.19: 48799726-48799726 on Build GRCh38

View Full Product Details