Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48813477-48813899 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSD17B14
Entrez Gene ID
51171
Gene Name
hydroxysteroid 17-beta dehydrogenase 14
Gene Aliases
DHRS10, SDR47C1, retSDR3
UniGene
Hs.18788
Gene Chromosome Location
Chr.19: 48813014-48836688 on Build GRCh38
Transcripts
NM_016246.2 XM_005258969.3 XM_005258970.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6682353 c.520C>T p.P174S Chr.19: 48813685-48813685 on Build GRCh38
4450321 c.543-1G>A p.? Chr.19: 48813553-48813553 on Build GRCh38
7329443 c.577G>T p.E193* Chr.19: 48813518-48813518 on Build GRCh38
4499730 c.548C>T p.S183F Chr.19: 48813547-48813547 on Build GRCh38
5416614 c.553G>A p.G185R Chr.19: 48813542-48813542 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769820095 Chr.19: 48813536-48813536 on Build GRCh38
rs574122888 Chr.19: 48813655-48813655 on Build GRCh38
rs763849664 Chr.19: 48813656-48813656 on Build GRCh38
rs761399964 Chr.19: 48813673-48813673 on Build GRCh38
rs762743428 Chr.19: 48813768-48813768 on Build GRCh38
rs997911792 Chr.19: 48813820-48813820 on Build GRCh38
rs541412439 Chr.19: 48813671-48813671 on Build GRCh38
rs776027257 Chr.19: 48813653-48813653 on Build GRCh38
rs372564188 Chr.19: 48813617-48813617 on Build GRCh38
rs749741184 Chr.19: 48813625-48813625 on Build GRCh38
rs757262802 Chr.19: 48813713-48813713 on Build GRCh38
rs78535883 Chr.19: 48813841-48813841 on Build GRCh38
rs759452711 Chr.19: 48813595-48813595 on Build GRCh38
rs374514062 Chr.19: 48813636-48813636 on Build GRCh38
rs751373181 Chr.19: 48813686-48813686 on Build GRCh38
rs767631684 Chr.19: 48813750-48813750 on Build GRCh38
rs375354399 Chr.19: 48813747-48813747 on Build GRCh38
rs1003695597 Chr.19: 48813705-48813705 on Build GRCh38
rs753402005 Chr.19: 48813592-48813592 on Build GRCh38
rs779944003 Chr.19: 48813619-48813619 on Build GRCh38
rs370106155 Chr.19: 48813767-48813767 on Build GRCh38
rs771957091 Chr.19: 48813520-48813520 on Build GRCh38
rs953586855 Chr.19: 48813798-48813798 on Build GRCh38
rs781263301 Chr.19: 48813526-48813526 on Build GRCh38
rs984340663 Chr.19: 48813610-48813610 on Build GRCh38
rs563318838 Chr.19: 48813681-48813681 on Build GRCh38
rs764383075 Chr.19: 48813860-48813860 on Build GRCh38
rs545979572 Chr.19: 48813757-48813757 on Build GRCh38
rs986154691 Chr.19: 48813809-48813809 on Build GRCh38
rs763656800 Chr.19: 48813591-48813591 on Build GRCh38
rs750438757 Chr.19: 48813618-48813618 on Build GRCh38
rs763387375 Chr.19: 48813549-48813549 on Build GRCh38
rs1018945517 Chr.19: 48813829-48813829 on Build GRCh38
rs775413921 Chr.19: 48813786-48813786 on Build GRCh38
rs774691125 Chr.19: 48813643-48813643 on Build GRCh38
rs138263977 Chr.19: 48813538-48813538 on Build GRCh38
rs759816626 Chr.19: 48813569-48813569 on Build GRCh38
rs375667860 Chr.19: 48813570-48813570 on Build GRCh38
rs769079266 Chr.19: 48813697-48813697 on Build GRCh38
rs564083251 Chr.19: 48813811-48813811 on Build GRCh38
rs745661373 Chr.19: 48813512-48813512 on Build GRCh38
rs760425770 Chr.19: 48813572-48813572 on Build GRCh38
rs772275418 Chr.19: 48813647-48813647 on Build GRCh38
rs558024141 Chr.19: 48813567-48813567 on Build GRCh38
rs977908819 Chr.19: 48813854-48813854 on Build GRCh38
rs764598752 Chr.19: 48813764-48813764 on Build GRCh38
rs766292281 Chr.19: 48813776-48813776 on Build GRCh38
rs149556198 Chr.19: 48813703-48813703 on Build GRCh38
rs375145835 Chr.19: 48813781-48813781 on Build GRCh38
rs748405562 Chr.19: 48813534-48813534 on Build GRCh38
rs748333829 Chr.19: 48813625-48813625 on Build GRCh38
rs1005794481 Chr.19: 48813869-48813869 on Build GRCh38
rs756337553 Chr.19: 48813629-48813629 on Build GRCh38
rs764839668 Chr.19: 48813674-48813674 on Build GRCh38
rs746321048 Chr.19: 48813742-48813742 on Build GRCh38
rs371808321 Chr.19: 48813773-48813773 on Build GRCh38
rs370704792 Chr.19: 48813559-48813559 on Build GRCh38
rs572019970 Chr.19: 48813690-48813690 on Build GRCh38
rs559833225 Chr.19: 48813684-48813684 on Build GRCh38
rs779072754 Chr.19: 48813720-48813720 on Build GRCh38
rs371851745 Chr.19: 48813762-48813762 on Build GRCh38
rs367675186 Chr.19: 48813753-48813753 on Build GRCh38
rs749554564 Chr.19: 48813706-48813706 on Build GRCh38
rs555639599 Chr.19: 48813642-48813642 on Build GRCh38
rs200003607 Chr.19: 48813644-48813644 on Build GRCh38

View Full Product Details