Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48875473-48875992 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP1R15A
Entrez Gene ID
23645
Gene Name
protein phosphatase 1 regulatory subunit 15A
Gene Aliases
GADD34
UniGene
Hs.631593
Gene Chromosome Location
Chr.19: 48872392-48876062 on Build GRCh38
Transcripts
NM_014330.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
163682 c.2024G>C p.*675S Chr.19: 48875972-48875972 on Build GRCh38
1612553 c.1889T>A p.L630H Chr.19: 48875837-48875837 on Build GRCh38
3797353 c.1976C>T p.S659L Chr.19: 48875924-48875924 on Build GRCh38
7306109 c.1992G>A p.A664A Chr.19: 48875940-48875940 on Build GRCh38
3893022 c.1788C>T p.I596I Chr.19: 48875736-48875736 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs77919834 Chr.19: 48875618-48875618 on Build GRCh38
rs774080120 Chr.19: 48875941-48875945 on Build GRCh38
rs747561869 Chr.19: 48875746-48875746 on Build GRCh38
rs767917201 Chr.19: 48875921-48875921 on Build GRCh38
rs748198214 Chr.19: 48875610-48875610 on Build GRCh38
rs927823179 Chr.19: 48875962-48875962 on Build GRCh38
rs771676165 Chr.19: 48875634-48875634 on Build GRCh38
rs540566506 Chr.19: 48875569-48875569 on Build GRCh38
rs548393011 Chr.19: 48875625-48875625 on Build GRCh38
rs500079 Chr.19: 48875737-48875737 on Build GRCh38
rs779098200 Chr.19: 48875759-48875760 on Build GRCh38
rs771308583 Chr.19: 48875924-48875924 on Build GRCh38
rs35007147 Chr.19: 48875920-48875920 on Build GRCh38
rs201219958 Chr.19: 48875609-48875609 on Build GRCh38
rs144725427 Chr.19: 48875741-48875741 on Build GRCh38
rs147256383 Chr.19: 48875731-48875731 on Build GRCh38
rs764387649 Chr.19: 48875720-48875720 on Build GRCh38
rs764179438 Chr.19: 48875783-48875783 on Build GRCh38
rs775150021 Chr.19: 48875635-48875635 on Build GRCh38
rs763906526 Chr.19: 48875940-48875940 on Build GRCh38
rs759405447 Chr.19: 48875770-48875770 on Build GRCh38
rs749232698 Chr.19: 48875760-48875760 on Build GRCh38
rs1009377941 Chr.19: 48875653-48875653 on Build GRCh38
rs758973673 Chr.19: 48875734-48875734 on Build GRCh38
rs780930149 Chr.19: 48875745-48875745 on Build GRCh38
rs755730793 Chr.19: 48875924-48875924 on Build GRCh38
rs929442445 Chr.19: 48875530-48875531 on Build GRCh38
rs758846641 Chr.19: 48875826-48875826 on Build GRCh38
rs957142430 Chr.19: 48875550-48875550 on Build GRCh38
rs970391984 Chr.19: 48875503-48875503 on Build GRCh38
rs772407341 Chr.19: 48875704-48875704 on Build GRCh38
rs566687328 Chr.19: 48875637-48875637 on Build GRCh38
rs906297445 Chr.19: 48875769-48875769 on Build GRCh38
rs770019481 Chr.19: 48875963-48875963 on Build GRCh38
rs147814056 Chr.19: 48875763-48875763 on Build GRCh38
rs370969118 Chr.19: 48875899-48875899 on Build GRCh38
rs777100478 Chr.19: 48875932-48875940 on Build GRCh38
rs186438232 Chr.19: 48875940-48875940 on Build GRCh38
rs757385027 Chr.19: 48875729-48875729 on Build GRCh38
rs759963976 Chr.19: 48875938-48875939 on Build GRCh38
rs7260416 Chr.19: 48875580-48875580 on Build GRCh38
rs201012471 Chr.19: 48875732-48875732 on Build GRCh38
rs755786656 Chr.19: 48875870-48875870 on Build GRCh38
rs200500833 Chr.19: 48875681-48875681 on Build GRCh38
rs755358847 Chr.19: 48875850-48875850 on Build GRCh38
rs1022571033 Chr.19: 48875831-48875831 on Build GRCh38
rs571123568 Chr.19: 48875692-48875692 on Build GRCh38
rs753354896 Chr.19: 48875939-48875939 on Build GRCh38
rs146665963 Chr.19: 48875945-48875945 on Build GRCh38
rs757438814 Chr.19: 48875912-48875912 on Build GRCh38
rs778047173 Chr.19: 48875617-48875617 on Build GRCh38
rs761644106 Chr.19: 48875665-48875665 on Build GRCh38
rs761985375 Chr.19: 48875796-48875796 on Build GRCh38
rs778609437 Chr.19: 48875941-48875941 on Build GRCh38
rs560195365 Chr.19: 48875871-48875871 on Build GRCh38
rs750120273 Chr.19: 48875668-48875668 on Build GRCh38
rs141370460 Chr.19: 48875607-48875607 on Build GRCh38
rs569259259 Chr.19: 48875616-48875616 on Build GRCh38
rs776893878 Chr.19: 48875794-48875794 on Build GRCh38
rs542853891 Chr.19: 48875542-48875542 on Build GRCh38
rs374535887 Chr.19: 48875719-48875719 on Build GRCh38
rs774919285 Chr.19: 48875577-48875577 on Build GRCh38
rs959024859 Chr.19: 48875700-48875700 on Build GRCh38
rs746254526 Chr.19: 48875627-48875627 on Build GRCh38
rs199878289 Chr.19: 48875654-48875654 on Build GRCh38
rs781458075 Chr.19: 48875863-48875863 on Build GRCh38
rs527 Chr.19: 48875925-48875925 on Build GRCh38
rs749055306 Chr.19: 48875662-48875663 on Build GRCh38
rs773446345 Chr.19: 48875969-48875969 on Build GRCh38
rs1050963345 Chr.19: 48875624-48875624 on Build GRCh38
rs148460066 Chr.19: 48875800-48875800 on Build GRCh38
rs748297223 Chr.19: 48875802-48875802 on Build GRCh38
rs368302218 Chr.19: 48875626-48875626 on Build GRCh38
rs771964492 Chr.19: 48875944-48875944 on Build GRCh38
rs577024195 Chr.19: 48875736-48875736 on Build GRCh38
rs752038705 Chr.19: 48875916-48875916 on Build GRCh38
rs775218534 Chr.19: 48875889-48875889 on Build GRCh38
rs906266172 Chr.19: 48875662-48875662 on Build GRCh38
rs139425008 Chr.19: 48875689-48875689 on Build GRCh38
rs561018347 Chr.19: 48875547-48875547 on Build GRCh38
rs779475446 Chr.19: 48875690-48875690 on Build GRCh38
rs760371750 Chr.19: 48875648-48875648 on Build GRCh38
rs751881103 Chr.19: 48875843-48875843 on Build GRCh38
rs373159566 Chr.19: 48875570-48875570 on Build GRCh38
rs752942887 Chr.19: 48875677-48875677 on Build GRCh38
rs756444344 Chr.19: 48875680-48875680 on Build GRCh38
rs746786955 Chr.19: 48875947-48875947 on Build GRCh38
rs753755934 Chr.19: 48875585-48875585 on Build GRCh38
rs371089180 Chr.19: 48875666-48875666 on Build GRCh38
rs909781098 Chr.19: 48875749-48875749 on Build GRCh38
rs767212164 Chr.19: 48875571-48875571 on Build GRCh38
rs764939326 Chr.19: 48875597-48875597 on Build GRCh38
rs2270946 Chr.19: 48875728-48875728 on Build GRCh38
rs772078537 Chr.19: 48875858-48875865 on Build GRCh38
rs528417901 Chr.19: 48875560-48875560 on Build GRCh38
rs749614844 Chr.19: 48875568-48875568 on Build GRCh38
rs903684196 Chr.19: 48875762-48875762 on Build GRCh38
rs953578810 Chr.19: 48875837-48875837 on Build GRCh38
rs763051525 Chr.19: 48875914-48875914 on Build GRCh38
rs373351648 Chr.19: 48875867-48875867 on Build GRCh38
rs372233957 Chr.19: 48875751-48875751 on Build GRCh38
rs745605469 Chr.19: 48875886-48875886 on Build GRCh38
rs764242076 Chr.19: 48875885-48875885 on Build GRCh38
rs766823908 Chr.19: 48875915-48875915 on Build GRCh38
rs773610977 Chr.19: 48875877-48875877 on Build GRCh38
rs776273456 Chr.19: 48875664-48875664 on Build GRCh38
rs761391876 Chr.19: 48875711-48875711 on Build GRCh38
rs548313458 Chr.19: 48875806-48875806 on Build GRCh38
rs756283354 Chr.19: 48875615-48875615 on Build GRCh38
rs769579706 Chr.19: 48875713-48875713 on Build GRCh38
rs763974981 Chr.19: 48875582-48875582 on Build GRCh38
rs755244404 Chr.19: 48875726-48875740 on Build GRCh38
rs897075419 Chr.19: 48875606-48875606 on Build GRCh38
rs747507817 Chr.19: 48875923-48875925 on Build GRCh38
rs766877382 Chr.19: 48875828-48875828 on Build GRCh38
rs769325327 Chr.19: 48875747-48875747 on Build GRCh38
rs1031267425 Chr.19: 48875548-48875548 on Build GRCh38
rs773886388 Chr.19: 48875563-48875563 on Build GRCh38
rs524 Chr.19: 48875910-48875910 on Build GRCh38
rs1031045647 Chr.19: 48875667-48875667 on Build GRCh38
rs386810065 Chr.19: 48875910-48875925 on Build GRCh38
rs575250363 Chr.19: 48875714-48875714 on Build GRCh38
rs142378074 Chr.19: 48875798-48875798 on Build GRCh38
rs146849578 Chr.19: 48875801-48875801 on Build GRCh38
rs536325560 Chr.19: 48875716-48875716 on Build GRCh38
rs111550347 Chr.19: 48875611-48875611 on Build GRCh38
rs915452570 Chr.19: 48875561-48875561 on Build GRCh38
rs202191597 Chr.19: 48875888-48875888 on Build GRCh38
rs556876971 Chr.19: 48875708-48875708 on Build GRCh38
rs1016548847 Chr.19: 48875839-48875839 on Build GRCh38
rs538298636 Chr.19: 48875707-48875707 on Build GRCh38
rs982163017 Chr.19: 48875537-48875537 on Build GRCh38
rs944707119 Chr.19: 48875596-48875596 on Build GRCh38
rs918204537 Chr.19: 48875505-48875505 on Build GRCh38

View Full Product Details