Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48961445-48961948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAX
Entrez Gene ID
581
Gene Name
BCL2 associated X, apoptosis regulator
Gene Aliases
BCL2L4
UniGene
Hs.624291
Gene Chromosome Location
Chr.19: 48954825-48961798 on Build GRCh38
Transcripts
NM_001291428.1 NM_001291431.1 NM_138761.3 NM_138763.3 NM_138764.4 NR_027882.1 XM_017027077.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7328210 c.575G>A p.G192D Chr.19: 48961632-48961632 on Build GRCh38
999162 c.484C>A p.L162I Chr.19: 48961541-48961541 on Build GRCh38
4806893 c.569A>G p.K190R Chr.19: 48961626-48961626 on Build GRCh38
4079954 c.533C>T p.A178V Chr.19: 48961590-48961590 on Build GRCh38
4285202 c.539T>G p.V180G Chr.19: 48961596-48961596 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530189252 Chr.19: 48961468-48961468 on Build GRCh38
rs1056271201 Chr.19: 48961479-48961479 on Build GRCh38
rs548474023 Chr.19: 48961502-48961502 on Build GRCh38
rs76091384 Chr.19: 48961907-48961907 on Build GRCh38
rs771956472 Chr.19: 48961564-48961564 on Build GRCh38
rs775345231 Chr.19: 48961570-48961570 on Build GRCh38
rs760063736 Chr.19: 48961596-48961596 on Build GRCh38
rs370817149 Chr.19: 48961627-48961627 on Build GRCh38
rs751630532 Chr.19: 48961594-48961594 on Build GRCh38
rs1009698093 Chr.19: 48961921-48961921 on Build GRCh38
rs983493121 Chr.19: 48961579-48961579 on Build GRCh38
rs917770251 Chr.19: 48961886-48961886 on Build GRCh38
rs566817615 Chr.19: 48961683-48961683 on Build GRCh38
rs964963858 Chr.19: 48961561-48961561 on Build GRCh38
rs112938655 Chr.19: 48961837-48961837 on Build GRCh38
rs745430809 Chr.19: 48961662-48961662 on Build GRCh38
rs908300907 Chr.19: 48961871-48961871 on Build GRCh38
rs937148426 Chr.19: 48961467-48961467 on Build GRCh38
rs377294979 Chr.19: 48961574-48961574 on Build GRCh38
rs909320036 Chr.19: 48961867-48961867 on Build GRCh38
rs778548071 Chr.19: 48961660-48961660 on Build GRCh38
rs766476483 Chr.19: 48961591-48961591 on Build GRCh38
rs768088908 Chr.19: 48961649-48961649 on Build GRCh38
rs577134354 Chr.19: 48961885-48961885 on Build GRCh38
rs773126142 Chr.19: 48961650-48961650 on Build GRCh38
rs556985719 Chr.19: 48961784-48961784 on Build GRCh38
rs778771537 Chr.19: 48961549-48961550 on Build GRCh38
rs572099252 Chr.19: 48961830-48961830 on Build GRCh38
rs766694177 Chr.19: 48961548-48961561 on Build GRCh38
rs558629161 Chr.19: 48961791-48961791 on Build GRCh38
rs1042218283 Chr.19: 48961911-48961911 on Build GRCh38
rs980080128 Chr.19: 48961875-48961875 on Build GRCh38
rs991326265 Chr.19: 48961831-48961831 on Build GRCh38
rs766781664 Chr.19: 48961753-48961753 on Build GRCh38
rs373493221 Chr.19: 48961558-48961558 on Build GRCh38
rs772045059 Chr.19: 48961667-48961667 on Build GRCh38
rs762967671 Chr.19: 48961582-48961582 on Build GRCh38
rs757014739 Chr.19: 48961652-48961652 on Build GRCh38
rs1020903317 Chr.19: 48961793-48961793 on Build GRCh38
rs937908625 Chr.19: 48961717-48961717 on Build GRCh38
rs1057369 Chr.19: 48961609-48961609 on Build GRCh38
rs750039474 Chr.19: 48961749-48961749 on Build GRCh38
rs373416805 Chr.19: 48961754-48961754 on Build GRCh38
rs765437837 Chr.19: 48961487-48961487 on Build GRCh38
rs748508290 Chr.19: 48961520-48961520 on Build GRCh38
rs1030951484 Chr.19: 48961843-48961843 on Build GRCh38
rs36101119 Chr.19: 48961866-48961866 on Build GRCh38
rs903614745 Chr.19: 48961928-48961928 on Build GRCh38
rs534360637 Chr.19: 48961704-48961704 on Build GRCh38
rs1002464330 Chr.19: 48961466-48961466 on Build GRCh38
rs781589645 Chr.19: 48961511-48961511 on Build GRCh38
rs1042145584 Chr.19: 48961802-48961802 on Build GRCh38
rs751106339 Chr.19: 48961874-48961874 on Build GRCh38
rs950899868 Chr.19: 48961854-48961854 on Build GRCh38
rs997529866 Chr.19: 48961887-48961887 on Build GRCh38
rs704243 Chr.19: 48961714-48961714 on Build GRCh38
rs777518890 Chr.19: 48961648-48961648 on Build GRCh38
rs117098111 Chr.19: 48961803-48961803 on Build GRCh38
rs926553930 Chr.19: 48961707-48961707 on Build GRCh38
rs955465774 Chr.19: 48961752-48961752 on Build GRCh38
rs909246907 Chr.19: 48961580-48961580 on Build GRCh38
rs1020669707 Chr.19: 48961510-48961510 on Build GRCh38
rs750494572 Chr.19: 48961494-48961494 on Build GRCh38
rs758475653 Chr.19: 48961495-48961495 on Build GRCh38
rs536406169 Chr.19: 48961790-48961790 on Build GRCh38
rs770587887 Chr.19: 48961535-48961535 on Build GRCh38
rs369789011 Chr.19: 48961552-48961552 on Build GRCh38
rs755777499 Chr.19: 48961524-48961524 on Build GRCh38
rs976666767 Chr.19: 48961563-48961563 on Build GRCh38
rs760456972 Chr.19: 48961573-48961573 on Build GRCh38
rs146366710 Chr.19: 48961534-48961534 on Build GRCh38
rs925867726 Chr.19: 48961827-48961827 on Build GRCh38
rs766809725 Chr.19: 48961497-48961497 on Build GRCh38
rs767966452 Chr.19: 48961603-48961603 on Build GRCh38
rs748073277 Chr.19: 48961621-48961623 on Build GRCh38
rs1044350955 Chr.19: 48961679-48961679 on Build GRCh38
rs546778551 Chr.19: 48961748-48961748 on Build GRCh38
rs369285586 Chr.19: 48961557-48961557 on Build GRCh38
rs575358546 Chr.19: 48961786-48961786 on Build GRCh38
rs983524490 Chr.19: 48961865-48961865 on Build GRCh38
rs937963038 Chr.19: 48961884-48961884 on Build GRCh38
rs1000548611 Chr.19: 48961828-48961828 on Build GRCh38
rs753039973 Chr.19: 48961618-48961618 on Build GRCh38
rs558702576 Chr.19: 48961883-48961883 on Build GRCh38
rs997777413 Chr.19: 48961838-48961838 on Build GRCh38
rs546174884 Chr.19: 48961705-48961705 on Build GRCh38
rs777573936 Chr.19: 48961531-48961531 on Build GRCh38

View Full Product Details