Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49015717-49016208 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LHB
Entrez Gene ID
3972
Gene Name
luteinizing hormone beta polypeptide
Gene Aliases
CGB4, HH23, LSH-B, LSH-beta
UniGene
Hs.154704
Gene Chromosome Location
Chr.19: 49015980-49017090 on Build GRCh38
Transcripts
NM_000894.2 XM_011526975.1 NM_001321190.1 NM_001321191.1 NM_006666.2 NR_135578.1 XM_011526330.1

Gene Symbol
RUVBL2
Entrez Gene ID
10856
Gene Name
RuvB like AAA ATPase 2
Gene Aliases
CGI-46, ECP-51, ECP51, INO80J, REPTIN, RVB2, TAP54-beta, TIH2, TIP48, TIP49B
UniGene
Hs.515846
Gene Chromosome Location
Chr.19: 48993448-49015995 on Build GRCh38
Transcripts
NM_000894.2 XM_011526975.1 NM_001321190.1 NM_001321191.1 NM_006666.2 NR_135578.1 XM_011526330.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5056814 c.354T>G p.S118S Chr.19: 49016140-49016140 on Build GRCh38
5499378 c.1372G>A p.E458K Chr.19: 49015822-49015822 on Build GRCh38
4920936 c.343C>G p.R115G Chr.19: 49016151-49016151 on Build GRCh38
7322879 c.407C>T p.S136L Chr.19: 49016087-49016087 on Build GRCh38
999186 c.294C>A p.P98P Chr.19: 49016200-49016200 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758865199 Chr.19: 49016000-49016000 on Build GRCh38
rs763016665 Chr.19: 49016153-49016153 on Build GRCh38
rs780562920 Chr.19: 49016009-49016009 on Build GRCh38
rs746347011 Chr.19: 49015998-49015998 on Build GRCh38
rs769154319 Chr.19: 49016018-49016018 on Build GRCh38
rs200647471 Chr.19: 49015783-49015783 on Build GRCh38
rs746584185 Chr.19: 49016077-49016079 on Build GRCh38
rs747407636 Chr.19: 49015951-49015951 on Build GRCh38
rs771737595 Chr.19: 49016140-49016140 on Build GRCh38
rs920437883 Chr.19: 49016054-49016054 on Build GRCh38
rs370430354 Chr.19: 49015815-49015815 on Build GRCh38
rs201720780 Chr.19: 49015829-49015829 on Build GRCh38
rs774432143 Chr.19: 49016035-49016035 on Build GRCh38
rs757996082 Chr.19: 49015835-49015837 on Build GRCh38
rs772806540 Chr.19: 49016024-49016024 on Build GRCh38
rs781649328 Chr.19: 49016101-49016101 on Build GRCh38
rs867597840 Chr.19: 49016121-49016121 on Build GRCh38
rs778081352 Chr.19: 49015865-49015865 on Build GRCh38
rs944338805 Chr.19: 49015891-49015891 on Build GRCh38
rs200507790 Chr.19: 49015985-49015985 on Build GRCh38
rs141729250 Chr.19: 49016014-49016014 on Build GRCh38
rs780045417 Chr.19: 49015859-49015859 on Build GRCh38
rs903471820 Chr.19: 49015940-49015940 on Build GRCh38
rs779016630 Chr.19: 49015994-49015994 on Build GRCh38
rs761161758 Chr.19: 49015967-49015967 on Build GRCh38
rs760411948 Chr.19: 49016146-49016146 on Build GRCh38
rs777561425 Chr.19: 49016168-49016168 on Build GRCh38
rs569880835 Chr.19: 49015949-49015949 on Build GRCh38
rs189656791 Chr.19: 49016012-49016012 on Build GRCh38
rs777337615 Chr.19: 49016013-49016013 on Build GRCh38
rs558017198 Chr.19: 49015856-49015856 on Build GRCh38
rs371692563 Chr.19: 49016159-49016159 on Build GRCh38
rs199984329 Chr.19: 49015798-49015798 on Build GRCh38
rs150244609 Chr.19: 49016079-49016079 on Build GRCh38
rs376444705 Chr.19: 49016162-49016162 on Build GRCh38
rs757275035 Chr.19: 49015986-49015986 on Build GRCh38
rs563409995 Chr.19: 49016068-49016068 on Build GRCh38
rs545246639 Chr.19: 49015751-49015751 on Build GRCh38
rs370781314 Chr.19: 49016182-49016182 on Build GRCh38
rs370235131 Chr.19: 49015782-49015782 on Build GRCh38
rs373012966 Chr.19: 49015892-49015892 on Build GRCh38
rs759571421 Chr.19: 49016134-49016134 on Build GRCh38
rs765025780 Chr.19: 49015849-49015849 on Build GRCh38
rs374676929 Chr.19: 49016057-49016057 on Build GRCh38
rs111564806 Chr.19: 49015860-49015860 on Build GRCh38
rs762509063 Chr.19: 49015957-49015957 on Build GRCh38
rs758281537 Chr.19: 49015767-49015767 on Build GRCh38
rs777579855 Chr.19: 49016127-49016127 on Build GRCh38
rs768146510 Chr.19: 49015956-49015956 on Build GRCh38
rs373687298 Chr.19: 49015787-49015787 on Build GRCh38
rs760365712 Chr.19: 49015802-49015802 on Build GRCh38
rs367773986 Chr.19: 49015835-49015835 on Build GRCh38
rs770410549 Chr.19: 49016140-49016141 on Build GRCh38
rs749776 Chr.19: 49015801-49015801 on Build GRCh38
rs775318829 Chr.19: 49016142-49016142 on Build GRCh38
rs766358514 Chr.19: 49016033-49016033 on Build GRCh38
rs764859975 Chr.19: 49015910-49015910 on Build GRCh38
rs773260282 Chr.19: 49015972-49015972 on Build GRCh38
rs2013039 Chr.19: 49015973-49015973 on Build GRCh38
rs549429148 Chr.19: 49015756-49015756 on Build GRCh38
rs2013040 Chr.19: 49015983-49015983 on Build GRCh38
rs778529225 Chr.19: 49016181-49016181 on Build GRCh38
rs755285445 Chr.19: 49016100-49016100 on Build GRCh38
rs368190391 Chr.19: 49016108-49016108 on Build GRCh38
rs200045996 Chr.19: 49016036-49016036 on Build GRCh38
rs775870202 Chr.19: 49015950-49015950 on Build GRCh38
rs563521941 Chr.19: 49015753-49015753 on Build GRCh38
rs749003379 Chr.19: 49016175-49016175 on Build GRCh38
rs763625411 Chr.19: 49015806-49015806 on Build GRCh38
rs138913384 Chr.19: 49016155-49016155 on Build GRCh38
rs781570619 Chr.19: 49016160-49016160 on Build GRCh38
rs1012974032 Chr.19: 49015974-49015974 on Build GRCh38
rs184781114 Chr.19: 49015984-49015984 on Build GRCh38
rs771999985 Chr.19: 49016183-49016183 on Build GRCh38
rs201749590 Chr.19: 49016151-49016151 on Build GRCh38
rs533242738 Chr.19: 49015741-49015741 on Build GRCh38
rs762572085 Chr.19: 49016031-49016031 on Build GRCh38
rs201196194 Chr.19: 49016066-49016066 on Build GRCh38
rs113453475 Chr.19: 49015927-49015927 on Build GRCh38
rs771296389 Chr.19: 49015791-49015791 on Build GRCh38
rs746428805 Chr.19: 49015894-49015894 on Build GRCh38
rs749272679 Chr.19: 49015880-49015880 on Build GRCh38
rs575749991 Chr.19: 49016125-49016125 on Build GRCh38
rs756253228 Chr.19: 49015863-49015863 on Build GRCh38
rs528776938 Chr.19: 49016154-49016154 on Build GRCh38
rs761406558 Chr.19: 49015846-49015846 on Build GRCh38
rs770539036 Chr.19: 49016176-49016176 on Build GRCh38
rs756549072 Chr.19: 49016167-49016167 on Build GRCh38
rs369933565 Chr.19: 49015773-49015773 on Build GRCh38
rs747619934 Chr.19: 49016099-49016099 on Build GRCh38
rs774939996 Chr.19: 49015792-49015792 on Build GRCh38
rs765387170 Chr.19: 49016150-49016150 on Build GRCh38
rs751348372 Chr.19: 49015982-49015982 on Build GRCh38
rs780310489 Chr.19: 49015800-49015802 on Build GRCh38
rs764860251 Chr.19: 49015968-49015968 on Build GRCh38
rs371411755 Chr.19: 49016056-49016056 on Build GRCh38
rs758255007 Chr.19: 49015857-49015857 on Build GRCh38
rs531145535 Chr.19: 49015754-49015754 on Build GRCh38
rs5030774 Chr.19: 49016130-49016130 on Build GRCh38
rs201570299 Chr.19: 49015888-49015888 on Build GRCh38
rs747317735 Chr.19: 49016013-49016013 on Build GRCh38
rs779287504 Chr.19: 49015893-49015893 on Build GRCh38
rs759832019 Chr.19: 49015794-49015794 on Build GRCh38
rs533700885 Chr.19: 49015924-49015924 on Build GRCh38
rs754218231 Chr.19: 49015809-49015809 on Build GRCh38
rs765477616 Chr.19: 49016064-49016064 on Build GRCh38

View Full Product Details