Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49063649-49064142 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NTF4
Entrez Gene ID
4909
Gene Name
neurotrophin 4
Gene Aliases
GLC10, GLC1O, NT-4, NT-4/5, NT-5, NT4, NT5, NTF5
UniGene
Hs.266902
Gene Chromosome Location
Chr.19: 49058284-49065076 on Build GRCh38
Transcripts
NM_006179.4 XM_005258962.3 XM_006723232.3 XM_011527009.2 XM_011527010.2

SNP Detail

SNP ID Chromosome Location
rs373084816 Chr.19: 49063898-49063899 on Build GRCh38
rs906604493 Chr.19: 49063917-49063917 on Build GRCh38
rs953659043 Chr.19: 49063908-49063908 on Build GRCh38
rs183084555 Chr.19: 49064088-49064088 on Build GRCh38
rs888259862 Chr.19: 49063754-49063754 on Build GRCh38
rs533326471 Chr.19: 49063722-49063722 on Build GRCh38
rs898814008 Chr.19: 49063791-49063792 on Build GRCh38
rs993608946 Chr.19: 49063906-49063906 on Build GRCh38
rs772745228 Chr.19: 49063678-49063678 on Build GRCh38
rs890685028 Chr.19: 49063797-49063797 on Build GRCh38
rs3810184 Chr.19: 49064035-49064035 on Build GRCh38
rs986266560 Chr.19: 49064000-49064000 on Build GRCh38
rs991124053 Chr.19: 49063727-49063727 on Build GRCh38
rs1050061904 Chr.19: 49064063-49064063 on Build GRCh38
rs531235522 Chr.19: 49063893-49063893 on Build GRCh38
rs866170466 Chr.19: 49063792-49063792 on Build GRCh38
rs1022484371 Chr.19: 49063894-49063894 on Build GRCh38
rs770847355 Chr.19: 49063738-49063738 on Build GRCh38
rs372197508 Chr.19: 49063831-49063831 on Build GRCh38
rs966059121 Chr.19: 49064014-49064014 on Build GRCh38
rs773385643 Chr.19: 49063873-49063873 on Build GRCh38
rs1037406368 Chr.19: 49063785-49063785 on Build GRCh38
rs932276297 Chr.19: 49063791-49063791 on Build GRCh38
rs1045209995 Chr.19: 49063827-49063827 on Build GRCh38
rs970579745 Chr.19: 49063981-49063981 on Build GRCh38
rs534407500 Chr.19: 49064109-49064109 on Build GRCh38
rs991476415 Chr.19: 49063883-49063883 on Build GRCh38
rs375215891 Chr.19: 49063901-49063901 on Build GRCh38
rs1020460293 Chr.19: 49063814-49063814 on Build GRCh38
rs565881813 Chr.19: 49064060-49064060 on Build GRCh38
rs541313373 Chr.19: 49063681-49063681 on Build GRCh38
rs906237902 Chr.19: 49063830-49063830 on Build GRCh38
rs984432800 Chr.19: 49064061-49064061 on Build GRCh38
rs552069291 Chr.19: 49063723-49063723 on Build GRCh38
rs912153705 Chr.19: 49064064-49064064 on Build GRCh38
rs981876397 Chr.19: 49063760-49063760 on Build GRCh38
rs567040169 Chr.19: 49064021-49064021 on Build GRCh38
rs548912973 Chr.19: 49063954-49063954 on Build GRCh38
rs1009079620 Chr.19: 49063804-49063804 on Build GRCh38
rs1018981498 Chr.19: 49064011-49064011 on Build GRCh38
rs559902354 Chr.19: 49063697-49063697 on Build GRCh38
rs1044865890 Chr.19: 49064099-49064099 on Build GRCh38
rs539228139 Chr.19: 49064086-49064086 on Build GRCh38
rs1011551785 Chr.19: 49063878-49063878 on Build GRCh38
rs949234097 Chr.19: 49063756-49063756 on Build GRCh38
rs553545465 Chr.19: 49064056-49064056 on Build GRCh38
rs34619524 Chr.19: 49064119-49064119 on Build GRCh38
rs1023516276 Chr.19: 49063948-49063948 on Build GRCh38
rs892737263 Chr.19: 49063869-49063869 on Build GRCh38
rs947878382 Chr.19: 49064091-49064091 on Build GRCh38
rs549351004 Chr.19: 49063982-49063982 on Build GRCh38
rs1030638911 Chr.19: 49063847-49063847 on Build GRCh38
rs981950311 Chr.19: 49063988-49063988 on Build GRCh38
rs190519951 Chr.19: 49064037-49064037 on Build GRCh38
rs1003212973 Chr.19: 49063840-49063840 on Build GRCh38
rs976675082 Chr.19: 49063707-49063707 on Build GRCh38
rs1054960728 Chr.19: 49064118-49064118 on Build GRCh38
rs940442633 Chr.19: 49063775-49063775 on Build GRCh38
rs1025087547 Chr.19: 49063912-49063912 on Build GRCh38
rs1000344385 Chr.19: 49064112-49064112 on Build GRCh38
rs947861171 Chr.19: 49063761-49063761 on Build GRCh38

View Full Product Details