Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49433734-49434148 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC17A7
Entrez Gene ID
57030
Gene Name
solute carrier family 17 member 7
Gene Aliases
BNPI, VGLUT1
UniGene
Hs.375616
Gene Chromosome Location
Chr.19: 49429398-49441551 on Build GRCh38
Transcripts
NM_020309.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4956148 c.724G>A p.G242S Chr.19: 49433960-49433960 on Build GRCh38
1225979 c.839A>G p.E280G Chr.19: 49433754-49433754 on Build GRCh38
999294 c.826G>A p.D276N Chr.19: 49433767-49433767 on Build GRCh38
6779919 c.782C>T p.A261V Chr.19: 49433811-49433811 on Build GRCh38
999295 c.688C>T p.Q230* Chr.19: 49433996-49433996 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755280190 Chr.19: 49434098-49434098 on Build GRCh38
rs778846385 Chr.19: 49433961-49433961 on Build GRCh38
rs754527729 Chr.19: 49433873-49433873 on Build GRCh38
rs773478101 Chr.19: 49433902-49433902 on Build GRCh38
rs1001025599 Chr.19: 49434116-49434116 on Build GRCh38
rs376116312 Chr.19: 49434075-49434075 on Build GRCh38
rs759194899 Chr.19: 49434052-49434052 on Build GRCh38
rs773814218 Chr.19: 49434022-49434022 on Build GRCh38
rs754254813 Chr.19: 49433937-49433937 on Build GRCh38
rs756038458 Chr.19: 49433943-49433943 on Build GRCh38
rs763482389 Chr.19: 49434083-49434083 on Build GRCh38
rs1024979174 Chr.19: 49434104-49434104 on Build GRCh38
rs768940081 Chr.19: 49433974-49433974 on Build GRCh38
rs764247943 Chr.19: 49433799-49433799 on Build GRCh38
rs756628316 Chr.19: 49433805-49433806 on Build GRCh38
rs971488971 Chr.19: 49433804-49433804 on Build GRCh38
rs373252240 Chr.19: 49433991-49433991 on Build GRCh38
rs775745022 Chr.19: 49433764-49433764 on Build GRCh38
rs199650774 Chr.19: 49434059-49434059 on Build GRCh38
rs141159987 Chr.19: 49433813-49433813 on Build GRCh38
rs752187419 Chr.19: 49433934-49433934 on Build GRCh38
rs757988017 Chr.19: 49433831-49433831 on Build GRCh38
rs370635021 Chr.19: 49433891-49433891 on Build GRCh38
rs767030567 Chr.19: 49433933-49433933 on Build GRCh38
rs763324674 Chr.19: 49433907-49433907 on Build GRCh38
rs1002640850 Chr.19: 49433963-49433963 on Build GRCh38
rs369525295 Chr.19: 49433932-49433932 on Build GRCh38
rs971665540 Chr.19: 49434115-49434115 on Build GRCh38
rs766859282 Chr.19: 49433870-49433871 on Build GRCh38
rs748216851 Chr.19: 49434005-49434005 on Build GRCh38
rs778864484 Chr.19: 49434093-49434093 on Build GRCh38
rs773049247 Chr.19: 49433759-49433759 on Build GRCh38
rs199595597 Chr.19: 49433760-49433760 on Build GRCh38
rs201240445 Chr.19: 49434051-49434051 on Build GRCh38
rs375509968 Chr.19: 49433821-49433821 on Build GRCh38
rs904043001 Chr.19: 49434095-49434095 on Build GRCh38
rs747202802 Chr.19: 49433890-49433890 on Build GRCh38
rs533858610 Chr.19: 49433967-49433967 on Build GRCh38
rs776852602 Chr.19: 49433986-49433986 on Build GRCh38
rs757247344 Chr.19: 49434085-49434085 on Build GRCh38
rs909051120 Chr.19: 49433872-49433872 on Build GRCh38
rs373858112 Chr.19: 49434006-49434006 on Build GRCh38
rs777718517 Chr.19: 49433946-49433946 on Build GRCh38
rs780679514 Chr.19: 49433875-49433875 on Build GRCh38
rs200629513 Chr.19: 49434053-49434053 on Build GRCh38
rs932475856 Chr.19: 49433911-49433911 on Build GRCh38
rs374850701 Chr.19: 49434017-49434017 on Build GRCh38
rs73931785 Chr.19: 49434120-49434120 on Build GRCh38
rs35661101 Chr.19: 49434120-49434120 on Build GRCh38
rs566161964 Chr.19: 49433830-49433830 on Build GRCh38
rs763333486 Chr.19: 49434027-49434027 on Build GRCh38
rs760789528 Chr.19: 49433792-49433792 on Build GRCh38
rs775904533 Chr.19: 49433885-49433885 on Build GRCh38
rs766791421 Chr.19: 49434030-49434030 on Build GRCh38
rs751087475 Chr.19: 49433865-49433865 on Build GRCh38
rs776836687 Chr.19: 49433894-49433894 on Build GRCh38
rs777436545 Chr.19: 49433878-49433878 on Build GRCh38
rs535780422 Chr.19: 49433782-49433782 on Build GRCh38
rs748145694 Chr.19: 49433877-49433877 on Build GRCh38
rs772207811 Chr.19: 49433884-49433884 on Build GRCh38
rs754436792 Chr.19: 49433811-49433811 on Build GRCh38
rs749159335 Chr.19: 49433951-49433951 on Build GRCh38
rs762186041 Chr.19: 49434001-49434001 on Build GRCh38
rs975754458 Chr.19: 49433777-49433777 on Build GRCh38
rs147368622 Chr.19: 49433863-49433863 on Build GRCh38
rs751283629 Chr.19: 49433920-49433920 on Build GRCh38
rs754615719 Chr.19: 49433924-49433924 on Build GRCh38
rs1041690920 Chr.19: 49434002-49434002 on Build GRCh38
rs749073234 Chr.19: 49433879-49433879 on Build GRCh38
rs753880830 Chr.19: 49434084-49434084 on Build GRCh38
rs200232443 Chr.19: 49433908-49433908 on Build GRCh38

View Full Product Details