Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49854382-49854875 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR4749
Entrez Gene ID
100616313
Gene Name
microRNA 4749
Gene Aliases
mir-4749
UniGene
Gene Chromosome Location
Chr.19: 49854591-49854651 on Build GRCh38
Transcripts
NR_039904.2 NM_001305105.1 NM_001305108.1 NM_017432.4 NR_130963.1 XM_005258998.4 XM_006723244.3 XM_011527033.1 XM_011527034.2 XM_011527035.1 XM_017026879.1 XM_017026880.1

Gene Symbol
PTOV1
Entrez Gene ID
53635
Gene Name
prostate tumor overexpressed 1
Gene Aliases
ACID2, PTOV-1
UniGene
Hs.587979
Gene Chromosome Location
Chr.19: 49850726-49860744 on Build GRCh38
Transcripts
NR_039904.2 NM_001305105.1 NM_001305108.1 NM_017432.4 NR_130963.1 XM_005258998.4 XM_006723244.3 XM_011527033.1 XM_011527034.2 XM_011527035.1 XM_017026879.1 XM_017026880.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4718881 c.182G>A p.R61Q Chr.19: 49854416-49854416 on Build GRCh38
5365383 c.93C>A p.F31L Chr.19: 49854423-49854423 on Build GRCh38
4285471 c.196G>A p.V66I Chr.19: 49854526-49854526 on Build GRCh38
6758760 c.94G>A p.G32R Chr.19: 49854424-49854424 on Build GRCh38
6198132 c.221C>T p.P74L Chr.19: 49854455-49854455 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149977568 Chr.19: 49854726-49854726 on Build GRCh38
rs765021487 Chr.19: 49854827-49854827 on Build GRCh38
rs779795928 Chr.19: 49854465-49854465 on Build GRCh38
rs770355657 Chr.19: 49854594-49854594 on Build GRCh38
rs533426566 Chr.19: 49854732-49854732 on Build GRCh38
rs750019888 Chr.19: 49854714-49854714 on Build GRCh38
rs761684383 Chr.19: 49854826-49854826 on Build GRCh38
rs373185339 Chr.19: 49854532-49854532 on Build GRCh38
rs778080118 Chr.19: 49854780-49854780 on Build GRCh38
rs747628312 Chr.19: 49854478-49854478 on Build GRCh38
rs776045338 Chr.19: 49854439-49854439 on Build GRCh38
rs754734913 Chr.19: 49854540-49854540 on Build GRCh38
rs946785560 Chr.19: 49854522-49854522 on Build GRCh38
rs368435954 Chr.19: 49854751-49854751 on Build GRCh38
rs769431489 Chr.19: 49854416-49854416 on Build GRCh38
rs745679865 Chr.19: 49854807-49854807 on Build GRCh38
rs760556849 Chr.19: 49854691-49854691 on Build GRCh38
rs766648202 Chr.19: 49854531-49854531 on Build GRCh38
rs201103722 Chr.19: 49854647-49854647 on Build GRCh38
rs1011867307 Chr.19: 49854504-49854504 on Build GRCh38
rs200363426 Chr.19: 49854469-49854469 on Build GRCh38
rs371913757 Chr.19: 49854406-49854406 on Build GRCh38
rs747328391 Chr.19: 49854582-49854582 on Build GRCh38
rs748925811 Chr.19: 49854486-49854486 on Build GRCh38
rs368120925 Chr.19: 49854818-49854818 on Build GRCh38
rs552910419 Chr.19: 49854596-49854596 on Build GRCh38
rs764000568 Chr.19: 49854749-49854749 on Build GRCh38
rs756168767 Chr.19: 49854545-49854545 on Build GRCh38
rs767922140 Chr.19: 49854567-49854567 on Build GRCh38
rs202190749 Chr.19: 49854722-49854722 on Build GRCh38
rs776558199 Chr.19: 49854824-49854824 on Build GRCh38
rs570340094 Chr.19: 49854636-49854636 on Build GRCh38
rs748573000 Chr.19: 49854644-49854644 on Build GRCh38
rs531528682 Chr.19: 49854493-49854493 on Build GRCh38
rs773885148 Chr.19: 49854578-49854578 on Build GRCh38
rs374463962 Chr.19: 49854727-49854727 on Build GRCh38
rs778438436 Chr.19: 49854643-49854643 on Build GRCh38
rs749720183 Chr.19: 49854788-49854788 on Build GRCh38
rs200987923 Chr.19: 49854446-49854446 on Build GRCh38
rs115799833 Chr.19: 49854743-49854743 on Build GRCh38
rs757689560 Chr.19: 49854790-49854790 on Build GRCh38
rs1035289193 Chr.19: 49854696-49854696 on Build GRCh38
rs751109977 Chr.19: 49854852-49854852 on Build GRCh38
rs745865849 Chr.19: 49854814-49854814 on Build GRCh38
rs750779261 Chr.19: 49854578-49854582 on Build GRCh38
rs993083687 Chr.19: 49854575-49854575 on Build GRCh38
rs1053562233 Chr.19: 49854804-49854804 on Build GRCh38
rs1049733815 Chr.19: 49854571-49854571 on Build GRCh38
rs777386239 Chr.19: 49854563-49854563 on Build GRCh38
rs778260462 Chr.19: 49854711-49854711 on Build GRCh38
rs761212020 Chr.19: 49854441-49854441 on Build GRCh38
rs774539379 Chr.19: 49854656-49854656 on Build GRCh38
rs746982022 Chr.19: 49854763-49854764 on Build GRCh38
rs763022646 Chr.19: 49854442-49854443 on Build GRCh38
rs752338633 Chr.19: 49854734-49854735 on Build GRCh38
rs140236059 Chr.19: 49854715-49854715 on Build GRCh38
rs139204161 Chr.19: 49854462-49854462 on Build GRCh38
rs374564420 Chr.19: 49854747-49854747 on Build GRCh38
rs775802191 Chr.19: 49854816-49854816 on Build GRCh38
rs553423378 Chr.19: 49854755-49854755 on Build GRCh38
rs200911767 Chr.19: 49854588-49854588 on Build GRCh38
rs901784650 Chr.19: 49854574-49854574 on Build GRCh38
rs761907415 Chr.19: 49854510-49854510 on Build GRCh38
rs143215505 Chr.19: 49854525-49854525 on Build GRCh38
rs748446625 Chr.19: 49854721-49854721 on Build GRCh38
rs752114387 Chr.19: 49854527-49854528 on Build GRCh38
rs764053837 Chr.19: 49854692-49854692 on Build GRCh38
rs770745504 Chr.19: 49854487-49854487 on Build GRCh38
rs370036159 Chr.19: 49854494-49854494 on Build GRCh38
rs770230641 Chr.19: 49854645-49854645 on Build GRCh38
rs370943862 Chr.19: 49854744-49854744 on Build GRCh38
rs777005025 Chr.19: 49854508-49854508 on Build GRCh38
rs773003272 Chr.19: 49854423-49854423 on Build GRCh38
rs201967614 Chr.19: 49854624-49854624 on Build GRCh38
rs766865204 Chr.19: 49854816-49854817 on Build GRCh38
rs765279767 Chr.19: 49854762-49854762 on Build GRCh38
rs756115404 Chr.19: 49854623-49854623 on Build GRCh38
rs759326327 Chr.19: 49854607-49854607 on Build GRCh38
rs373959297 Chr.19: 49854773-49854773 on Build GRCh38
rs753338524 Chr.19: 49854626-49854626 on Build GRCh38
rs746017107 Chr.19: 49854733-49854733 on Build GRCh38
rs756463596 Chr.19: 49854581-49854582 on Build GRCh38
rs764100094 Chr.19: 49854525-49854526 on Build GRCh38
rs773234830 Chr.19: 49854838-49854838 on Build GRCh38
rs1022832210 Chr.19: 49854515-49854515 on Build GRCh38
rs11558837 Chr.19: 49854668-49854668 on Build GRCh38
rs946762278 Chr.19: 49854701-49854701 on Build GRCh38
rs201591948 Chr.19: 49854763-49854763 on Build GRCh38
rs759939370 Chr.19: 49854424-49854424 on Build GRCh38
rs769461066 Chr.19: 49854480-49854480 on Build GRCh38
rs372605957 Chr.19: 49854772-49854772 on Build GRCh38
rs545720828 Chr.19: 49854468-49854468 on Build GRCh38
rs1040241071 Chr.19: 49854556-49854556 on Build GRCh38
rs768929363 Chr.19: 49854583-49854583 on Build GRCh38
rs765282722 Chr.19: 49854521-49854521 on Build GRCh38
rs748872444 Chr.19: 49854564-49854564 on Build GRCh38
rs751761421 Chr.19: 49854605-49854605 on Build GRCh38
rs1035180089 Chr.19: 49854810-49854810 on Build GRCh38
rs374560183 Chr.19: 49854617-49854617 on Build GRCh38
rs549336919 Chr.19: 49854526-49854526 on Build GRCh38
rs377247875 Chr.19: 49854654-49854654 on Build GRCh38
rs762322759 Chr.19: 49854848-49854848 on Build GRCh38
rs769070734 Chr.19: 49854820-49854820 on Build GRCh38
rs147943327 Chr.19: 49854632-49854632 on Build GRCh38
rs377218952 Chr.19: 49854455-49854455 on Build GRCh38
rs1002340336 Chr.19: 49854800-49854800 on Build GRCh38
rs370169892 Chr.19: 49854697-49854697 on Build GRCh38
rs371342339 Chr.19: 49854849-49854849 on Build GRCh38
rs752525642 Chr.19: 49854661-49854661 on Build GRCh38
rs967370174 Chr.19: 49854694-49854694 on Build GRCh38
rs147535884 Chr.19: 49854660-49854660 on Build GRCh38
rs757931333 Chr.19: 49854768-49854768 on Build GRCh38
rs919385983 Chr.19: 49854550-49854550 on Build GRCh38
rs772222630 Chr.19: 49854815-49854815 on Build GRCh38
rs200056596 Chr.19: 49854635-49854635 on Build GRCh38
rs532879959 Chr.19: 49854729-49854729 on Build GRCh38
rs113897016 Chr.19: 49854830-49854830 on Build GRCh38
rs375276416 Chr.19: 49854519-49854519 on Build GRCh38
rs148982635 Chr.19: 49854633-49854633 on Build GRCh38
rs969799824 Chr.19: 49854794-49854794 on Build GRCh38
rs779487402 Chr.19: 49854808-49854808 on Build GRCh38
rs757293505 Chr.19: 49854454-49854454 on Build GRCh38
rs753853560 Chr.19: 49854700-49854700 on Build GRCh38
rs750669912 Chr.19: 49854456-49854456 on Build GRCh38
rs752576001 Chr.19: 49854621-49854621 on Build GRCh38
rs781132685 Chr.19: 49854775-49854776 on Build GRCh38
rs757237405 Chr.19: 49854765-49854766 on Build GRCh38
rs747312801 Chr.19: 49854611-49854611 on Build GRCh38
rs770938530 Chr.19: 49854655-49854655 on Build GRCh38
rs761288567 Chr.19: 49854413-49854413 on Build GRCh38
rs756773891 Chr.19: 49854705-49854705 on Build GRCh38
rs762281105 Chr.19: 49854591-49854591 on Build GRCh38
rs768014871 Chr.19: 49854430-49854430 on Build GRCh38
rs764600206 Chr.19: 49854495-49854495 on Build GRCh38
rs770692076 Chr.19: 49854568-49854568 on Build GRCh38
rs913955289 Chr.19: 49854516-49854516 on Build GRCh38
rs781144832 Chr.19: 49854542-49854542 on Build GRCh38
rs370929811 Chr.19: 49854595-49854595 on Build GRCh38
rs1052440687 Chr.19: 49854498-49854498 on Build GRCh38
rs372882504 Chr.19: 49854634-49854634 on Build GRCh38
rs764573893 Chr.19: 49854442-49854442 on Build GRCh38
rs758042401 Chr.19: 49854749-49854763 on Build GRCh38
rs766523467 Chr.19: 49854599-49854599 on Build GRCh38
rs777418918 Chr.19: 49854757-49854758 on Build GRCh38
rs762299709 Chr.19: 49854556-49854556 on Build GRCh38
rs759645419 Chr.19: 49854657-49854657 on Build GRCh38
rs772950016 Chr.19: 49854481-49854481 on Build GRCh38

View Full Product Details