Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49864076-49864577 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PNKP
Entrez Gene ID
11284
Gene Name
polynucleotide kinase 3'-phosphatase
Gene Aliases
AOA4, EIEE10, MCSZ, PNK
UniGene
Hs.78016
Gene Chromosome Location
Chr.19: 49861203-49867565 on Build GRCh38
Transcripts
NM_007254.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6751257 c.602G>A p.R201H Chr.19: 49864213-49864213 on Build GRCh38
7384779 c.538C>A p.R180S Chr.19: 49864364-49864364 on Build GRCh38
6751255 c.524C>T p.T175M Chr.19: 49864378-49864378 on Build GRCh38
4644232 c.624C>T p.A208A Chr.19: 49864191-49864191 on Build GRCh38
3835404 c.625G>A p.E209K Chr.19: 49864190-49864190 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751065721 Chr.19: 49864440-49864440 on Build GRCh38
rs796052859 Chr.19: 49864211-49864212 on Build GRCh38
rs142770020 Chr.19: 49864382-49864382 on Build GRCh38
rs1006890687 Chr.19: 49864555-49864555 on Build GRCh38
rs904492234 Chr.19: 49864541-49864541 on Build GRCh38
rs760819552 Chr.19: 49864321-49864321 on Build GRCh38
rs926857628 Chr.19: 49864282-49864282 on Build GRCh38
rs767753048 Chr.19: 49864241-49864241 on Build GRCh38
rs531793511 Chr.19: 49864529-49864529 on Build GRCh38
rs771458611 Chr.19: 49864375-49864375 on Build GRCh38
rs770185057 Chr.19: 49864278-49864278 on Build GRCh38
rs1044770289 Chr.19: 49864476-49864476 on Build GRCh38
rs138931842 Chr.19: 49864228-49864228 on Build GRCh38
rs758697522 Chr.19: 49864422-49864423 on Build GRCh38
rs796052850 Chr.19: 49864205-49864205 on Build GRCh38
rs778015264 Chr.19: 49864279-49864279 on Build GRCh38
rs780365093 Chr.19: 49864327-49864327 on Build GRCh38
rs371834726 Chr.19: 49864240-49864240 on Build GRCh38
rs771585511 Chr.19: 49864289-49864289 on Build GRCh38
rs754563867 Chr.19: 49864445-49864445 on Build GRCh38
rs767378244 Chr.19: 49864209-49864209 on Build GRCh38
rs768950578 Chr.19: 49864167-49864167 on Build GRCh38
rs929287033 Chr.19: 49864545-49864545 on Build GRCh38
rs71353341 Chr.19: 49864447-49864447 on Build GRCh38
rs909512959 Chr.19: 49864410-49864410 on Build GRCh38
rs368556374 Chr.19: 49864137-49864137 on Build GRCh38
rs939139668 Chr.19: 49864491-49864491 on Build GRCh38
rs545573584 Chr.19: 49864407-49864407 on Build GRCh38
rs375294634 Chr.19: 49864149-49864149 on Build GRCh38
rs773988123 Chr.19: 49864164-49864164 on Build GRCh38
rs906272600 Chr.19: 49864483-49864483 on Build GRCh38
rs778146955 Chr.19: 49864370-49864370 on Build GRCh38
rs3739186 Chr.19: 49864229-49864229 on Build GRCh38
rs3739185 Chr.19: 49864364-49864364 on Build GRCh38
rs186103446 Chr.19: 49864507-49864507 on Build GRCh38
rs746434170 Chr.19: 49864381-49864381 on Build GRCh38
rs372737887 Chr.19: 49864417-49864417 on Build GRCh38
rs774972325 Chr.19: 49864156-49864156 on Build GRCh38
rs926779502 Chr.19: 49864516-49864516 on Build GRCh38
rs368764301 Chr.19: 49864255-49864255 on Build GRCh38
rs766512638 Chr.19: 49864237-49864237 on Build GRCh38
rs1026094855 Chr.19: 49864131-49864131 on Build GRCh38
rs758501201 Chr.19: 49864142-49864142 on Build GRCh38
rs756105740 Chr.19: 49864155-49864155 on Build GRCh38
rs571119317 Chr.19: 49864191-49864191 on Build GRCh38
rs772780281 Chr.19: 49864118-49864118 on Build GRCh38
rs199541652 Chr.19: 49864254-49864254 on Build GRCh38
rs759654539 Chr.19: 49864307-49864307 on Build GRCh38
rs994228667 Chr.19: 49864112-49864112 on Build GRCh38
rs749250190 Chr.19: 49864218-49864218 on Build GRCh38
rs773641701 Chr.19: 49864190-49864190 on Build GRCh38
rs3739187 Chr.19: 49864172-49864172 on Build GRCh38
rs761565319 Chr.19: 49864409-49864409 on Build GRCh38
rs781246709 Chr.19: 49864097-49864097 on Build GRCh38
rs774039394 Chr.19: 49864128-49864128 on Build GRCh38
rs749959202 Chr.19: 49864257-49864257 on Build GRCh38
rs267606957 Chr.19: 49864376-49864376 on Build GRCh38
rs367553967 Chr.19: 49864206-49864206 on Build GRCh38
rs3739188 Chr.19: 49864098-49864098 on Build GRCh38
rs938215122 Chr.19: 49864517-49864517 on Build GRCh38
rs929865929 Chr.19: 49864389-49864389 on Build GRCh38
rs201968000 Chr.19: 49864226-49864226 on Build GRCh38
rs145904995 Chr.19: 49864236-49864236 on Build GRCh38
rs745459391 Chr.19: 49864227-49864227 on Build GRCh38
rs760910540 Chr.19: 49864392-49864392 on Build GRCh38
rs570975244 Chr.19: 49864336-49864336 on Build GRCh38
rs762469707 Chr.19: 49864121-49864121 on Build GRCh38
rs751820948 Chr.19: 49864204-49864204 on Build GRCh38
rs749769920 Chr.19: 49864280-49864280 on Build GRCh38
rs144284975 Chr.19: 49864383-49864383 on Build GRCh38
rs757925060 Chr.19: 49864357-49864357 on Build GRCh38
rs779727048 Chr.19: 49864268-49864268 on Build GRCh38
rs150253619 Chr.19: 49864213-49864213 on Build GRCh38
rs763852240 Chr.19: 49864247-49864247 on Build GRCh38
rs755907454 Chr.19: 49864212-49864212 on Build GRCh38
rs748805586 Chr.19: 49864161-49864161 on Build GRCh38
rs968726480 Chr.19: 49864266-49864266 on Build GRCh38
rs777095183 Chr.19: 49864160-49864160 on Build GRCh38
rs745577158 Chr.19: 49864166-49864166 on Build GRCh38
rs752898407 Chr.19: 49864242-49864242 on Build GRCh38
rs1030278703 Chr.19: 49864462-49864463 on Build GRCh38
rs770100322 Chr.19: 49864230-49864230 on Build GRCh38
rs770468060 Chr.19: 49864162-49864162 on Build GRCh38
rs532550120 Chr.19: 49864188-49864188 on Build GRCh38
rs767243528 Chr.19: 49864454-49864454 on Build GRCh38
rs749645305 Chr.19: 49864374-49864374 on Build GRCh38
rs955732346 Chr.19: 49864245-49864245 on Build GRCh38
rs751873804 Chr.19: 49864148-49864148 on Build GRCh38
rs1044286634 Chr.19: 49864276-49864276 on Build GRCh38
rs988493693 Chr.19: 49864505-49864505 on Build GRCh38
rs774601739 Chr.19: 49864239-49864239 on Build GRCh38
rs574408360 Chr.19: 49864338-49864338 on Build GRCh38
rs911037129 Chr.19: 49864179-49864179 on Build GRCh38
rs1035086677 Chr.19: 49864125-49864125 on Build GRCh38
rs767085452 Chr.19: 49864154-49864154 on Build GRCh38
rs765084625 Chr.19: 49864413-49864413 on Build GRCh38
rs149947403 Chr.19: 49864290-49864290 on Build GRCh38
rs997841657 Chr.19: 49864455-49864455 on Build GRCh38
rs779428180 Chr.19: 49864392-49864392 on Build GRCh38
rs3739184 Chr.19: 49864430-49864430 on Build GRCh38
rs779492301 Chr.19: 49864363-49864363 on Build GRCh38
rs1018297810 Chr.19: 49864556-49864556 on Build GRCh38
rs1021889154 Chr.19: 49864530-49864530 on Build GRCh38
rs753093330 Chr.19: 49864420-49864421 on Build GRCh38
rs556261718 Chr.19: 49864534-49864535 on Build GRCh38
rs180965613 Chr.19: 49864143-49864143 on Build GRCh38
rs755931733 Chr.19: 49864246-49864246 on Build GRCh38
rs201129262 Chr.19: 49864335-49864335 on Build GRCh38
rs190908648 Chr.19: 49864513-49864513 on Build GRCh38
rs376534063 Chr.19: 49864129-49864129 on Build GRCh38
rs201738905 Chr.19: 49864133-49864133 on Build GRCh38
rs772984464 Chr.19: 49864514-49864515 on Build GRCh38
rs769125579 Chr.19: 49864116-49864116 on Build GRCh38
rs765850622 Chr.19: 49864439-49864439 on Build GRCh38
rs1056629962 Chr.19: 49864525-49864525 on Build GRCh38
rs369818814 Chr.19: 49864350-49864350 on Build GRCh38
rs772109789 Chr.19: 49864311-49864311 on Build GRCh38
rs868590428 Chr.19: 49864422-49864422 on Build GRCh38
rs142143566 Chr.19: 49864401-49864401 on Build GRCh38
rs202138901 Chr.19: 49864269-49864269 on Build GRCh38
rs200789718 Chr.19: 49864263-49864263 on Build GRCh38
rs770588540 Chr.19: 49864123-49864123 on Build GRCh38
rs755418385 Chr.19: 49864132-49864157 on Build GRCh38
rs1056754589 Chr.19: 49864324-49864324 on Build GRCh38
rs189056874 Chr.19: 49864138-49864138 on Build GRCh38
rs779554596 Chr.19: 49864380-49864380 on Build GRCh38
rs886042466 Chr.19: 49864408-49864408 on Build GRCh38
rs773769080 Chr.19: 49864231-49864231 on Build GRCh38
rs775768360 Chr.19: 49864314-49864314 on Build GRCh38
rs766637329 Chr.19: 49864193-49864193 on Build GRCh38

View Full Product Details