Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 50443626-50444102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYBPC2
Entrez Gene ID
4606
Gene Name
myosin binding protein C, fast type
Gene Aliases
MYBPC, MYBPCF
UniGene
Hs.85937
Gene Chromosome Location
Chr.19: 50432903-50466326 on Build GRCh38
Transcripts
NM_004533.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5929925 c.1087G>A p.V363M Chr.19: 50443770-50443770 on Build GRCh38
6525206 c.1030C>A p.P344T Chr.19: 50443713-50443713 on Build GRCh38
7378198 c.1064A>T p.Q355L Chr.19: 50443747-50443747 on Build GRCh38
6895989 c.1028-1G>A p.? Chr.19: 50443710-50443710 on Build GRCh38
7134844 c.1133+1G>A p.? Chr.19: 50443817-50443817 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868265430 Chr.19: 50444035-50444035 on Build GRCh38
rs945922961 Chr.19: 50443923-50443923 on Build GRCh38
rs767716644 Chr.19: 50443657-50443657 on Build GRCh38
rs759897532 Chr.19: 50443653-50443653 on Build GRCh38
rs763152892 Chr.19: 50443820-50443820 on Build GRCh38
rs781087868 Chr.19: 50443721-50443721 on Build GRCh38
rs751362189 Chr.19: 50443834-50443834 on Build GRCh38
rs534923875 Chr.19: 50443817-50443817 on Build GRCh38
rs377450077 Chr.19: 50443743-50443743 on Build GRCh38
rs747685663 Chr.19: 50443730-50443730 on Build GRCh38
rs950918476 Chr.19: 50443799-50443799 on Build GRCh38
rs777626792 Chr.19: 50443777-50443778 on Build GRCh38
rs771856464 Chr.19: 50443761-50443786 on Build GRCh38
rs754912829 Chr.19: 50443714-50443714 on Build GRCh38
rs1043257442 Chr.19: 50443964-50443964 on Build GRCh38
rs563859405 Chr.19: 50443913-50443913 on Build GRCh38
rs374722391 Chr.19: 50444051-50444051 on Build GRCh38
rs142489516 Chr.19: 50443832-50443832 on Build GRCh38
rs1054939465 Chr.19: 50444069-50444069 on Build GRCh38
rs964936728 Chr.19: 50443762-50443762 on Build GRCh38
rs761075304 Chr.19: 50443779-50443779 on Build GRCh38
rs748097385 Chr.19: 50443753-50443756 on Build GRCh38
rs375664625 Chr.19: 50443990-50444001 on Build GRCh38
rs904282912 Chr.19: 50444011-50444011 on Build GRCh38
rs531276596 Chr.19: 50443950-50443950 on Build GRCh38
rs368586553 Chr.19: 50443837-50443837 on Build GRCh38
rs1033478370 Chr.19: 50443668-50443668 on Build GRCh38
rs537673479 Chr.19: 50443830-50443830 on Build GRCh38
rs76966642 Chr.19: 50443869-50443869 on Build GRCh38
rs997684324 Chr.19: 50443790-50443790 on Build GRCh38
rs750610045 Chr.19: 50443692-50443692 on Build GRCh38
rs761149352 Chr.19: 50443671-50443671 on Build GRCh38
rs764718000 Chr.19: 50443800-50443800 on Build GRCh38
rs775905702 Chr.19: 50443670-50443670 on Build GRCh38
rs992784626 Chr.19: 50443885-50443885 on Build GRCh38
rs770608151 Chr.19: 50443768-50443768 on Build GRCh38
rs754323043 Chr.19: 50443927-50443927 on Build GRCh38
rs573288749 Chr.19: 50443691-50443691 on Build GRCh38
rs75028924 Chr.19: 50443789-50443789 on Build GRCh38
rs201433983 Chr.19: 50443767-50443767 on Build GRCh38
rs765354092 Chr.19: 50443809-50443809 on Build GRCh38
rs980139949 Chr.19: 50443868-50443868 on Build GRCh38
rs987354257 Chr.19: 50443965-50443965 on Build GRCh38
rs912685128 Chr.19: 50443974-50443974 on Build GRCh38
rs866872359 Chr.19: 50443874-50443874 on Build GRCh38
rs552817311 Chr.19: 50443801-50443801 on Build GRCh38
rs146785361 Chr.19: 50443986-50443997 on Build GRCh38
rs188593033 Chr.19: 50444050-50444050 on Build GRCh38
rs779770854 Chr.19: 50443697-50443697 on Build GRCh38
rs761992019 Chr.19: 50443804-50443804 on Build GRCh38
rs373358196 Chr.19: 50443681-50443681 on Build GRCh38
rs950502104 Chr.19: 50443746-50443746 on Build GRCh38
rs754017850 Chr.19: 50443672-50443672 on Build GRCh38
rs962957967 Chr.19: 50443878-50443878 on Build GRCh38
rs1030536853 Chr.19: 50443792-50443792 on Build GRCh38
rs758231305 Chr.19: 50443694-50443694 on Build GRCh38
rs772864373 Chr.19: 50443760-50443760 on Build GRCh38
rs778862941 Chr.19: 50443667-50443668 on Build GRCh38
rs764585595 Chr.19: 50443814-50443814 on Build GRCh38
rs775754598 Chr.19: 50443771-50443771 on Build GRCh38
rs561299893 Chr.19: 50444059-50444059 on Build GRCh38
rs772809439 Chr.19: 50443651-50443651 on Build GRCh38
rs746816703 Chr.19: 50443698-50443698 on Build GRCh38
rs781110615 Chr.19: 50443857-50443857 on Build GRCh38
rs752572282 Chr.19: 50443859-50443859 on Build GRCh38
rs754760396 Chr.19: 50443854-50443854 on Build GRCh38

View Full Product Details