Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55302616-55303122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRSK1
Entrez Gene ID
84446
Gene Name
BR serine/threonine kinase 1
Gene Aliases
hSAD1
UniGene
Hs.182081
Gene Chromosome Location
Chr.19: 55284101-55312562 on Build GRCh38
Transcripts
NM_032430.1 XM_005259327.3 XM_011527395.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3404641 c.980G>C p.G327A Chr.19: 55302819-55302819 on Build GRCh38
144391 c.1019G>A p.R340H Chr.19: 55302858-55302858 on Build GRCh38
7357768 c.1026G>T p.E342D Chr.19: 55302865-55302865 on Build GRCh38
4983028 c.908G>A p.R303Q Chr.19: 55302747-55302747 on Build GRCh38
6612988 c.948C>T p.D316D Chr.19: 55302787-55302787 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs957943278 Chr.19: 55303034-55303034 on Build GRCh38
rs778088324 Chr.19: 55302899-55302899 on Build GRCh38
rs139469654 Chr.19: 55302835-55302835 on Build GRCh38
rs747186655 Chr.19: 55303060-55303060 on Build GRCh38
rs138976142 Chr.19: 55302766-55302766 on Build GRCh38
rs767793522 Chr.19: 55302749-55302749 on Build GRCh38
rs749274433 Chr.19: 55302825-55302825 on Build GRCh38
rs536845893 Chr.19: 55303033-55303033 on Build GRCh38
rs1006353038 Chr.19: 55302961-55302961 on Build GRCh38
rs752251740 Chr.19: 55302892-55302892 on Build GRCh38
rs769339374 Chr.19: 55302815-55302815 on Build GRCh38
rs932738413 Chr.19: 55302760-55302760 on Build GRCh38
rs946111526 Chr.19: 55302851-55302851 on Build GRCh38
rs762774044 Chr.19: 55302739-55302739 on Build GRCh38
rs745487860 Chr.19: 55302809-55302809 on Build GRCh38
rs773610849 Chr.19: 55302850-55302850 on Build GRCh38
rs1035469680 Chr.19: 55303067-55303067 on Build GRCh38
rs778404767 Chr.19: 55302709-55302710 on Build GRCh38
rs754419549 Chr.19: 55302772-55302772 on Build GRCh38
rs147341219 Chr.19: 55302817-55302817 on Build GRCh38
rs1053904925 Chr.19: 55303013-55303013 on Build GRCh38
rs748684276 Chr.19: 55302909-55302909 on Build GRCh38
rs746696435 Chr.19: 55302698-55302698 on Build GRCh38
rs1024591768 Chr.19: 55302642-55302642 on Build GRCh38
rs146532428 Chr.19: 55302958-55302958 on Build GRCh38
rs371888974 Chr.19: 55302922-55302922 on Build GRCh38
rs779616409 Chr.19: 55302908-55302908 on Build GRCh38
rs746809522 Chr.19: 55302990-55302990 on Build GRCh38
rs775445066 Chr.19: 55302727-55302727 on Build GRCh38
rs753110336 Chr.19: 55302649-55302649 on Build GRCh38
rs893806975 Chr.19: 55303016-55303016 on Build GRCh38
rs958424399 Chr.19: 55302949-55302949 on Build GRCh38
rs757321396 Chr.19: 55302913-55302914 on Build GRCh38
rs768407957 Chr.19: 55302834-55302834 on Build GRCh38
rs758877732 Chr.19: 55302682-55302683 on Build GRCh38
rs760094147 Chr.19: 55302884-55302884 on Build GRCh38
rs770130862 Chr.19: 55302758-55302758 on Build GRCh38
rs766537322 Chr.19: 55302770-55302770 on Build GRCh38
rs944312011 Chr.19: 55302997-55302997 on Build GRCh38
rs753822124 Chr.19: 55302883-55302883 on Build GRCh38
rs1041220945 Chr.19: 55302712-55302712 on Build GRCh38
rs780693381 Chr.19: 55302705-55302705 on Build GRCh38
rs142894930 Chr.19: 55302804-55302804 on Build GRCh38
rs375928229 Chr.19: 55302903-55302903 on Build GRCh38
rs756706326 Chr.19: 55302650-55302650 on Build GRCh38
rs1003084352 Chr.19: 55302926-55302926 on Build GRCh38
rs1053765698 Chr.19: 55302845-55302845 on Build GRCh38
rs755411496 Chr.19: 55302683-55302683 on Build GRCh38
rs921451853 Chr.19: 55303011-55303011 on Build GRCh38
rs893838211 Chr.19: 55302880-55302880 on Build GRCh38
rs757026588 Chr.19: 55302999-55302999 on Build GRCh38
rs762094970 Chr.19: 55302747-55302747 on Build GRCh38
rs767069878 Chr.19: 55302666-55302666 on Build GRCh38
rs952415799 Chr.19: 55302962-55302962 on Build GRCh38
rs549017341 Chr.19: 55302769-55302769 on Build GRCh38
rs769287871 Chr.19: 55302725-55302725 on Build GRCh38
rs141186658 Chr.19: 55302988-55302988 on Build GRCh38
rs1002810663 Chr.19: 55303061-55303061 on Build GRCh38
rs766590610 Chr.19: 55302876-55302876 on Build GRCh38
rs745440598 Chr.19: 55302720-55302720 on Build GRCh38
rs976148015 Chr.19: 55303095-55303095 on Build GRCh38
rs779367841 Chr.19: 55302787-55302787 on Build GRCh38
rs765722079 Chr.19: 55302890-55302890 on Build GRCh38
rs760959221 Chr.19: 55302859-55302859 on Build GRCh38
rs770450285 Chr.19: 55302700-55302700 on Build GRCh38
rs747494431 Chr.19: 55302905-55302905 on Build GRCh38
rs565644434 Chr.19: 55302947-55302947 on Build GRCh38
rs755391793 Chr.19: 55302775-55302775 on Build GRCh38
rs756168876 Chr.19: 55302902-55302902 on Build GRCh38
rs908749098 Chr.19: 55302996-55302996 on Build GRCh38
rs755721806 Chr.19: 55302907-55302907 on Build GRCh38
rs141095323 Chr.19: 55303051-55303051 on Build GRCh38
rs531758783 Chr.19: 55302950-55302950 on Build GRCh38
rs189108034 Chr.19: 55303035-55303035 on Build GRCh38
rs1035499464 Chr.19: 55302948-55302948 on Build GRCh38
rs571393535 Chr.19: 55302990-55302990 on Build GRCh38
rs771990360 Chr.19: 55302841-55302841 on Build GRCh38
rs932276475 Chr.19: 55302933-55302933 on Build GRCh38
rs899885867 Chr.19: 55302759-55302759 on Build GRCh38
rs768515985 Chr.19: 55302743-55302743 on Build GRCh38
rs752945445 Chr.19: 55302795-55302795 on Build GRCh38
rs779127224 Chr.19: 55302687-55302687 on Build GRCh38
rs372675247 Chr.19: 55302836-55302836 on Build GRCh38
rs780150120 Chr.19: 55303012-55303012 on Build GRCh38
rs969440173 Chr.19: 55303000-55303000 on Build GRCh38
rs749810972 Chr.19: 55302674-55302674 on Build GRCh38
rs754501327 Chr.19: 55302910-55302910 on Build GRCh38
rs758748714 Chr.19: 55302803-55302803 on Build GRCh38
rs758801610 Chr.19: 55302897-55302897 on Build GRCh38
rs144130246 Chr.19: 55302746-55302746 on Build GRCh38
rs897616433 Chr.19: 55302967-55302967 on Build GRCh38

View Full Product Details