Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55390117-55390627 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPL28
Entrez Gene ID
6158
Gene Name
ribosomal protein L28
Gene Aliases
L28
UniGene
Hs.652114
Gene Chromosome Location
Chr.19: 55385932-55392085 on Build GRCh38
Transcripts
NM_000991.4 NM_001136134.1 NM_001136135.1

SNP Detail

SNP ID Chromosome Location
rs982467086 Chr.19: 55390181-55390181 on Build GRCh38
rs35860013 Chr.19: 55390583-55390584 on Build GRCh38
rs148318618 Chr.19: 55390497-55390497 on Build GRCh38
rs1008918715 Chr.19: 55390245-55390245 on Build GRCh38
rs997199930 Chr.19: 55390253-55390253 on Build GRCh38
rs981861662 Chr.19: 55390552-55390552 on Build GRCh38
rs4806669 Chr.19: 55390285-55390285 on Build GRCh38
rs45621533 Chr.19: 55390548-55390548 on Build GRCh38
rs143306924 Chr.19: 55390489-55390489 on Build GRCh38
rs925550859 Chr.19: 55390248-55390248 on Build GRCh38
rs548190178 Chr.19: 55390478-55390478 on Build GRCh38
rs937684690 Chr.19: 55390315-55390315 on Build GRCh38
rs548360952 Chr.19: 55390484-55390484 on Build GRCh38
rs969566068 Chr.19: 55390547-55390547 on Build GRCh38
rs987900363 Chr.19: 55390292-55390292 on Build GRCh38
rs1003852779 Chr.19: 55390466-55390466 on Build GRCh38
rs1020767564 Chr.19: 55390537-55390537 on Build GRCh38
rs970427909 Chr.19: 55390307-55390307 on Build GRCh38
rs974104921 Chr.19: 55390321-55390321 on Build GRCh38
rs73054982 Chr.19: 55390567-55390567 on Build GRCh38
rs929471195 Chr.19: 55390395-55390395 on Build GRCh38
rs564734707 Chr.19: 55390266-55390266 on Build GRCh38
rs993664751 Chr.19: 55390218-55390218 on Build GRCh38
rs981807952 Chr.19: 55390308-55390308 on Build GRCh38
rs45477992 Chr.19: 55390167-55390167 on Build GRCh38
rs141179850 Chr.19: 55390379-55390379 on Build GRCh38
rs888378743 Chr.19: 55390454-55390454 on Build GRCh38
rs890608417 Chr.19: 55390468-55390468 on Build GRCh38
rs959789657 Chr.19: 55390217-55390217 on Build GRCh38
rs890512260 Chr.19: 55390235-55390235 on Build GRCh38
rs1009373872 Chr.19: 55390526-55390526 on Build GRCh38
rs918089007 Chr.19: 55390231-55390231 on Build GRCh38
rs1052402033 Chr.19: 55390276-55390276 on Build GRCh38
rs562880759 Chr.19: 55390387-55390387 on Build GRCh38
rs981034075 Chr.19: 55390242-55390242 on Build GRCh38
rs753818848 Chr.19: 55390163-55390163 on Build GRCh38
rs761282360 Chr.19: 55390207-55390207 on Build GRCh38
rs781337616 Chr.19: 55390267-55390267 on Build GRCh38
rs1020674123 Chr.19: 55390303-55390303 on Build GRCh38
rs1017639034 Chr.19: 55390246-55390246 on Build GRCh38
rs973061588 Chr.19: 55390583-55390583 on Build GRCh38
rs79869003 Chr.19: 55390234-55390234 on Build GRCh38
rs751405713 Chr.19: 55390534-55390534 on Build GRCh38
rs10425107 Chr.19: 55390184-55390184 on Build GRCh38
rs567943034 Chr.19: 55390488-55390488 on Build GRCh38
rs1023166522 Chr.19: 55390504-55390505 on Build GRCh38
rs11540466 Chr.19: 55390249-55390249 on Build GRCh38
rs952398710 Chr.19: 55390174-55390174 on Build GRCh38
rs778710053 Chr.19: 55390178-55390178 on Build GRCh38
rs771557881 Chr.19: 55390413-55390413 on Build GRCh38
rs944798742 Chr.19: 55390471-55390471 on Build GRCh38
rs186183165 Chr.19: 55390309-55390309 on Build GRCh38
rs777955955 Chr.19: 55390335-55390335 on Build GRCh38
rs370183612 Chr.19: 55390420-55390420 on Build GRCh38
rs11299036 Chr.19: 55390411-55390411 on Build GRCh38
rs45495700 Chr.19: 55390553-55390553 on Build GRCh38
rs553775119 Chr.19: 55390494-55390494 on Build GRCh38
rs376684360 Chr.19: 55390187-55390187 on Build GRCh38
rs749847501 Chr.19: 55390463-55390463 on Build GRCh38
rs529939140 Chr.19: 55390357-55390357 on Build GRCh38
rs1039107662 Chr.19: 55390472-55390472 on Build GRCh38
rs899364961 Chr.19: 55390211-55390211 on Build GRCh38
rs369971781 Chr.19: 55390205-55390205 on Build GRCh38
rs866456176 Chr.19: 55390490-55390490 on Build GRCh38
rs770043264 Chr.19: 55390330-55390330 on Build GRCh38
rs1032256378 Chr.19: 55390254-55390254 on Build GRCh38
rs992289489 Chr.19: 55390224-55390224 on Build GRCh38

View Full Product Details