Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 56621760-56622250 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF71
Entrez Gene ID
58491
Gene Name
zinc finger protein 71
Gene Aliases
EZFIT
UniGene
Hs.301431
Gene Chromosome Location
Chr.19: 56595259-56624481 on Build GRCh38
Transcripts
NM_021216.4 XM_011527193.2 XM_017027078.1 XM_017027079.1 XM_017027080.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5022054 c.857C>T p.T286I Chr.19: 56622144-56622144 on Build GRCh38
6832613 c.911G>A p.G304D Chr.19: 56622198-56622198 on Build GRCh38
1712932 c.818C>T p.P273L Chr.19: 56622105-56622105 on Build GRCh38
6832615 c.648C>T p.C216C Chr.19: 56621935-56621935 on Build GRCh38
6832609 c.734G>T p.G245V Chr.19: 56622021-56622021 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs8106568 Chr.19: 56621965-56621965 on Build GRCh38
rs867534279 Chr.19: 56622228-56622228 on Build GRCh38
rs781265021 Chr.19: 56621806-56621806 on Build GRCh38
rs538802556 Chr.19: 56622043-56622043 on Build GRCh38
rs947916623 Chr.19: 56621986-56621986 on Build GRCh38
rs371614088 Chr.19: 56622001-56622001 on Build GRCh38
rs773577607 Chr.19: 56621830-56621830 on Build GRCh38
rs768970719 Chr.19: 56622016-56622016 on Build GRCh38
rs148112500 Chr.19: 56621823-56621823 on Build GRCh38
rs779093808 Chr.19: 56622145-56622145 on Build GRCh38
rs536103931 Chr.19: 56621988-56621988 on Build GRCh38
rs781427340 Chr.19: 56621997-56621997 on Build GRCh38
rs756067721 Chr.19: 56621873-56621873 on Build GRCh38
rs755112007 Chr.19: 56621799-56621799 on Build GRCh38
rs147869247 Chr.19: 56621800-56621800 on Build GRCh38
rs752226323 Chr.19: 56622027-56622027 on Build GRCh38
rs746243538 Chr.19: 56622055-56622055 on Build GRCh38
rs201252925 Chr.19: 56622076-56622076 on Build GRCh38
rs368689449 Chr.19: 56622104-56622104 on Build GRCh38
rs764436296 Chr.19: 56621932-56621932 on Build GRCh38
rs897526784 Chr.19: 56622031-56622031 on Build GRCh38
rs868356457 Chr.19: 56621867-56621867 on Build GRCh38
rs879176975 Chr.19: 56621917-56621917 on Build GRCh38
rs756127762 Chr.19: 56621810-56621810 on Build GRCh38
rs769532421 Chr.19: 56622164-56622164 on Build GRCh38
rs760729510 Chr.19: 56621792-56621792 on Build GRCh38
rs763196663 Chr.19: 56621926-56621926 on Build GRCh38
rs767356203 Chr.19: 56621852-56621852 on Build GRCh38
rs765107648 Chr.19: 56622025-56622025 on Build GRCh38
rs199560535 Chr.19: 56621925-56621925 on Build GRCh38
rs781585548 Chr.19: 56622053-56622053 on Build GRCh38
rs763175149 Chr.19: 56622015-56622015 on Build GRCh38
rs754884155 Chr.19: 56621951-56621951 on Build GRCh38
rs750770330 Chr.19: 56622130-56622130 on Build GRCh38
rs750278640 Chr.19: 56621861-56621861 on Build GRCh38
rs780467724 Chr.19: 56622061-56622061 on Build GRCh38
rs968087811 Chr.19: 56622183-56622183 on Build GRCh38
rs766899741 Chr.19: 56621833-56621833 on Build GRCh38
rs756858548 Chr.19: 56622113-56622113 on Build GRCh38
rs777447946 Chr.19: 56621984-56621984 on Build GRCh38
rs772314811 Chr.19: 56622153-56622153 on Build GRCh38
rs200779625 Chr.19: 56622169-56622169 on Build GRCh38
rs773343246 Chr.19: 56622154-56622154 on Build GRCh38
rs745746304 Chr.19: 56621809-56621809 on Build GRCh38
rs781589174 Chr.19: 56621896-56621896 on Build GRCh38
rs772212199 Chr.19: 56622092-56622092 on Build GRCh38
rs141818908 Chr.19: 56622023-56622023 on Build GRCh38
rs562627527 Chr.19: 56621887-56621887 on Build GRCh38
rs773832910 Chr.19: 56622178-56622178 on Build GRCh38
rs755195979 Chr.19: 56621866-56621866 on Build GRCh38
rs548176437 Chr.19: 56621947-56621947 on Build GRCh38
rs759526820 Chr.19: 56621924-56621924 on Build GRCh38
rs762840279 Chr.19: 56622168-56622168 on Build GRCh38
rs776436135 Chr.19: 56622093-56622093 on Build GRCh38
rs748259634 Chr.19: 56622147-56622147 on Build GRCh38
rs1000934597 Chr.19: 56622184-56622184 on Build GRCh38
rs756502875 Chr.19: 56622133-56622133 on Build GRCh38
rs777743310 Chr.19: 56621970-56621970 on Build GRCh38
rs867101572 Chr.19: 56622212-56622212 on Build GRCh38
rs752789564 Chr.19: 56622229-56622229 on Build GRCh38
rs753872644 Chr.19: 56621949-56621949 on Build GRCh38
rs753917345 Chr.19: 56621795-56621795 on Build GRCh38
rs772545203 Chr.19: 56621829-56621829 on Build GRCh38
rs879041857 Chr.19: 56622060-56622060 on Build GRCh38
rs865841041 Chr.19: 56621936-56621936 on Build GRCh38
rs767195075 Chr.19: 56622190-56622190 on Build GRCh38
rs748854480 Chr.19: 56621819-56621819 on Build GRCh38
rs768915565 Chr.19: 56622079-56622079 on Build GRCh38
rs144761166 Chr.19: 56622007-56622007 on Build GRCh38
rs137860470 Chr.19: 56622163-56622163 on Build GRCh38
rs747913275 Chr.19: 56621935-56621935 on Build GRCh38
rs17851130 Chr.19: 56622223-56622223 on Build GRCh38
rs201233218 Chr.19: 56622203-56622203 on Build GRCh38
rs757941704 Chr.19: 56622029-56622029 on Build GRCh38
rs375499921 Chr.19: 56621977-56621977 on Build GRCh38
rs763727830 Chr.19: 56622036-56622036 on Build GRCh38
rs138773922 Chr.19: 56622019-56622019 on Build GRCh38
rs923968108 Chr.19: 56621878-56621878 on Build GRCh38
rs748314344 Chr.19: 56622085-56622085 on Build GRCh38
rs747393285 Chr.19: 56621908-56621908 on Build GRCh38
rs868096549 Chr.19: 56622210-56622210 on Build GRCh38
rs768173155 Chr.19: 56621820-56621820 on Build GRCh38
rs780191454 Chr.19: 56621880-56621880 on Build GRCh38
rs985908162 Chr.19: 56621791-56621791 on Build GRCh38
rs765187724 Chr.19: 56622097-56622097 on Build GRCh38
rs775273656 Chr.19: 56622167-56622167 on Build GRCh38
rs548728280 Chr.19: 56622098-56622098 on Build GRCh38
rs566518390 Chr.19: 56622042-56622042 on Build GRCh38
rs753025411 Chr.19: 56621845-56621845 on Build GRCh38
rs759476055 Chr.19: 56622083-56622083 on Build GRCh38
rs766217639 Chr.19: 56622022-56622022 on Build GRCh38
rs755544863 Chr.19: 56622144-56622144 on Build GRCh38
rs936418461 Chr.19: 56622211-56622211 on Build GRCh38
rs764289300 Chr.19: 56621939-56621940 on Build GRCh38
rs759295864 Chr.19: 56622094-56622094 on Build GRCh38
rs771402320 Chr.19: 56621909-56621909 on Build GRCh38
rs780599197 Chr.19: 56622142-56622142 on Build GRCh38
rs765342654 Chr.19: 56621836-56621836 on Build GRCh38
rs760589190 Chr.19: 56621945-56621945 on Build GRCh38
rs747778809 Chr.19: 56621903-56621903 on Build GRCh38
rs760390542 Chr.19: 56622199-56622199 on Build GRCh38
rs376573351 Chr.19: 56622006-56622006 on Build GRCh38
rs748456963 Chr.19: 56621824-56621824 on Build GRCh38
rs147430593 Chr.19: 56622226-56622226 on Build GRCh38
rs753256167 Chr.19: 56621883-56621883 on Build GRCh38
rs143688776 Chr.19: 56621822-56621822 on Build GRCh38
rs146186559 Chr.19: 56622059-56622059 on Build GRCh38
rs761101566 Chr.19: 56621831-56621831 on Build GRCh38
rs756122097 Chr.19: 56621821-56621821 on Build GRCh38
rs373155014 Chr.19: 56621998-56621998 on Build GRCh38
rs751103602 Chr.19: 56622105-56622105 on Build GRCh38
rs771714841 Chr.19: 56621907-56621907 on Build GRCh38
rs552420419 Chr.19: 56621803-56621803 on Build GRCh38
rs750223436 Chr.19: 56621944-56621944 on Build GRCh38
rs202008254 Chr.19: 56622161-56622161 on Build GRCh38
rs201305988 Chr.19: 56621781-56621782 on Build GRCh38
rs778418524 Chr.19: 56621958-56621958 on Build GRCh38
rs761273823 Chr.19: 56622182-56622182 on Build GRCh38
rs758566804 Chr.19: 56621848-56621848 on Build GRCh38
rs779611801 Chr.19: 56622227-56622227 on Build GRCh38
rs752178775 Chr.19: 56621963-56621963 on Build GRCh38
rs777158137 Chr.19: 56621914-56621914 on Build GRCh38
rs372609144 Chr.19: 56621834-56621834 on Build GRCh38
rs767128079 Chr.19: 56622115-56622115 on Build GRCh38

View Full Product Details