Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57487619-57488113 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF419
Entrez Gene ID
79744
Gene Name
zinc finger protein 419
Gene Aliases
ZAPHIR, ZNF419A
UniGene
Hs.744339
Gene Chromosome Location
Chr.19: 57487711-57494680 on Build GRCh38
Transcripts
NM_001098491.1 NM_001098492.1 NM_001098493.1 NM_001098494.1 NM_001098495.1 NM_001098496.1 NM_001291743.1 NM_001291744.1 NM_001291745.1 NM_024691.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6830025 c.9G>A p.A3A Chr.19: 57487959-57487959 on Build GRCh38
7294441 c.5C>T p.A2V Chr.19: 57487955-57487955 on Build GRCh38
3389450 c.15C>G p.A5A Chr.19: 57487965-57487965 on Build GRCh38
6830024 c.9G>A p.A3A Chr.19: 57487959-57487959 on Build GRCh38
6308426 c.22G>A p.D8N Chr.19: 57487972-57487972 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1037235286 Chr.19: 57487953-57487953 on Build GRCh38
rs1015833518 Chr.19: 57487802-57487802 on Build GRCh38
rs185296279 Chr.19: 57488071-57488071 on Build GRCh38
rs970172230 Chr.19: 57487845-57487845 on Build GRCh38
rs766545317 Chr.19: 57487931-57487931 on Build GRCh38
rs8113039 Chr.19: 57487814-57487814 on Build GRCh38
rs768981716 Chr.19: 57487720-57487720 on Build GRCh38
rs549713481 Chr.19: 57488000-57488000 on Build GRCh38
rs979894932 Chr.19: 57487853-57487853 on Build GRCh38
rs959854134 Chr.19: 57487878-57487878 on Build GRCh38
rs753900077 Chr.19: 57487998-57487998 on Build GRCh38
rs374521709 Chr.19: 57487911-57487911 on Build GRCh38
rs113628862 Chr.19: 57487965-57487965 on Build GRCh38
rs750498241 Chr.19: 57487926-57487926 on Build GRCh38
rs774711232 Chr.19: 57487920-57487920 on Build GRCh38
rs370176097 Chr.19: 57487910-57487910 on Build GRCh38
rs564578685 Chr.19: 57487637-57487637 on Build GRCh38
rs748945108 Chr.19: 57487966-57487966 on Build GRCh38
rs903962522 Chr.19: 57487644-57487644 on Build GRCh38
rs377623996 Chr.19: 57487917-57487917 on Build GRCh38
rs778665896 Chr.19: 57488026-57488026 on Build GRCh38
rs1037162058 Chr.19: 57488037-57488037 on Build GRCh38
rs552816224 Chr.19: 57487890-57487890 on Build GRCh38
rs533590046 Chr.19: 57487659-57487671 on Build GRCh38
rs781233306 Chr.19: 57487963-57487963 on Build GRCh38
rs369007634 Chr.19: 57487745-57487746 on Build GRCh38
rs147294415 Chr.19: 57487745-57487745 on Build GRCh38
rs375849290 Chr.19: 57487899-57487899 on Build GRCh38
rs1026659376 Chr.19: 57487818-57487818 on Build GRCh38
rs762242745 Chr.19: 57487922-57487922 on Build GRCh38
rs879023444 Chr.19: 57487904-57487904 on Build GRCh38
rs541370432 Chr.19: 57487941-57487941 on Build GRCh38
rs181262420 Chr.19: 57487999-57487999 on Build GRCh38
rs761250123 Chr.19: 57487981-57487981 on Build GRCh38
rs897346156 Chr.19: 57488049-57488049 on Build GRCh38
rs955781832 Chr.19: 57487701-57487701 on Build GRCh38
rs200962952 Chr.19: 57487925-57487925 on Build GRCh38
rs530606612 Chr.19: 57488085-57488085 on Build GRCh38
rs768241137 Chr.19: 57487972-57487972 on Build GRCh38
rs374429622 Chr.19: 57487977-57487977 on Build GRCh38
rs756301318 Chr.19: 57487929-57487929 on Build GRCh38
rs921870263 Chr.19: 57488006-57488006 on Build GRCh38
rs149140649 Chr.19: 57487888-57487888 on Build GRCh38
rs988629136 Chr.19: 57487886-57487886 on Build GRCh38
rs111473875 Chr.19: 57487753-57487753 on Build GRCh38
rs772544949 Chr.19: 57488032-57488032 on Build GRCh38
rs1015237769 Chr.19: 57487719-57487719 on Build GRCh38
rs371959035 Chr.19: 57488027-57488027 on Build GRCh38
rs550306590 Chr.19: 57487752-57487752 on Build GRCh38
rs757127375 Chr.19: 57488003-57488003 on Build GRCh38
rs534566039 Chr.19: 57487782-57487782 on Build GRCh38
rs188853075 Chr.19: 57487800-57487800 on Build GRCh38
rs751308508 Chr.19: 57487944-57487944 on Build GRCh38
rs997423463 Chr.19: 57487659-57487659 on Build GRCh38
rs111825322 Chr.19: 57487770-57487770 on Build GRCh38
rs549025351 Chr.19: 57487776-57487776 on Build GRCh38
rs762584724 Chr.19: 57487912-57487912 on Build GRCh38
rs757315252 Chr.19: 57487676-57487676 on Build GRCh38
rs1048627113 Chr.19: 57488054-57488054 on Build GRCh38
rs768330924 Chr.19: 57488011-57488011 on Build GRCh38
rs750445130 Chr.19: 57488004-57488004 on Build GRCh38
rs374962089 Chr.19: 57488010-57488010 on Build GRCh38
rs2158011 Chr.19: 57488002-57488002 on Build GRCh38
rs2158012 Chr.19: 57488079-57488079 on Build GRCh38
rs1009945940 Chr.19: 57488084-57488084 on Build GRCh38
rs536889965 Chr.19: 57487801-57487801 on Build GRCh38
rs1035354219 Chr.19: 57488076-57488076 on Build GRCh38
rs1035427752 Chr.19: 57487980-57487980 on Build GRCh38
rs996134139 Chr.19: 57488050-57488050 on Build GRCh38
rs1008396889 Chr.19: 57487942-57487942 on Build GRCh38
rs1028526610 Chr.19: 57487669-57487669 on Build GRCh38
rs754170928 Chr.19: 57487935-57487935 on Build GRCh38
rs112712302 Chr.19: 57487780-57487780 on Build GRCh38
rs948531963 Chr.19: 57487745-57487745 on Build GRCh38
rs891743319 Chr.19: 57487761-57487761 on Build GRCh38
rs755243267 Chr.19: 57487940-57487940 on Build GRCh38
rs755979883 Chr.19: 57488008-57488008 on Build GRCh38
rs143577485 Chr.19: 57487896-57487896 on Build GRCh38
rs987067421 Chr.19: 57487702-57487702 on Build GRCh38
rs1049820153 Chr.19: 57487913-57487913 on Build GRCh38
rs973756411 Chr.19: 57487728-57487728 on Build GRCh38
rs199880706 Chr.19: 57487975-57487975 on Build GRCh38
rs533367744 Chr.19: 57487751-57487751 on Build GRCh38
rs546181518 Chr.19: 57487774-57487774 on Build GRCh38
rs762196703 Chr.19: 57487976-57487976 on Build GRCh38
rs919666646 Chr.19: 57487744-57487744 on Build GRCh38
rs370981223 Chr.19: 57487945-57487945 on Build GRCh38
rs906133383 Chr.19: 57488070-57488070 on Build GRCh38
rs573626222 Chr.19: 57487872-57487872 on Build GRCh38
rs551208415 Chr.19: 57487795-57487795 on Build GRCh38
rs369148882 Chr.19: 57487965-57487970 on Build GRCh38
rs201210233 Chr.19: 57487984-57487984 on Build GRCh38
rs373519441 Chr.19: 57487964-57487964 on Build GRCh38

View Full Product Details