Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57572285-57572796 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF416
Entrez Gene ID
55659
Gene Name
zinc finger protein 416
Gene Aliases
-
UniGene
Hs.247711
Gene Chromosome Location
Chr.19: 57571015-57578938 on Build GRCh38
Transcripts
NM_017879.1 XM_005259051.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6026304 c.1499G>A p.G500E Chr.19: 57572405-57572405 on Build GRCh38
2972704 c.1162G>C p.G388R Chr.19: 57572742-57572742 on Build GRCh38
286854 c.1207A>T p.I403F Chr.19: 57572697-57572697 on Build GRCh38
5787503 c.1363A>T p.K455* Chr.19: 57572541-57572541 on Build GRCh38
5748135 c.1393C>T p.P465S Chr.19: 57572511-57572511 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371407051 Chr.19: 57572371-57572371 on Build GRCh38
rs201445198 Chr.19: 57572750-57572750 on Build GRCh38
rs565987759 Chr.19: 57572394-57572394 on Build GRCh38
rs142385856 Chr.19: 57572483-57572483 on Build GRCh38
rs867026265 Chr.19: 57572625-57572625 on Build GRCh38
rs749552148 Chr.19: 57572405-57572405 on Build GRCh38
rs761223467 Chr.19: 57572533-57572533 on Build GRCh38
rs367665355 Chr.19: 57572490-57572490 on Build GRCh38
rs557448143 Chr.19: 57572335-57572335 on Build GRCh38
rs955123748 Chr.19: 57572696-57572696 on Build GRCh38
rs779184173 Chr.19: 57572560-57572560 on Build GRCh38
rs746818643 Chr.19: 57572440-57572440 on Build GRCh38
rs1030472044 Chr.19: 57572692-57572692 on Build GRCh38
rs140295213 Chr.19: 57572643-57572643 on Build GRCh38
rs374204072 Chr.19: 57572419-57572419 on Build GRCh38
rs369374347 Chr.19: 57572690-57572690 on Build GRCh38
rs778244417 Chr.19: 57572761-57572761 on Build GRCh38
rs553291159 Chr.19: 57572518-57572518 on Build GRCh38
rs777435081 Chr.19: 57572436-57572436 on Build GRCh38
rs372260953 Chr.19: 57572574-57572574 on Build GRCh38
rs757707608 Chr.19: 57572379-57572379 on Build GRCh38
rs772518379 Chr.19: 57572648-57572648 on Build GRCh38
rs756238265 Chr.19: 57572396-57572396 on Build GRCh38
rs746934322 Chr.19: 57572337-57572337 on Build GRCh38
rs950127482 Chr.19: 57572520-57572520 on Build GRCh38
rs918003479 Chr.19: 57572401-57572401 on Build GRCh38
rs143138224 Chr.19: 57572575-57572575 on Build GRCh38
rs767956487 Chr.19: 57572522-57572522 on Build GRCh38
rs768459376 Chr.19: 57572384-57572384 on Build GRCh38
rs151206783 Chr.19: 57572628-57572628 on Build GRCh38
rs761270256 Chr.19: 57572668-57572668 on Build GRCh38
rs372856157 Chr.19: 57572497-57572497 on Build GRCh38
rs965861394 Chr.19: 57572382-57572383 on Build GRCh38
rs753937550 Chr.19: 57572758-57572759 on Build GRCh38
rs767934523 Chr.19: 57572496-57572496 on Build GRCh38
rs762221420 Chr.19: 57572521-57572521 on Build GRCh38
rs375281655 Chr.19: 57572682-57572682 on Build GRCh38
rs75651199 Chr.19: 57572338-57572338 on Build GRCh38
rs886466976 Chr.19: 57572559-57572559 on Build GRCh38
rs957007753 Chr.19: 57572341-57572341 on Build GRCh38
rs537011734 Chr.19: 57572323-57572323 on Build GRCh38
rs745832528 Chr.19: 57572604-57572604 on Build GRCh38
rs915970808 Chr.19: 57572519-57572519 on Build GRCh38
rs780069616 Chr.19: 57572732-57572734 on Build GRCh38
rs780463257 Chr.19: 57572398-57572398 on Build GRCh38
rs754031914 Chr.19: 57572686-57572686 on Build GRCh38
rs893414603 Chr.19: 57572737-57572737 on Build GRCh38
rs781214946 Chr.19: 57572600-57572600 on Build GRCh38
rs372568260 Chr.19: 57572503-57572503 on Build GRCh38
rs543250032 Chr.19: 57572343-57572343 on Build GRCh38
rs765421147 Chr.19: 57572697-57572697 on Build GRCh38
rs372660403 Chr.19: 57572712-57572712 on Build GRCh38
rs866246566 Chr.19: 57572681-57572681 on Build GRCh38
rs950549618 Chr.19: 57572485-57572485 on Build GRCh38
rs866444409 Chr.19: 57572623-57572623 on Build GRCh38
rs748842940 Chr.19: 57572756-57572756 on Build GRCh38
rs941322784 Chr.19: 57572499-57572499 on Build GRCh38
rs141943092 Chr.19: 57572512-57572512 on Build GRCh38
rs1017304540 Chr.19: 57572758-57572758 on Build GRCh38
rs770832508 Chr.19: 57572461-57572461 on Build GRCh38
rs748003734 Chr.19: 57572322-57572322 on Build GRCh38
rs769665166 Chr.19: 57572618-57572618 on Build GRCh38
rs762050977 Chr.19: 57572455-57572455 on Build GRCh38
rs577622214 Chr.19: 57572320-57572320 on Build GRCh38
rs759074213 Chr.19: 57572342-57572342 on Build GRCh38
rs200576929 Chr.19: 57572749-57572749 on Build GRCh38
rs781057139 Chr.19: 57572710-57572710 on Build GRCh38
rs747915397 Chr.19: 57572651-57572651 on Build GRCh38
rs150383989 Chr.19: 57572663-57572663 on Build GRCh38
rs867009043 Chr.19: 57572432-57572432 on Build GRCh38
rs750524836 Chr.19: 57572392-57572392 on Build GRCh38
rs762122403 Chr.19: 57572644-57572644 on Build GRCh38
rs200315346 Chr.19: 57572513-57572513 on Build GRCh38
rs867698187 Chr.19: 57572314-57572314 on Build GRCh38
rs900753030 Chr.19: 57572608-57572608 on Build GRCh38
rs752996956 Chr.19: 57572698-57572698 on Build GRCh38
rs768226707 Chr.19: 57572507-57572507 on Build GRCh38
rs867679924 Chr.19: 57572373-57572373 on Build GRCh38
rs545727805 Chr.19: 57572641-57572641 on Build GRCh38
rs762557103 Chr.19: 57572372-57572372 on Build GRCh38
rs781169978 Chr.19: 57572473-57572473 on Build GRCh38
rs766989850 Chr.19: 57572671-57572671 on Build GRCh38
rs371954172 Chr.19: 57572377-57572377 on Build GRCh38
rs111662574 Chr.19: 57572374-57572374 on Build GRCh38
rs768062028 Chr.19: 57572389-57572389 on Build GRCh38
rs1007955589 Chr.19: 57572705-57572705 on Build GRCh38
rs755318810 Chr.19: 57572549-57572549 on Build GRCh38
rs773331470 Chr.19: 57572313-57572313 on Build GRCh38
rs766588301 Chr.19: 57572536-57572536 on Build GRCh38
rs754478768 Chr.19: 57572760-57572760 on Build GRCh38
rs758505218 Chr.19: 57572709-57572709 on Build GRCh38
rs369719582 Chr.19: 57572523-57572523 on Build GRCh38
rs750874169 Chr.19: 57572544-57572573 on Build GRCh38
rs764468301 Chr.19: 57572376-57572376 on Build GRCh38
rs753904201 Chr.19: 57572537-57572537 on Build GRCh38
rs756106771 Chr.19: 57572724-57572724 on Build GRCh38
rs979395334 Chr.19: 57572361-57572361 on Build GRCh38
rs780812977 Chr.19: 57572587-57572587 on Build GRCh38
rs747858780 Chr.19: 57572773-57572773 on Build GRCh38
rs756619850 Chr.19: 57572579-57572584 on Build GRCh38

View Full Product Details