Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57753207-57753715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF776
Entrez Gene ID
284309
Gene Name
zinc finger protein 776
Gene Aliases
-
UniGene
Hs.720254
Gene Chromosome Location
Chr.19: 57746796-57758159 on Build GRCh38
Transcripts
NM_173632.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1590377 c.161-7C>A p.? Chr.19: 57753284-57753284 on Build GRCh38
1397173 c.233delA p.K80fs*31 Chr.19: 57753489-57753489 on Build GRCh38
1590375 c.417G>T p.E139D Chr.19: 57753547-57753547 on Build GRCh38
6038269 c.352G>A p.A118T Chr.19: 57753482-57753482 on Build GRCh38
3971313 c.76A>G p.T26A Chr.19: 57753332-57753332 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769475706 Chr.19: 57753532-57753532 on Build GRCh38
rs369025164 Chr.19: 57753539-57753539 on Build GRCh38
rs1011222390 Chr.19: 57753268-57753268 on Build GRCh38
rs771784384 Chr.19: 57753317-57753317 on Build GRCh38
rs983660752 Chr.19: 57753482-57753482 on Build GRCh38
rs751862092 Chr.19: 57753447-57753447 on Build GRCh38
rs745838415 Chr.19: 57753601-57753601 on Build GRCh38
rs542332791 Chr.19: 57753452-57753452 on Build GRCh38
rs753106467 Chr.19: 57753250-57753250 on Build GRCh38
rs781021790 Chr.19: 57753516-57753516 on Build GRCh38
rs779765582 Chr.19: 57753606-57753606 on Build GRCh38
rs777065864 Chr.19: 57753669-57753669 on Build GRCh38
rs767886595 Chr.19: 57753552-57753552 on Build GRCh38
rs769399644 Chr.19: 57753629-57753629 on Build GRCh38
rs745768250 Chr.19: 57753519-57753519 on Build GRCh38
rs754110429 Chr.19: 57753501-57753501 on Build GRCh38
rs930354440 Chr.19: 57753284-57753284 on Build GRCh38
rs776263066 Chr.19: 57753435-57753435 on Build GRCh38
rs750878978 Chr.19: 57753456-57753456 on Build GRCh38
rs775305722 Chr.19: 57753438-57753438 on Build GRCh38
rs747480521 Chr.19: 57753261-57753261 on Build GRCh38
rs371849767 Chr.19: 57753445-57753445 on Build GRCh38
rs375490927 Chr.19: 57753656-57753656 on Build GRCh38
rs766486964 Chr.19: 57753564-57753564 on Build GRCh38
rs757382664 Chr.19: 57753289-57753289 on Build GRCh38
rs748817322 Chr.19: 57753607-57753607 on Build GRCh38
rs762093594 Chr.19: 57753550-57753550 on Build GRCh38
rs1050580190 Chr.19: 57753313-57753313 on Build GRCh38
rs756221170 Chr.19: 57753471-57753471 on Build GRCh38
rs113199212 Chr.19: 57753368-57753368 on Build GRCh38
rs200360104 Chr.19: 57753324-57753324 on Build GRCh38
rs769934945 Chr.19: 57753536-57753536 on Build GRCh38
rs201309717 Chr.19: 57753383-57753383 on Build GRCh38
rs372947500 Chr.19: 57753594-57753594 on Build GRCh38
rs772427582 Chr.19: 57753675-57753676 on Build GRCh38
rs777691910 Chr.19: 57753505-57753505 on Build GRCh38
rs376101331 Chr.19: 57753514-57753514 on Build GRCh38
rs773991887 Chr.19: 57753541-57753541 on Build GRCh38
rs1012960233 Chr.19: 57753490-57753490 on Build GRCh38
rs773597287 Chr.19: 57753650-57753650 on Build GRCh38
rs779016907 Chr.19: 57753583-57753583 on Build GRCh38
rs576412938 Chr.19: 57753436-57753436 on Build GRCh38
rs1053388945 Chr.19: 57753237-57753237 on Build GRCh38
rs747005562 Chr.19: 57753409-57753409 on Build GRCh38
rs139142470 Chr.19: 57753251-57753251 on Build GRCh38
rs769907451 Chr.19: 57753315-57753315 on Build GRCh38
rs759643836 Chr.19: 57753670-57753670 on Build GRCh38
rs756125145 Chr.19: 57753379-57753379 on Build GRCh38
rs755162710 Chr.19: 57753503-57753503 on Build GRCh38
rs746768569 Chr.19: 57753506-57753506 on Build GRCh38
rs201807991 Chr.19: 57753258-57753258 on Build GRCh38
rs149185851 Chr.19: 57753611-57753611 on Build GRCh38
rs780256099 Chr.19: 57753380-57753380 on Build GRCh38
rs762670227 Chr.19: 57753444-57753444 on Build GRCh38
rs759299523 Chr.19: 57753335-57753335 on Build GRCh38
rs777519447 Chr.19: 57753399-57753399 on Build GRCh38
rs762436281 Chr.19: 57753448-57753448 on Build GRCh38
rs865965438 Chr.19: 57753682-57753682 on Build GRCh38
rs927830183 Chr.19: 57753228-57753228 on Build GRCh38
rs772781781 Chr.19: 57753639-57753639 on Build GRCh38
rs113708527 Chr.19: 57753355-57753355 on Build GRCh38
rs755252301 Chr.19: 57753568-57753568 on Build GRCh38
rs752331564 Chr.19: 57753344-57753344 on Build GRCh38
rs760477650 Chr.19: 57753323-57753323 on Build GRCh38
rs750259202 Chr.19: 57753488-57753489 on Build GRCh38
rs757023667 Chr.19: 57753591-57753591 on Build GRCh38
rs781113420 Chr.19: 57753292-57753292 on Build GRCh38
rs752910362 Chr.19: 57753680-57753680 on Build GRCh38
rs765310602 Chr.19: 57753674-57753674 on Build GRCh38
rs770768919 Chr.19: 57753412-57753412 on Build GRCh38
rs775971584 Chr.19: 57753328-57753328 on Build GRCh38
rs151216270 Chr.19: 57753352-57753352 on Build GRCh38
rs771874157 Chr.19: 57753397-57753397 on Build GRCh38
rs147594107 Chr.19: 57753497-57753497 on Build GRCh38
rs142022067 Chr.19: 57753540-57753540 on Build GRCh38
rs747947453 Chr.19: 57753316-57753316 on Build GRCh38
rs781403724 Chr.19: 57753369-57753369 on Build GRCh38
rs776341830 Chr.19: 57753293-57753293 on Build GRCh38
rs145378642 Chr.19: 57753571-57753571 on Build GRCh38
rs764994384 Chr.19: 57753341-57753341 on Build GRCh38
rs80320382 Chr.19: 57753480-57753480 on Build GRCh38
rs750918354 Chr.19: 57753553-57753553 on Build GRCh38
rs746144561 Chr.19: 57753298-57753298 on Build GRCh38
rs954600196 Chr.19: 57753690-57753690 on Build GRCh38
rs181594464 Chr.19: 57753658-57753658 on Build GRCh38
rs184557756 Chr.19: 57753320-57753320 on Build GRCh38
rs745927043 Chr.19: 57753370-57753370 on Build GRCh38
rs201024433 Chr.19: 57753343-57753343 on Build GRCh38
rs756113588 Chr.19: 57753603-57753603 on Build GRCh38
rs758612939 Chr.19: 57753691-57753691 on Build GRCh38
rs766858854 Chr.19: 57753662-57753662 on Build GRCh38
rs752923444 Chr.19: 57753589-57753589 on Build GRCh38
rs776147026 Chr.19: 57753687-57753687 on Build GRCh38
rs111750364 Chr.19: 57753566-57753566 on Build GRCh38
rs773911379 Chr.19: 57753614-57753614 on Build GRCh38

View Full Product Details