Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 58436956-58437456 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF132
Entrez Gene ID
7691
Gene Name
zinc finger protein 132
Gene Aliases
pHZ-12
UniGene
Hs.156169
Gene Chromosome Location
Chr.19: 58432814-58440222 on Build GRCh38
Transcripts
NM_003433.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3797642 c.177G>C p.Q59H Chr.19: 58437102-58437102 on Build GRCh38
1002731 c.223A>G p.T75A Chr.19: 58437056-58437056 on Build GRCh38
4478585 c.224C>T p.T75I Chr.19: 58437055-58437055 on Build GRCh38
4817521 c.153G>A p.E51E Chr.19: 58437126-58437126 on Build GRCh38
3673043 c.150G>T p.E50D Chr.19: 58437129-58437129 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770497613 Chr.19: 58437073-58437073 on Build GRCh38
rs950298551 Chr.19: 58437023-58437023 on Build GRCh38
rs781515273 Chr.19: 58437090-58437090 on Build GRCh38
rs765002621 Chr.19: 58437169-58437169 on Build GRCh38
rs76048958 Chr.19: 58437145-58437145 on Build GRCh38
rs199860736 Chr.19: 58437174-58437174 on Build GRCh38
rs765209327 Chr.19: 58437260-58437260 on Build GRCh38
rs763615450 Chr.19: 58437225-58437228 on Build GRCh38
rs1043632356 Chr.19: 58437274-58437274 on Build GRCh38
rs545353587 Chr.19: 58437301-58437301 on Build GRCh38
rs370065697 Chr.19: 58437211-58437211 on Build GRCh38
rs767665317 Chr.19: 58437142-58437142 on Build GRCh38
rs553399013 Chr.19: 58437195-58437195 on Build GRCh38
rs141827864 Chr.19: 58437221-58437221 on Build GRCh38
rs756755255 Chr.19: 58437217-58437217 on Build GRCh38
rs759464105 Chr.19: 58437259-58437259 on Build GRCh38
rs201404130 Chr.19: 58437040-58437040 on Build GRCh38
rs765023777 Chr.19: 58436996-58436996 on Build GRCh38
rs753651752 Chr.19: 58437155-58437155 on Build GRCh38
rs770228284 Chr.19: 58437093-58437093 on Build GRCh38
rs775792083 Chr.19: 58437000-58437000 on Build GRCh38
rs777356204 Chr.19: 58437197-58437197 on Build GRCh38
rs749716895 Chr.19: 58437234-58437234 on Build GRCh38
rs192896475 Chr.19: 58437028-58437028 on Build GRCh38
rs1019228148 Chr.19: 58437136-58437136 on Build GRCh38
rs532400800 Chr.19: 58437329-58437329 on Build GRCh38
rs199724943 Chr.19: 58437015-58437015 on Build GRCh38
rs746395623 Chr.19: 58437091-58437091 on Build GRCh38
rs761746270 Chr.19: 58437118-58437118 on Build GRCh38
rs753832067 Chr.19: 58437038-58437038 on Build GRCh38
rs1040584394 Chr.19: 58437357-58437357 on Build GRCh38
rs562607098 Chr.19: 58437406-58437406 on Build GRCh38
rs139059426 Chr.19: 58437051-58437051 on Build GRCh38
rs141889506 Chr.19: 58437179-58437179 on Build GRCh38
rs148123600 Chr.19: 58437163-58437163 on Build GRCh38
rs756659604 Chr.19: 58437203-58437203 on Build GRCh38
rs972087936 Chr.19: 58437296-58437296 on Build GRCh38
rs760074980 Chr.19: 58437340-58437340 on Build GRCh38
rs200726338 Chr.19: 58437240-58437240 on Build GRCh38
rs1008373106 Chr.19: 58437344-58437344 on Build GRCh38
rs561159599 Chr.19: 58437095-58437095 on Build GRCh38
rs940127225 Chr.19: 58437041-58437041 on Build GRCh38
rs76156338 Chr.19: 58437243-58437243 on Build GRCh38
rs759771584 Chr.19: 58436984-58436984 on Build GRCh38
rs778717641 Chr.19: 58437102-58437102 on Build GRCh38
rs774464256 Chr.19: 58437115-58437115 on Build GRCh38
rs752682135 Chr.19: 58437037-58437038 on Build GRCh38
rs915575204 Chr.19: 58437294-58437294 on Build GRCh38
rs773397107 Chr.19: 58437246-58437246 on Build GRCh38
rs758103270 Chr.19: 58437055-58437055 on Build GRCh38
rs759428485 Chr.19: 58436992-58436992 on Build GRCh38
rs751257425 Chr.19: 58437201-58437201 on Build GRCh38
rs769038063 Chr.19: 58437106-58437106 on Build GRCh38
rs202147179 Chr.19: 58437036-58437036 on Build GRCh38
rs774392140 Chr.19: 58437239-58437239 on Build GRCh38
rs1008813001 Chr.19: 58437112-58437112 on Build GRCh38
rs897986013 Chr.19: 58437358-58437358 on Build GRCh38
rs1027689277 Chr.19: 58437086-58437086 on Build GRCh38
rs769160945 Chr.19: 58437235-58437235 on Build GRCh38
rs889034367 Chr.19: 58437109-58437109 on Build GRCh38
rs1038471141 Chr.19: 58437342-58437342 on Build GRCh38
rs920017861 Chr.19: 58437347-58437347 on Build GRCh38
rs760627533 Chr.19: 58437021-58437021 on Build GRCh38
rs775430796 Chr.19: 58437266-58437266 on Build GRCh38
rs763259968 Chr.19: 58437099-58437099 on Build GRCh38
rs746349878 Chr.19: 58437216-58437216 on Build GRCh38
rs775234080 Chr.19: 58437125-58437125 on Build GRCh38
rs576916320 Chr.19: 58437011-58437011 on Build GRCh38
rs991131610 Chr.19: 58436997-58436997 on Build GRCh38
rs777032589 Chr.19: 58436979-58436979 on Build GRCh38
rs777235591 Chr.19: 58437062-58437062 on Build GRCh38
rs749580045 Chr.19: 58437221-58437224 on Build GRCh38
rs142948922 Chr.19: 58437068-58437068 on Build GRCh38
rs770229673 Chr.19: 58436990-58436990 on Build GRCh38
rs993632239 Chr.19: 58437389-58437389 on Build GRCh38
rs373221733 Chr.19: 58437014-58437014 on Build GRCh38
rs373902742 Chr.19: 58437050-58437050 on Build GRCh38
rs763284255 Chr.19: 58437002-58437002 on Build GRCh38
rs775088403 Chr.19: 58437300-58437300 on Build GRCh38

View Full Product Details