Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 58477932-58478385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF446
Entrez Gene ID
55663
Gene Name
zinc finger protein 446
Gene Aliases
ZKSCAN20, ZSCAN30, ZSCAN52
UniGene
Hs.710284
Gene Chromosome Location
Chr.19: 58475855-58489533 on Build GRCh38
Transcripts
NM_001304453.1 NM_017908.3 XM_005259052.4 XM_005259053.4 XM_006723266.3 XM_017026949.1 XM_017026950.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6003781 c.627+8G>A p.? Chr.19: 58478189-58478189 on Build GRCh38
4082441 c.575A>G p.N192S Chr.19: 58478129-58478129 on Build GRCh38
4001036 c.574A>C p.N192H Chr.19: 58478128-58478128 on Build GRCh38
4844151 c.621G>A p.R207R Chr.19: 58478175-58478175 on Build GRCh38
3540795 c.576C>T p.N192N Chr.19: 58478130-58478130 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770977056 Chr.19: 58478192-58478192 on Build GRCh38
rs955920782 Chr.19: 58478001-58478001 on Build GRCh38
rs766878658 Chr.19: 58478160-58478160 on Build GRCh38
rs763804131 Chr.19: 58478151-58478151 on Build GRCh38
rs955972978 Chr.19: 58478254-58478254 on Build GRCh38
rs1040370073 Chr.19: 58478309-58478309 on Build GRCh38
rs748263163 Chr.19: 58478214-58478214 on Build GRCh38
rs539349658 Chr.19: 58478236-58478236 on Build GRCh38
rs749622182 Chr.19: 58478190-58478190 on Build GRCh38
rs758083195 Chr.19: 58478075-58478075 on Build GRCh38
rs752543886 Chr.19: 58478073-58478073 on Build GRCh38
rs924549196 Chr.19: 58478000-58478000 on Build GRCh38
rs77446414 Chr.19: 58478125-58478125 on Build GRCh38
rs868419441 Chr.19: 58477996-58477996 on Build GRCh38
rs780358614 Chr.19: 58478106-58478106 on Build GRCh38
rs550089034 Chr.19: 58478322-58478322 on Build GRCh38
rs938373587 Chr.19: 58478090-58478090 on Build GRCh38
rs934326306 Chr.19: 58478007-58478007 on Build GRCh38
rs757297472 Chr.19: 58478089-58478089 on Build GRCh38
rs139673627 Chr.19: 58478139-58478139 on Build GRCh38
rs928433094 Chr.19: 58478348-58478348 on Build GRCh38
rs762887094 Chr.19: 58478150-58478150 on Build GRCh38
rs368246732 Chr.19: 58478226-58478226 on Build GRCh38
rs750675997 Chr.19: 58478190-58478190 on Build GRCh38
rs746417197 Chr.19: 58478096-58478096 on Build GRCh38
rs1004727156 Chr.19: 58478356-58478356 on Build GRCh38
rs773358170 Chr.19: 58478138-58478138 on Build GRCh38
rs372284895 Chr.19: 58478124-58478124 on Build GRCh38
rs775673931 Chr.19: 58478037-58478037 on Build GRCh38
rs777590204 Chr.19: 58478084-58478084 on Build GRCh38
rs1051298799 Chr.19: 58478331-58478331 on Build GRCh38
rs751237339 Chr.19: 58478155-58478155 on Build GRCh38
rs978476518 Chr.19: 58477995-58477995 on Build GRCh38
rs763522218 Chr.19: 58478040-58478041 on Build GRCh38
rs769828755 Chr.19: 58478229-58478229 on Build GRCh38
rs768899913 Chr.19: 58478041-58478041 on Build GRCh38
rs761852342 Chr.19: 58478052-58478052 on Build GRCh38
rs921394566 Chr.19: 58478314-58478314 on Build GRCh38
rs900014327 Chr.19: 58478111-58478112 on Build GRCh38
rs781486524 Chr.19: 58478351-58478351 on Build GRCh38
rs749679297 Chr.19: 58478109-58478109 on Build GRCh38
rs148708008 Chr.19: 58478087-58478087 on Build GRCh38
rs781718561 Chr.19: 58478089-58478089 on Build GRCh38
rs990073762 Chr.19: 58478256-58478256 on Build GRCh38
rs893185 Chr.19: 58478128-58478128 on Build GRCh38
rs759363087 Chr.19: 58478059-58478059 on Build GRCh38
rs111330625 Chr.19: 58477990-58477990 on Build GRCh38
rs774369730 Chr.19: 58478051-58478051 on Build GRCh38
rs546492259 Chr.19: 58478014-58478016 on Build GRCh38
rs547175483 Chr.19: 58478140-58478140 on Build GRCh38
rs756777272 Chr.19: 58478159-58478159 on Build GRCh38
rs780589148 Chr.19: 58478187-58478187 on Build GRCh38
rs368910040 Chr.19: 58478111-58478111 on Build GRCh38
rs764574275 Chr.19: 58478053-58478054 on Build GRCh38
rs763096857 Chr.19: 58478039-58478039 on Build GRCh38
rs756655988 Chr.19: 58478102-58478102 on Build GRCh38
rs370033116 Chr.19: 58478043-58478043 on Build GRCh38
rs764884458 Chr.19: 58478067-58478067 on Build GRCh38
rs990021174 Chr.19: 58478017-58478017 on Build GRCh38
rs914243512 Chr.19: 58478282-58478282 on Build GRCh38
rs751644473 Chr.19: 58478059-58478060 on Build GRCh38
rs989814315 Chr.19: 58478267-58478267 on Build GRCh38
rs731259 Chr.19: 58478057-58478057 on Build GRCh38
rs561870851 Chr.19: 58478078-58478078 on Build GRCh38
rs773123974 Chr.19: 58478218-58478218 on Build GRCh38
rs779098105 Chr.19: 58478211-58478211 on Build GRCh38
rs775373065 Chr.19: 58478146-58478146 on Build GRCh38
rs980313080 Chr.19: 58478341-58478341 on Build GRCh38
rs924357492 Chr.19: 58478253-58478255 on Build GRCh38
rs1007433237 Chr.19: 58478168-58478168 on Build GRCh38
rs931563495 Chr.19: 58478133-58478133 on Build GRCh38
rs755407529 Chr.19: 58478196-58478196 on Build GRCh38
rs151261644 Chr.19: 58478130-58478130 on Build GRCh38
rs1021511227 Chr.19: 58478293-58478293 on Build GRCh38
rs756531418 Chr.19: 58478186-58478186 on Build GRCh38
rs375347224 Chr.19: 58478216-58478216 on Build GRCh38
rs1054564501 Chr.19: 58478014-58478014 on Build GRCh38
rs749982843 Chr.19: 58478184-58478184 on Build GRCh38
rs914399121 Chr.19: 58478019-58478019 on Build GRCh38
rs781409891 Chr.19: 58478120-58478121 on Build GRCh38
rs763903992 Chr.19: 58477985-58477985 on Build GRCh38

View Full Product Details