Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 1791739-1792241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYT1L
Entrez Gene ID
23040
Gene Name
myelin transcription factor 1 like
Gene Aliases
MRD39, NZF1, ZC2H2C2, ZC2HC4B
UniGene
Hs.434418
Gene Chromosome Location
Chr.2: 1789113-2331388 on Build GRCh38
Transcripts
NM_001303052.1 NM_015025.3 XM_011510318.1 XM_011510319.1 XM_011510320.2 XM_011510321.2 XM_011510322.2 XM_011510323.1 XM_011510324.1 XM_011510325.1 XM_011510326.1 XM_011510327.1 XM_011510328.2 XM_017003604.1 XM_017003605.1 XM_017003606.1 XM_017003607.1 XM_017003609.1 XM_017003610.1 XM_017003611.1 XM_017003612.1 XM_017003613.1 XM_017003614.1 XM_017003615.1 XM_017003616.1 XM_017003617.1 XM_017003618.1 XM_017003619.1 XM_017003620.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7224070 c.459G>C p.L153F Chr.2: 1791963-1791963 on Build GRCh38
3798326 c.3498G>C p.Q1166H Chr.2: 1791930-1791930 on Build GRCh38
5834327 c.490C>G p.Q164E Chr.2: 1791932-1791932 on Build GRCh38
6054736 c.3427G>T p.E1143* Chr.2: 1791995-1791995 on Build GRCh38
6222412 c.3485G>A p.R1162H Chr.2: 1791937-1791937 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34259720 Chr.2: 1792140-1792141 on Build GRCh38
rs978806752 Chr.2: 1792066-1792066 on Build GRCh38
rs1055970058 Chr.2: 1792054-1792054 on Build GRCh38
rs753741286 Chr.2: 1791856-1791857 on Build GRCh38
rs759998216 Chr.2: 1791997-1791997 on Build GRCh38
rs764343593 Chr.2: 1792001-1792001 on Build GRCh38
rs754552615 Chr.2: 1791864-1791864 on Build GRCh38
rs549304346 Chr.2: 1791853-1791853 on Build GRCh38
rs777864846 Chr.2: 1791834-1791834 on Build GRCh38
rs747777960 Chr.2: 1791923-1791923 on Build GRCh38
rs1051725480 Chr.2: 1791814-1791814 on Build GRCh38
rs749800637 Chr.2: 1791960-1791960 on Build GRCh38
rs1016543178 Chr.2: 1792183-1792183 on Build GRCh38
rs772863694 Chr.2: 1791938-1791938 on Build GRCh38
rs370800836 Chr.2: 1791843-1791843 on Build GRCh38
rs775893555 Chr.2: 1792117-1792117 on Build GRCh38
rs994783337 Chr.2: 1792209-1792209 on Build GRCh38
rs756173319 Chr.2: 1791823-1791823 on Build GRCh38
rs756789003 Chr.2: 1792046-1792046 on Build GRCh38
rs200657355 Chr.2: 1791886-1791886 on Build GRCh38
rs1024570209 Chr.2: 1792123-1792123 on Build GRCh38
rs771632725 Chr.2: 1791961-1791961 on Build GRCh38
rs1004619562 Chr.2: 1792161-1792161 on Build GRCh38
rs1002227291 Chr.2: 1792044-1792044 on Build GRCh38
rs933303393 Chr.2: 1791790-1791790 on Build GRCh38
rs780946663 Chr.2: 1791872-1791872 on Build GRCh38
rs944521961 Chr.2: 1792125-1792125 on Build GRCh38
rs527917269 Chr.2: 1791799-1791799 on Build GRCh38
rs913314468 Chr.2: 1791930-1791930 on Build GRCh38
rs530061364 Chr.2: 1792207-1792207 on Build GRCh38
rs950540688 Chr.2: 1792124-1792124 on Build GRCh38
rs758342649 Chr.2: 1791847-1791847 on Build GRCh38
rs1013213851 Chr.2: 1792102-1792102 on Build GRCh38
rs531865082 Chr.2: 1792194-1792194 on Build GRCh38
rs760813830 Chr.2: 1792042-1792042 on Build GRCh38
rs946122650 Chr.2: 1791946-1791946 on Build GRCh38
rs896033507 Chr.2: 1792092-1792092 on Build GRCh38
rs779808153 Chr.2: 1791850-1791850 on Build GRCh38
rs1043635336 Chr.2: 1791957-1791957 on Build GRCh38
rs774825001 Chr.2: 1791974-1791974 on Build GRCh38
rs186589929 Chr.2: 1792105-1792105 on Build GRCh38
rs769387462 Chr.2: 1791928-1791928 on Build GRCh38

View Full Product Details