Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 3580565-3581071 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS7
Entrez Gene ID
6201
Gene Name
ribosomal protein S7
Gene Aliases
DBA8, S7
UniGene
Hs.534346
Gene Chromosome Location
Chr.2: 3575263-3580919 on Build GRCh38
Transcripts
NM_001011.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3581351 c.569C>A p.P190Q Chr.2: 3580866-3580866 on Build GRCh38
7158418 c.511G>A p.E171K Chr.2: 3580808-3580808 on Build GRCh38
4499471 c.542C>T p.T181M Chr.2: 3580839-3580839 on Build GRCh38
5032001 c.528C>G p.V176V Chr.2: 3580825-3580825 on Build GRCh38
3581350 c.523G>A p.G175S Chr.2: 3580820-3580820 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs899834416 Chr.2: 3580958-3580958 on Build GRCh38
rs560628799 Chr.2: 3580939-3580939 on Build GRCh38
rs34789745 Chr.2: 3580591-3580592 on Build GRCh38
rs112774737 Chr.2: 3580993-3580993 on Build GRCh38
rs991837840 Chr.2: 3580737-3580737 on Build GRCh38
rs746435863 Chr.2: 3580800-3580800 on Build GRCh38
rs1030739238 Chr.2: 3580715-3580715 on Build GRCh38
rs773757917 Chr.2: 3580884-3580884 on Build GRCh38
rs776954129 Chr.2: 3580840-3580840 on Build GRCh38
rs897162807 Chr.2: 3580660-3580660 on Build GRCh38
rs1039700406 Chr.2: 3580941-3580941 on Build GRCh38
rs534558419 Chr.2: 3580617-3580617 on Build GRCh38
rs552693637 Chr.2: 3581037-3581037 on Build GRCh38
rs1047151498 Chr.2: 3580638-3580638 on Build GRCh38
rs149236744 Chr.2: 3580988-3580988 on Build GRCh38
rs914955808 Chr.2: 3580827-3580827 on Build GRCh38
rs770248786 Chr.2: 3580866-3580866 on Build GRCh38
rs535285909 Chr.2: 3580736-3580736 on Build GRCh38
rs994180622 Chr.2: 3580661-3580661 on Build GRCh38
rs541050041 Chr.2: 3580620-3580620 on Build GRCh38
rs10793 Chr.2: 3580862-3580862 on Build GRCh38
rs572802131 Chr.2: 3580957-3580957 on Build GRCh38
rs191506876 Chr.2: 3580913-3580913 on Build GRCh38
rs1007119207 Chr.2: 3580684-3580684 on Build GRCh38
rs947596429 Chr.2: 3580958-3580958 on Build GRCh38
rs772565785 Chr.2: 3580683-3580683 on Build GRCh38
rs919734855 Chr.2: 3580839-3580839 on Build GRCh38
rs891091515 Chr.2: 3580640-3580640 on Build GRCh38
rs184336880 Chr.2: 3581039-3581039 on Build GRCh38
rs528482260 Chr.2: 3581048-3581048 on Build GRCh38
rs1030633706 Chr.2: 3580744-3580744 on Build GRCh38
rs14742 Chr.2: 3580818-3580818 on Build GRCh38
rs113112388 Chr.2: 3580739-3580739 on Build GRCh38
rs903467467 Chr.2: 3580965-3580965 on Build GRCh38
rs59060135 Chr.2: 3580608-3580608 on Build GRCh38
rs541707217 Chr.2: 3580972-3580972 on Build GRCh38
rs753839949 Chr.2: 3580757-3580757 on Build GRCh38
rs546281280 Chr.2: 3580994-3580994 on Build GRCh38
rs528925798 Chr.2: 3580722-3580722 on Build GRCh38
rs1057034808 Chr.2: 3580657-3580657 on Build GRCh38
rs1027021190 Chr.2: 3580672-3580672 on Build GRCh38
rs938895045 Chr.2: 3580953-3580953 on Build GRCh38
rs774730828 Chr.2: 3580920-3580920 on Build GRCh38
rs539208774 Chr.2: 3580789-3580789 on Build GRCh38
rs553721392 Chr.2: 3580978-3580978 on Build GRCh38
rs532510798 Chr.2: 3580992-3580992 on Build GRCh38
rs983048833 Chr.2: 3580864-3580864 on Build GRCh38
rs748888092 Chr.2: 3580922-3580923 on Build GRCh38
rs999007789 Chr.2: 3580980-3580980 on Build GRCh38
rs976684410 Chr.2: 3580743-3580743 on Build GRCh38
rs1040132443 Chr.2: 3580652-3580652 on Build GRCh38
rs755591294 Chr.2: 3580916-3580917 on Build GRCh38
rs967558502 Chr.2: 3580684-3580686 on Build GRCh38
rs916485183 Chr.2: 3580591-3580591 on Build GRCh38
rs201281565 Chr.2: 3580798-3580798 on Build GRCh38
rs747706059 Chr.2: 3580787-3580790 on Build GRCh38
rs1057326108 Chr.2: 3580963-3580963 on Build GRCh38
rs563943008 Chr.2: 3580991-3580991 on Build GRCh38
rs777435945 Chr.2: 3580920-3580921 on Build GRCh38
rs897448285 Chr.2: 3580979-3580979 on Build GRCh38
rs748715934 Chr.2: 3580841-3580841 on Build GRCh38
rs575059031 Chr.2: 3580987-3580987 on Build GRCh38
rs778461388 Chr.2: 3580618-3580618 on Build GRCh38
rs764394331 Chr.2: 3580849-3580849 on Build GRCh38
rs187061405 Chr.2: 3580665-3580665 on Build GRCh38
rs763251793 Chr.2: 3580890-3580890 on Build GRCh38
rs980646629 Chr.2: 3580934-3580934 on Build GRCh38
rs2071639 Chr.2: 3580793-3580793 on Build GRCh38
rs906094564 Chr.2: 3580619-3580619 on Build GRCh38
rs768225488 Chr.2: 3580805-3580805 on Build GRCh38
rs1008215576 Chr.2: 3580646-3580646 on Build GRCh38
rs989184252 Chr.2: 3580821-3580821 on Build GRCh38

View Full Product Details