Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 8683990-8684500 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ID2
Entrez Gene ID
3398
Gene Name
inhibitor of DNA binding 2, HLH protein
Gene Aliases
GIG8, ID2A, ID2H, bHLHb26
UniGene
Hs.180919
Gene Chromosome Location
Chr.2: 8681983-8684453 on Build GRCh38
Transcripts
NM_002166.4

SNP Detail

SNP ID Chromosome Location
rs1054240102 Chr.2: 8684457-8684457 on Build GRCh38
rs150333363 Chr.2: 8684340-8684341 on Build GRCh38
rs868005555 Chr.2: 8684135-8684135 on Build GRCh38
rs955700680 Chr.2: 8684040-8684040 on Build GRCh38
rs574798390 Chr.2: 8684030-8684030 on Build GRCh38
rs943678400 Chr.2: 8684433-8684433 on Build GRCh38
rs1015482299 Chr.2: 8684464-8684464 on Build GRCh38
rs925510168 Chr.2: 8684373-8684373 on Build GRCh38
rs543520892 Chr.2: 8684063-8684063 on Build GRCh38
rs892162200 Chr.2: 8684020-8684020 on Build GRCh38
rs13401658 Chr.2: 8684240-8684240 on Build GRCh38
rs561735228 Chr.2: 8684024-8684024 on Build GRCh38
rs938169886 Chr.2: 8684101-8684101 on Build GRCh38
rs577296674 Chr.2: 8684219-8684219 on Build GRCh38
rs1037543763 Chr.2: 8684424-8684425 on Build GRCh38
rs1050114924 Chr.2: 8684210-8684210 on Build GRCh38
rs184949920 Chr.2: 8684238-8684238 on Build GRCh38
rs1025027599 Chr.2: 8684341-8684341 on Build GRCh38
rs926783899 Chr.2: 8684095-8684095 on Build GRCh38
rs56050726 Chr.2: 8684422-8684422 on Build GRCh38
rs982788644 Chr.2: 8684092-8684094 on Build GRCh38
rs373341223 Chr.2: 8684240-8684243 on Build GRCh38
rs375466228 Chr.2: 8684382-8684382 on Build GRCh38
rs781685596 Chr.2: 8684337-8684337 on Build GRCh38
rs748235392 Chr.2: 8684171-8684174 on Build GRCh38
rs939998187 Chr.2: 8684139-8684139 on Build GRCh38
rs1026778380 Chr.2: 8684128-8684128 on Build GRCh38
rs147763639 Chr.2: 8684439-8684439 on Build GRCh38
rs949684920 Chr.2: 8684092-8684092 on Build GRCh38
rs955818332 Chr.2: 8684247-8684247 on Build GRCh38
rs922593048 Chr.2: 8684428-8684428 on Build GRCh38
rs56114166 Chr.2: 8684174-8684177 on Build GRCh38
rs996601385 Chr.2: 8684125-8684125 on Build GRCh38
rs1040749488 Chr.2: 8684079-8684079 on Build GRCh38
rs888359269 Chr.2: 8684132-8684132 on Build GRCh38
rs192055392 Chr.2: 8684204-8684204 on Build GRCh38
rs988470415 Chr.2: 8684043-8684043 on Build GRCh38
rs140487184 Chr.2: 8684064-8684068 on Build GRCh38
rs984325448 Chr.2: 8684388-8684388 on Build GRCh38
rs988436839 Chr.2: 8684417-8684417 on Build GRCh38
rs1004360299 Chr.2: 8684473-8684473 on Build GRCh38
rs899605677 Chr.2: 8684105-8684105 on Build GRCh38
rs909958590 Chr.2: 8684389-8684392 on Build GRCh38
rs899553506 Chr.2: 8684442-8684442 on Build GRCh38
rs527712287 Chr.2: 8684108-8684108 on Build GRCh38
rs969462019 Chr.2: 8684345-8684345 on Build GRCh38
rs371228469 Chr.2: 8684188-8684188 on Build GRCh38
rs56392909 Chr.2: 8684208-8684209 on Build GRCh38
rs1037066212 Chr.2: 8684140-8684140 on Build GRCh38
rs932443603 Chr.2: 8684444-8684444 on Build GRCh38
rs1000021926 Chr.2: 8684237-8684237 on Build GRCh38
rs936908465 Chr.2: 8684403-8684403 on Build GRCh38
rs762424092 Chr.2: 8684303-8684303 on Build GRCh38
rs959526220 Chr.2: 8684382-8684383 on Build GRCh38
rs189748725 Chr.2: 8684456-8684456 on Build GRCh38
rs992173044 Chr.2: 8684221-8684221 on Build GRCh38
rs755302298 Chr.2: 8684054-8684054 on Build GRCh38
rs982353060 Chr.2: 8684371-8684375 on Build GRCh38
rs563781764 Chr.2: 8684229-8684231 on Build GRCh38
rs998091229 Chr.2: 8684177-8684178 on Build GRCh38
rs914126176 Chr.2: 8684073-8684073 on Build GRCh38
rs577807398 Chr.2: 8684139-8684144 on Build GRCh38
rs1004471687 Chr.2: 8684459-8684459 on Build GRCh38
rs980932018 Chr.2: 8684372-8684372 on Build GRCh38
rs971484370 Chr.2: 8684348-8684348 on Build GRCh38
rs1048470546 Chr.2: 8684116-8684116 on Build GRCh38
rs1009837922 Chr.2: 8684303-8684303 on Build GRCh38
rs889586230 Chr.2: 8684229-8684229 on Build GRCh38
rs527938224 Chr.2: 8684461-8684461 on Build GRCh38
rs1048689180 Chr.2: 8684455-8684455 on Build GRCh38
rs901069258 Chr.2: 8684148-8684148 on Build GRCh38
rs926871010 Chr.2: 8684375-8684376 on Build GRCh38

View Full Product Details