Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 11192006-11192523 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ROCK2
Entrez Gene ID
9475
Gene Name
Rho associated coiled-coil containing protein kinase 2
Gene Aliases
ROCK-II
UniGene
Hs.681743
Gene Chromosome Location
Chr.2: 11179759-11345407 on Build GRCh38
Transcripts
NM_001321643.1 NM_004850.4 XM_005246190.3 XM_011510417.2 XM_017005378.1 XM_017005379.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3960992 c.4082C>T p.S1361L Chr.2: 11192229-11192229 on Build GRCh38
2157015 c.4136G>A p.S1379N Chr.2: 11192175-11192175 on Build GRCh38
3565342 c.4070C>T p.P1357L Chr.2: 11192241-11192241 on Build GRCh38
6362199 c.3888C>A p.C1296* Chr.2: 11192512-11192512 on Build GRCh38
1398807 c.4052delA p.K1351fs*17 Chr.2: 11192259-11192259 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200299225 Chr.2: 11192431-11192431 on Build GRCh38
rs751806275 Chr.2: 11192251-11192251 on Build GRCh38
rs768685944 Chr.2: 11192126-11192126 on Build GRCh38
rs748794221 Chr.2: 11192146-11192146 on Build GRCh38
rs748832043 Chr.2: 11192113-11192114 on Build GRCh38
rs371469035 Chr.2: 11192365-11192365 on Build GRCh38
rs1039258337 Chr.2: 11192113-11192113 on Build GRCh38
rs759293945 Chr.2: 11192180-11192180 on Build GRCh38
rs777992763 Chr.2: 11192425-11192425 on Build GRCh38
rs978117111 Chr.2: 11192068-11192068 on Build GRCh38
rs763583939 Chr.2: 11192192-11192192 on Build GRCh38
rs766761794 Chr.2: 11192250-11192250 on Build GRCh38
rs1053880825 Chr.2: 11192088-11192088 on Build GRCh38
rs770411219 Chr.2: 11192128-11192130 on Build GRCh38
rs766740594 Chr.2: 11192376-11192376 on Build GRCh38
rs761424684 Chr.2: 11192396-11192396 on Build GRCh38
rs780178584 Chr.2: 11192115-11192115 on Build GRCh38
rs971004382 Chr.2: 11192413-11192413 on Build GRCh38
rs769221330 Chr.2: 11192439-11192439 on Build GRCh38
rs923930866 Chr.2: 11192078-11192078 on Build GRCh38
rs374585992 Chr.2: 11192389-11192389 on Build GRCh38
rs756095805 Chr.2: 11192113-11192113 on Build GRCh38
rs758366619 Chr.2: 11192443-11192443 on Build GRCh38
rs760901332 Chr.2: 11192378-11192379 on Build GRCh38
rs755243536 Chr.2: 11192252-11192252 on Build GRCh38
rs771818299 Chr.2: 11192366-11192367 on Build GRCh38
rs757763750 Chr.2: 11192100-11192100 on Build GRCh38
rs748181951 Chr.2: 11192137-11192137 on Build GRCh38
rs779333705 Chr.2: 11192102-11192102 on Build GRCh38
rs758748287 Chr.2: 11192232-11192232 on Build GRCh38
rs764703058 Chr.2: 11192349-11192349 on Build GRCh38
rs936778339 Chr.2: 11192079-11192079 on Build GRCh38
rs752949053 Chr.2: 11192408-11192408 on Build GRCh38
rs970950658 Chr.2: 11192065-11192065 on Build GRCh38
rs374630605 Chr.2: 11192172-11192172 on Build GRCh38
rs756323979 Chr.2: 11192424-11192424 on Build GRCh38
rs746721665 Chr.2: 11192327-11192327 on Build GRCh38
rs1025135510 Chr.2: 11192392-11192392 on Build GRCh38
rs748271839 Chr.2: 11192266-11192266 on Build GRCh38
rs763520895 Chr.2: 11192372-11192372 on Build GRCh38
rs200094303 Chr.2: 11192418-11192418 on Build GRCh38
rs745564499 Chr.2: 11192167-11192168 on Build GRCh38
rs761352733 Chr.2: 11192344-11192344 on Build GRCh38
rs367590981 Chr.2: 11192342-11192342 on Build GRCh38
rs775328443 Chr.2: 11192343-11192343 on Build GRCh38
rs376590769 Chr.2: 11192186-11192186 on Build GRCh38
rs776128031 Chr.2: 11192201-11192201 on Build GRCh38
rs201188133 Chr.2: 11192129-11192129 on Build GRCh38
rs754406833 Chr.2: 11192099-11192099 on Build GRCh38
rs1047418299 Chr.2: 11192277-11192277 on Build GRCh38
rs376149475 Chr.2: 11192326-11192326 on Build GRCh38
rs374395637 Chr.2: 11192278-11192278 on Build GRCh38
rs750898507 Chr.2: 11192223-11192223 on Build GRCh38
rs960536932 Chr.2: 11192311-11192311 on Build GRCh38
rs200825805 Chr.2: 11192212-11192212 on Build GRCh38
rs761262690 Chr.2: 11192204-11192204 on Build GRCh38
rs771825593 Chr.2: 11192290-11192290 on Build GRCh38
rs892184561 Chr.2: 11192055-11192055 on Build GRCh38
rs747242470 Chr.2: 11192123-11192123 on Build GRCh38
rs560570357 Chr.2: 11192397-11192397 on Build GRCh38
rs750806790 Chr.2: 11192107-11192107 on Build GRCh38
rs771017362 Chr.2: 11192161-11192161 on Build GRCh38
rs758749627 Chr.2: 11192111-11192111 on Build GRCh38
rs777511531 Chr.2: 11192112-11192113 on Build GRCh38
rs778552121 Chr.2: 11192141-11192143 on Build GRCh38
rs770509824 Chr.2: 11192214-11192214 on Build GRCh38
rs1008636707 Chr.2: 11192030-11192030 on Build GRCh38
rs751977482 Chr.2: 11192395-11192395 on Build GRCh38
rs371119461 Chr.2: 11192273-11192273 on Build GRCh38
rs781518081 Chr.2: 11192255-11192255 on Build GRCh38
rs201801097 Chr.2: 11192296-11192296 on Build GRCh38
rs911264045 Chr.2: 11192092-11192092 on Build GRCh38
rs745801053 Chr.2: 11192283-11192283 on Build GRCh38
rs773775703 Chr.2: 11192358-11192358 on Build GRCh38
rs779803586 Chr.2: 11192480-11192480 on Build GRCh38
rs1016022786 Chr.2: 11192335-11192335 on Build GRCh38
rs760366234 Chr.2: 11192190-11192190 on Build GRCh38
rs561135077 Chr.2: 11192379-11192379 on Build GRCh38
rs1040110165 Chr.2: 11192051-11192051 on Build GRCh38
rs2230775 Chr.2: 11192282-11192282 on Build GRCh38
rs968013894 Chr.2: 11192253-11192253 on Build GRCh38

View Full Product Details