Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 11658100-11658609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NTSR2
Entrez Gene ID
23620
Gene Name
neurotensin receptor 2
Gene Aliases
NTR2
UniGene
Hs.131138
Gene Chromosome Location
Chr.2: 11658178-11670206 on Build GRCh38
Transcripts
NM_012344.3 XM_005246156.3 XM_006711876.3 XM_006711877.3 XM_017003738.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
716097 c.1145C>A p.P382H Chr.2: 11658567-11658567 on Build GRCh38
350357 c.1194A>T p.T398T Chr.2: 11658518-11658518 on Build GRCh38
6902970 c.1110G>A p.L370L Chr.2: 11658602-11658602 on Build GRCh38
1690205 c.1181C>T p.T394I Chr.2: 11658531-11658531 on Build GRCh38
6872100 c.1104C>T p.L368L Chr.2: 11658608-11658608 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567504056 Chr.2: 11658207-11658207 on Build GRCh38
rs555315421 Chr.2: 11658333-11658333 on Build GRCh38
rs750364405 Chr.2: 11658407-11658407 on Build GRCh38
rs373736747 Chr.2: 11658332-11658332 on Build GRCh38
rs760557313 Chr.2: 11658552-11658552 on Build GRCh38
rs773177188 Chr.2: 11658553-11658553 on Build GRCh38
rs755612016 Chr.2: 11658502-11658502 on Build GRCh38
rs145638723 Chr.2: 11658488-11658488 on Build GRCh38
rs760238001 Chr.2: 11658258-11658258 on Build GRCh38
rs1030407653 Chr.2: 11658223-11658223 on Build GRCh38
rs537064989 Chr.2: 11658292-11658292 on Build GRCh38
rs972488546 Chr.2: 11658401-11658401 on Build GRCh38
rs144468085 Chr.2: 11658558-11658558 on Build GRCh38
rs202058616 Chr.2: 11658436-11658436 on Build GRCh38
rs146832454 Chr.2: 11658571-11658571 on Build GRCh38
rs748915884 Chr.2: 11658519-11658519 on Build GRCh38
rs766670836 Chr.2: 11658478-11658478 on Build GRCh38
rs1000783701 Chr.2: 11658341-11658341 on Build GRCh38
rs138316342 Chr.2: 11658276-11658276 on Build GRCh38
rs747259673 Chr.2: 11658561-11658561 on Build GRCh38
rs138784927 Chr.2: 11658548-11658548 on Build GRCh38
rs61753869 Chr.2: 11658549-11658549 on Build GRCh38
rs150650740 Chr.2: 11658503-11658503 on Build GRCh38
rs775445585 Chr.2: 11658551-11658551 on Build GRCh38
rs919474578 Chr.2: 11658180-11658180 on Build GRCh38
rs774822478 Chr.2: 11658499-11658500 on Build GRCh38
rs1052710462 Chr.2: 11658416-11658416 on Build GRCh38
rs1011546387 Chr.2: 11658132-11658132 on Build GRCh38
rs949047199 Chr.2: 11658154-11658154 on Build GRCh38
rs770696565 Chr.2: 11658531-11658531 on Build GRCh38
rs13423238 Chr.2: 11658241-11658241 on Build GRCh38
rs370951012 Chr.2: 11658576-11658576 on Build GRCh38
rs558237486 Chr.2: 11658256-11658256 on Build GRCh38
rs77419493 Chr.2: 11658568-11658568 on Build GRCh38
rs567064665 Chr.2: 11658440-11658440 on Build GRCh38
rs1020975864 Chr.2: 11658309-11658309 on Build GRCh38
rs371954886 Chr.2: 11658379-11658379 on Build GRCh38
rs763164358 Chr.2: 11658476-11658476 on Build GRCh38
rs146997854 Chr.2: 11658211-11658211 on Build GRCh38
rs765250739 Chr.2: 11658438-11658438 on Build GRCh38
rs775333750 Chr.2: 11658431-11658431 on Build GRCh38
rs935152560 Chr.2: 11658382-11658382 on Build GRCh38
rs755134638 Chr.2: 11658570-11658570 on Build GRCh38
rs999771864 Chr.2: 11658124-11658124 on Build GRCh38
rs539592729 Chr.2: 11658212-11658212 on Build GRCh38
rs143800347 Chr.2: 11658371-11658371 on Build GRCh38
rs774916831 Chr.2: 11658254-11658254 on Build GRCh38
rs947737804 Chr.2: 11658369-11658369 on Build GRCh38
rs188211375 Chr.2: 11658164-11658164 on Build GRCh38
rs778778739 Chr.2: 11658540-11658540 on Build GRCh38
rs6742234 Chr.2: 11658355-11658355 on Build GRCh38
rs768274285 Chr.2: 11658434-11658434 on Build GRCh38
rs200683246 Chr.2: 11658486-11658486 on Build GRCh38
rs766582840 Chr.2: 11658555-11658555 on Build GRCh38
rs767804665 Chr.2: 11658559-11658559 on Build GRCh38
rs771467024 Chr.2: 11658435-11658437 on Build GRCh38
rs958211443 Chr.2: 11658131-11658131 on Build GRCh38
rs142498936 Chr.2: 11658566-11658566 on Build GRCh38
rs199915986 Chr.2: 11658557-11658557 on Build GRCh38
rs1030392575 Chr.2: 11658550-11658550 on Build GRCh38
rs751670591 Chr.2: 11658291-11658291 on Build GRCh38
rs753167901 Chr.2: 11658499-11658499 on Build GRCh38
rs1025104826 Chr.2: 11658387-11658387 on Build GRCh38
rs113853897 Chr.2: 11658320-11658320 on Build GRCh38
rs150611318 Chr.2: 11658445-11658445 on Build GRCh38
rs1030876790 Chr.2: 11658128-11658128 on Build GRCh38
rs374173549 Chr.2: 11658496-11658496 on Build GRCh38
rs970509973 Chr.2: 11658396-11658396 on Build GRCh38
rs906275635 Chr.2: 11658372-11658372 on Build GRCh38
rs984995366 Chr.2: 11658451-11658451 on Build GRCh38
rs954861996 Chr.2: 11658252-11658252 on Build GRCh38

View Full Product Details