Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 15603704-15604234 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX1
Entrez Gene ID
1653
Gene Name
DEAD/H-box helicase 1
Gene Aliases
DBP-RB, UKVH5d
UniGene
Hs.440599
Gene Chromosome Location
Chr.2: 15591621-15631111 on Build GRCh38
Transcripts
NM_004939.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3391024 c.478A>C p.T160P Chr.2: 15603816-15603816 on Build GRCh38
4954454 c.492A>C p.G164G Chr.2: 15603830-15603830 on Build GRCh38
5587601 c.539A>G p.D180G Chr.2: 15603877-15603877 on Build GRCh38
7077884 c.538G>C p.D180H Chr.2: 15603876-15603876 on Build GRCh38
4965154 c.550G>T p.E184* Chr.2: 15603888-15603888 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs568370525 Chr.2: 15603782-15603782 on Build GRCh38
rs889334487 Chr.2: 15604154-15604154 on Build GRCh38
rs756651702 Chr.2: 15603792-15603792 on Build GRCh38
rs776365618 Chr.2: 15603896-15603896 on Build GRCh38
rs754378722 Chr.2: 15603784-15603784 on Build GRCh38
rs772548001 Chr.2: 15604040-15604040 on Build GRCh38
rs879017488 Chr.2: 15603799-15603799 on Build GRCh38
rs557977571 Chr.2: 15604080-15604080 on Build GRCh38
rs1032573908 Chr.2: 15603920-15603920 on Build GRCh38
rs566154510 Chr.2: 15604007-15604007 on Build GRCh38
rs746753084 Chr.2: 15603866-15603866 on Build GRCh38
rs368177836 Chr.2: 15603926-15603926 on Build GRCh38
rs944100060 Chr.2: 15603984-15603984 on Build GRCh38
rs373610152 Chr.2: 15603917-15603917 on Build GRCh38
rs766485782 Chr.2: 15603763-15603763 on Build GRCh38
rs191827026 Chr.2: 15604125-15604125 on Build GRCh38
rs375565209 Chr.2: 15603922-15603922 on Build GRCh38
rs745493947 Chr.2: 15603807-15603807 on Build GRCh38
rs368104792 Chr.2: 15603900-15603900 on Build GRCh38
rs918412974 Chr.2: 15604031-15604031 on Build GRCh38
rs370867394 Chr.2: 15603897-15603897 on Build GRCh38
rs759815852 Chr.2: 15603769-15603769 on Build GRCh38
rs756499280 Chr.2: 15603777-15603777 on Build GRCh38
rs373674722 Chr.2: 15603862-15603862 on Build GRCh38
rs888376397 Chr.2: 15603733-15603733 on Build GRCh38
rs749992701 Chr.2: 15603942-15603942 on Build GRCh38
rs753103696 Chr.2: 15603772-15603772 on Build GRCh38
rs3213793 Chr.2: 15603999-15603999 on Build GRCh38
rs760968716 Chr.2: 15603915-15603915 on Build GRCh38
rs1056498283 Chr.2: 15603946-15603959 on Build GRCh38
rs895507773 Chr.2: 15604102-15604102 on Build GRCh38
rs747691915 Chr.2: 15604003-15604003 on Build GRCh38
rs550402553 Chr.2: 15603757-15603757 on Build GRCh38
rs751730431 Chr.2: 15603765-15603765 on Build GRCh38
rs916469266 Chr.2: 15603967-15603967 on Build GRCh38
rs766726669 Chr.2: 15604075-15604075 on Build GRCh38
rs568691723 Chr.2: 15604196-15604196 on Build GRCh38
rs897980692 Chr.2: 15604210-15604210 on Build GRCh38
rs371993353 Chr.2: 15603903-15603903 on Build GRCh38
rs895563045 Chr.2: 15603884-15603884 on Build GRCh38
rs772546236 Chr.2: 15603776-15603780 on Build GRCh38
rs779962064 Chr.2: 15603830-15603830 on Build GRCh38
rs770906033 Chr.2: 15603898-15603898 on Build GRCh38
rs767848591 Chr.2: 15603771-15603771 on Build GRCh38
rs537081248 Chr.2: 15604126-15604126 on Build GRCh38
rs947884869 Chr.2: 15603978-15603978 on Build GRCh38
rs1000065221 Chr.2: 15604212-15604212 on Build GRCh38
rs752934252 Chr.2: 15603910-15603910 on Build GRCh38
rs771898232 Chr.2: 15603808-15603808 on Build GRCh38
rs1033985448 Chr.2: 15603790-15603790 on Build GRCh38
rs985422824 Chr.2: 15603948-15603948 on Build GRCh38
rs778551487 Chr.2: 15603805-15603805 on Build GRCh38
rs769320944 Chr.2: 15604037-15604037 on Build GRCh38
rs1055384802 Chr.2: 15604099-15604099 on Build GRCh38
rs911258209 Chr.2: 15603971-15603971 on Build GRCh38
rs535806313 Chr.2: 15603961-15603961 on Build GRCh38
rs371545709 Chr.2: 15603904-15603904 on Build GRCh38
rs11695856 Chr.2: 15604060-15604060 on Build GRCh38
rs1006818360 Chr.2: 15604166-15604166 on Build GRCh38
rs767682755 Chr.2: 15603907-15603907 on Build GRCh38
rs140238349 Chr.2: 15603889-15603889 on Build GRCh38
rs3815690 Chr.2: 15603755-15603755 on Build GRCh38
rs960401941 Chr.2: 15603937-15603937 on Build GRCh38

View Full Product Details