Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 19353433-19353925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OSR1
Entrez Gene ID
130497
Gene Name
odd-skipped related transciption factor 1
Gene Aliases
ODD
UniGene
Hs.123933
Gene Chromosome Location
Chr.2: 19351485-19358611 on Build GRCh38
Transcripts
NM_145260.2 XM_006711942.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2904692 c.226G>A p.V76M Chr.2: 19353580-19353580 on Build GRCh38
4710466 c.47T>C p.L16P Chr.2: 19353759-19353759 on Build GRCh38
2904691 c.245C>T p.A82V Chr.2: 19353561-19353561 on Build GRCh38
4710465 c.105G>T p.S35S Chr.2: 19353701-19353701 on Build GRCh38
4710464 c.344C>T p.P115L Chr.2: 19353462-19353462 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767950855 Chr.2: 19353755-19353755 on Build GRCh38
rs951855533 Chr.2: 19353787-19353787 on Build GRCh38
rs765266778 Chr.2: 19353696-19353696 on Build GRCh38
rs770726635 Chr.2: 19353662-19353662 on Build GRCh38
rs764299282 Chr.2: 19353514-19353514 on Build GRCh38
rs753222267 Chr.2: 19353759-19353759 on Build GRCh38
rs886665270 Chr.2: 19353714-19353714 on Build GRCh38
rs937035681 Chr.2: 19353905-19353905 on Build GRCh38
rs772016590 Chr.2: 19353807-19353807 on Build GRCh38
rs774303008 Chr.2: 19353589-19353589 on Build GRCh38
rs777186968 Chr.2: 19353766-19353766 on Build GRCh38
rs759532279 Chr.2: 19353671-19353671 on Build GRCh38
rs375559109 Chr.2: 19353833-19353833 on Build GRCh38
rs530028849 Chr.2: 19353732-19353732 on Build GRCh38
rs755578955 Chr.2: 19353762-19353762 on Build GRCh38
rs780326478 Chr.2: 19353627-19353627 on Build GRCh38
rs748922957 Chr.2: 19353661-19353661 on Build GRCh38
rs1056276138 Chr.2: 19353538-19353538 on Build GRCh38
rs562873645 Chr.2: 19353477-19353477 on Build GRCh38
rs769498692 Chr.2: 19353573-19353573 on Build GRCh38
rs775583465 Chr.2: 19353687-19353687 on Build GRCh38
rs45620831 Chr.2: 19353889-19353889 on Build GRCh38
rs759842137 Chr.2: 19353836-19353836 on Build GRCh38
rs928284019 Chr.2: 19353893-19353893 on Build GRCh38
rs947794638 Chr.2: 19353588-19353588 on Build GRCh38
rs34464817 Chr.2: 19353693-19353694 on Build GRCh38
rs778633008 Chr.2: 19353796-19353796 on Build GRCh38
rs757601212 Chr.2: 19353524-19353524 on Build GRCh38
rs148662449 Chr.2: 19353559-19353559 on Build GRCh38
rs775530378 Chr.2: 19353808-19353808 on Build GRCh38
rs766269293 Chr.2: 19353499-19353499 on Build GRCh38
rs1011740676 Chr.2: 19353621-19353621 on Build GRCh38
rs764998863 Chr.2: 19353470-19353470 on Build GRCh38
rs139101150 Chr.2: 19353599-19353599 on Build GRCh38
rs776229107 Chr.2: 19353459-19353459 on Build GRCh38
rs747587694 Chr.2: 19353643-19353643 on Build GRCh38
rs978008192 Chr.2: 19353864-19353864 on Build GRCh38
rs868677006 Chr.2: 19353597-19353597 on Build GRCh38
rs45587636 Chr.2: 19353461-19353461 on Build GRCh38
rs767897619 Chr.2: 19353844-19353844 on Build GRCh38
rs199763633 Chr.2: 19353561-19353561 on Build GRCh38
rs142245985 Chr.2: 19353581-19353581 on Build GRCh38
rs777431965 Chr.2: 19353656-19353656 on Build GRCh38
rs375985128 Chr.2: 19353612-19353612 on Build GRCh38
rs560393889 Chr.2: 19353821-19353821 on Build GRCh38
rs763136719 Chr.2: 19353706-19353706 on Build GRCh38
rs745601259 Chr.2: 19353802-19353802 on Build GRCh38
rs756917295 Chr.2: 19353783-19353783 on Build GRCh38
rs752989791 Chr.2: 19353848-19353850 on Build GRCh38
rs907421037 Chr.2: 19353580-19353580 on Build GRCh38
rs140787234 Chr.2: 19353547-19353547 on Build GRCh38
rs200422832 Chr.2: 19353587-19353587 on Build GRCh38
rs780564066 Chr.2: 19353545-19353545 on Build GRCh38
rs779415526 Chr.2: 19353528-19353528 on Build GRCh38
rs772756106 Chr.2: 19353623-19353623 on Build GRCh38
rs762792163 Chr.2: 19353485-19353485 on Build GRCh38
rs370575190 Chr.2: 19353679-19353679 on Build GRCh38
rs962112706 Chr.2: 19353879-19353879 on Build GRCh38
rs373968597 Chr.2: 19353686-19353686 on Build GRCh38
rs750733636 Chr.2: 19353530-19353530 on Build GRCh38
rs939201029 Chr.2: 19353476-19353476 on Build GRCh38
rs747789775 Chr.2: 19353566-19353566 on Build GRCh38
rs752087418 Chr.2: 19353628-19353628 on Build GRCh38
rs758705032 Chr.2: 19353543-19353543 on Build GRCh38
rs759585249 Chr.2: 19353592-19353592 on Build GRCh38
rs748946826 Chr.2: 19353769-19353769 on Build GRCh38
rs372422320 Chr.2: 19353609-19353609 on Build GRCh38
rs755336350 Chr.2: 19353746-19353746 on Build GRCh38
rs898159433 Chr.2: 19353607-19353607 on Build GRCh38
rs773456081 Chr.2: 19353701-19353701 on Build GRCh38
rs1026552672 Chr.2: 19353692-19353692 on Build GRCh38
rs368651434 Chr.2: 19353728-19353728 on Build GRCh38
rs769304282 Chr.2: 19353648-19353648 on Build GRCh38
rs755540358 Chr.2: 19353633-19353633 on Build GRCh38
rs780671260 Chr.2: 19353638-19353638 on Build GRCh38
rs45535040 Chr.2: 19353809-19353809 on Build GRCh38
rs923800427 Chr.2: 19353466-19353466 on Build GRCh38
rs371504366 Chr.2: 19353817-19353817 on Build GRCh38
rs370211451 Chr.2: 19353558-19353558 on Build GRCh38
rs34456793 Chr.2: 19353668-19353668 on Build GRCh38
rs762748716 Chr.2: 19353582-19353582 on Build GRCh38
rs758751821 Chr.2: 19353626-19353626 on Build GRCh38
rs965166465 Chr.2: 19353770-19353770 on Build GRCh38
rs543452352 Chr.2: 19353520-19353520 on Build GRCh38
rs756364623 Chr.2: 19353852-19353858 on Build GRCh38
rs761936888 Chr.2: 19353691-19353691 on Build GRCh38
rs774565727 Chr.2: 19353835-19353835 on Build GRCh38
rs751462463 Chr.2: 19353506-19353506 on Build GRCh38
rs368681707 Chr.2: 19353834-19353834 on Build GRCh38

View Full Product Details