Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 25093009-25093553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFR3B
Entrez Gene ID
22979
Gene Name
EFR3 homolog B
Gene Aliases
KIAA0953
UniGene
Hs.4892
Gene Chromosome Location
Chr.2: 25042104-25159137 on Build GRCh38
Transcripts
NM_001319099.1 NM_014971.1 XM_011532701.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6503850 c.196G>A p.G66S Chr.2: 25093114-25093114 on Build GRCh38
5530381 c.154C>T p.R52C Chr.2: 25093072-25093072 on Build GRCh38
6293826 c.178A>G p.R60G Chr.2: 25093096-25093096 on Build GRCh38
3962689 c.210T>A p.Y70* Chr.2: 25093128-25093128 on Build GRCh38
7365935 c.193G>A p.V65M Chr.2: 25093111-25093111 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs894379188 Chr.2: 25093098-25093098 on Build GRCh38
rs894415230 Chr.2: 25093327-25093327 on Build GRCh38
rs976769444 Chr.2: 25093225-25093225 on Build GRCh38
rs535181000 Chr.2: 25093185-25093185 on Build GRCh38
rs922559380 Chr.2: 25093232-25093232 on Build GRCh38
rs1055619292 Chr.2: 25093293-25093293 on Build GRCh38
rs575203233 Chr.2: 25093449-25093449 on Build GRCh38
rs753036713 Chr.2: 25093154-25093154 on Build GRCh38
rs890129832 Chr.2: 25093308-25093308 on Build GRCh38
rs1029914224 Chr.2: 25093526-25093526 on Build GRCh38
rs993618185 Chr.2: 25093118-25093118 on Build GRCh38
rs2917916 Chr.2: 25093467-25093467 on Build GRCh38
rs549771956 Chr.2: 25093270-25093270 on Build GRCh38
rs1027397016 Chr.2: 25093124-25093124 on Build GRCh38
rs1009039411 Chr.2: 25093339-25093339 on Build GRCh38
rs10495751 Chr.2: 25093176-25093176 on Build GRCh38
rs897528186 Chr.2: 25093389-25093389 on Build GRCh38
rs772911555 Chr.2: 25093081-25093081 on Build GRCh38
rs368952443 Chr.2: 25093110-25093110 on Build GRCh38
rs984285860 Chr.2: 25093175-25093175 on Build GRCh38
rs949930919 Chr.2: 25093344-25093344 on Build GRCh38
rs370067502 Chr.2: 25093043-25093043 on Build GRCh38
rs535729915 Chr.2: 25093304-25093304 on Build GRCh38
rs1053648829 Chr.2: 25093216-25093216 on Build GRCh38
rs371551570 Chr.2: 25093095-25093095 on Build GRCh38
rs868534907 Chr.2: 25093335-25093335 on Build GRCh38
rs1000026291 Chr.2: 25093080-25093080 on Build GRCh38
rs556949999 Chr.2: 25093434-25093434 on Build GRCh38
rs763547780 Chr.2: 25093263-25093263 on Build GRCh38
rs756920067 Chr.2: 25093305-25093305 on Build GRCh38
rs762477103 Chr.2: 25093248-25093248 on Build GRCh38
rs558190450 Chr.2: 25093515-25093515 on Build GRCh38
rs751388167 Chr.2: 25093172-25093172 on Build GRCh38
rs964262713 Chr.2: 25093214-25093214 on Build GRCh38
rs766165766 Chr.2: 25093146-25093146 on Build GRCh38
rs946161881 Chr.2: 25093247-25093247 on Build GRCh38
rs976969856 Chr.2: 25093492-25093492 on Build GRCh38
rs55917417 Chr.2: 25093050-25093050 on Build GRCh38
rs568463019 Chr.2: 25093280-25093280 on Build GRCh38
rs1045380375 Chr.2: 25093378-25093378 on Build GRCh38
rs977476460 Chr.2: 25093505-25093506 on Build GRCh38
rs924679315 Chr.2: 25093506-25093506 on Build GRCh38
rs897581859 Chr.2: 25093114-25093114 on Build GRCh38
rs1018433194 Chr.2: 25093451-25093451 on Build GRCh38
rs993174115 Chr.2: 25093411-25093411 on Build GRCh38
rs936095106 Chr.2: 25093506-25093506 on Build GRCh38
rs1027839242 Chr.2: 25093427-25093427 on Build GRCh38
rs1013888577 Chr.2: 25093104-25093104 on Build GRCh38
rs1027816668 Chr.2: 25093418-25093418 on Build GRCh38
rs1014639272 Chr.2: 25093347-25093347 on Build GRCh38
rs1041828742 Chr.2: 25093177-25093177 on Build GRCh38
rs953030523 Chr.2: 25093148-25093148 on Build GRCh38
rs967823143 Chr.2: 25093527-25093527 on Build GRCh38

View Full Product Details