Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 25741315-25741827 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASXL2
Entrez Gene ID
55252
Gene Name
additional sex combs like 2, transcriptional regulator
Gene Aliases
ASXH2
UniGene
Hs.119815
Gene Chromosome Location
Chr.2: 25733758-25878485 on Build GRCh38
Transcripts
NM_018263.4 XM_006712039.3 XM_006712040.2 XM_011532950.2 XM_011532951.2 XM_017004429.1 XM_017004430.1

SNP Detail

SNP ID Chromosome Location
rs1023439853 Chr.2: 25741429-25741429 on Build GRCh38
rs561001715 Chr.2: 25741407-25741407 on Build GRCh38
rs1008485243 Chr.2: 25741415-25741415 on Build GRCh38
rs567310887 Chr.2: 25741622-25741622 on Build GRCh38
rs893070432 Chr.2: 25741696-25741696 on Build GRCh38
rs981332443 Chr.2: 25741649-25741650 on Build GRCh38
rs950918432 Chr.2: 25741522-25741522 on Build GRCh38
rs2033173 Chr.2: 25741377-25741377 on Build GRCh38
rs772458095 Chr.2: 25741765-25741765 on Build GRCh38
rs368126212 Chr.2: 25741701-25741701 on Build GRCh38
rs148225214 Chr.2: 25741365-25741365 on Build GRCh38
rs141131150 Chr.2: 25741550-25741550 on Build GRCh38
rs139932342 Chr.2: 25741791-25741792 on Build GRCh38
rs956085857 Chr.2: 25741740-25741740 on Build GRCh38
rs564330767 Chr.2: 25741551-25741551 on Build GRCh38
rs755853607 Chr.2: 25741664-25741664 on Build GRCh38
rs970151910 Chr.2: 25741467-25741467 on Build GRCh38
rs571158285 Chr.2: 25741468-25741468 on Build GRCh38
rs764237396 Chr.2: 25741395-25741395 on Build GRCh38
rs112007704 Chr.2: 25741665-25741665 on Build GRCh38
rs549775642 Chr.2: 25741450-25741450 on Build GRCh38
rs981956407 Chr.2: 25741489-25741489 on Build GRCh38
rs186630033 Chr.2: 25741625-25741625 on Build GRCh38
rs973778675 Chr.2: 25741590-25741590 on Build GRCh38
rs547814965 Chr.2: 25741544-25741544 on Build GRCh38
rs983633861 Chr.2: 25741525-25741525 on Build GRCh38
rs1052999970 Chr.2: 25741670-25741670 on Build GRCh38
rs182487740 Chr.2: 25741385-25741385 on Build GRCh38
rs558485239 Chr.2: 25741739-25741739 on Build GRCh38
rs577212127 Chr.2: 25741791-25741791 on Build GRCh38
rs190316584 Chr.2: 25741346-25741346 on Build GRCh38
rs890957798 Chr.2: 25741662-25741662 on Build GRCh38
rs576091963 Chr.2: 25741711-25741711 on Build GRCh38
rs537167785 Chr.2: 25741726-25741726 on Build GRCh38
rs1050937651 Chr.2: 25741358-25741358 on Build GRCh38
rs17047139 Chr.2: 25741421-25741421 on Build GRCh38
rs188996748 Chr.2: 25741636-25741636 on Build GRCh38
rs539278424 Chr.2: 25741514-25741514 on Build GRCh38
rs1035860390 Chr.2: 25741506-25741506 on Build GRCh38
rs934491331 Chr.2: 25741669-25741669 on Build GRCh38
rs1053293449 Chr.2: 25741349-25741349 on Build GRCh38

View Full Product Details