Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26262942-26263484 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HADHB
Entrez Gene ID
3032
Gene Name
hydroxyacyl-CoA dehydrogenase/3-ketoacyl-CoA thiolase/enoyl-CoA hydratase (trifunctional protein), beta subunit
Gene Aliases
ECHB, MSTP029, MTPB, TP-BETA
UniGene
Hs.515848
Gene Chromosome Location
Chr.2: 26244748-26290465 on Build GRCh38
Transcripts
NM_000183.2 NM_001281512.1 NM_001281513.1 XM_011532803.1 XM_017003939.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6677469 c.170T>A p.V57E Chr.2: 26263440-26263440 on Build GRCh38
1407195 c.184A>G p.T62A Chr.2: 26263454-26263454 on Build GRCh38
385048 c.207T>A p.T69T Chr.2: 26263477-26263477 on Build GRCh38
6677471 c.144A>C p.K48N Chr.2: 26263414-26263414 on Build GRCh38
3910086 c.181C>T p.R61C Chr.2: 26263451-26263451 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879094220 Chr.2: 26263317-26263317 on Build GRCh38
rs749703431 Chr.2: 26263299-26263299 on Build GRCh38
rs534682304 Chr.2: 26263337-26263337 on Build GRCh38
rs780351691 Chr.2: 26263451-26263451 on Build GRCh38
rs569553174 Chr.2: 26263235-26263235 on Build GRCh38
rs576889882 Chr.2: 26263290-26263290 on Build GRCh38
rs1007318629 Chr.2: 26262993-26262993 on Build GRCh38
rs553040477 Chr.2: 26263382-26263382 on Build GRCh38
rs546691836 Chr.2: 26263070-26263070 on Build GRCh38
rs121913132 Chr.2: 26263452-26263452 on Build GRCh38
rs1048447556 Chr.2: 26263426-26263426 on Build GRCh38
rs998297141 Chr.2: 26263126-26263126 on Build GRCh38
rs990909709 Chr.2: 26263285-26263285 on Build GRCh38
rs1032514131 Chr.2: 26263248-26263248 on Build GRCh38
rs201804764 Chr.2: 26263441-26263441 on Build GRCh38
rs937101675 Chr.2: 26263291-26263292 on Build GRCh38
rs558674244 Chr.2: 26263256-26263256 on Build GRCh38
rs1037913162 Chr.2: 26263281-26263281 on Build GRCh38
rs762111652 Chr.2: 26263341-26263341 on Build GRCh38
rs564433178 Chr.2: 26262985-26262985 on Build GRCh38
rs752631689 Chr.2: 26263185-26263185 on Build GRCh38
rs202105626 Chr.2: 26263071-26263071 on Build GRCh38
rs147599321 Chr.2: 26263027-26263027 on Build GRCh38
rs916481934 Chr.2: 26263306-26263306 on Build GRCh38
rs977769743 Chr.2: 26263230-26263230 on Build GRCh38
rs754385341 Chr.2: 26263261-26263261 on Build GRCh38
rs920334279 Chr.2: 26263286-26263286 on Build GRCh38
rs1014554092 Chr.2: 26263384-26263384 on Build GRCh38
rs376739522 Chr.2: 26263062-26263062 on Build GRCh38
rs754537494 Chr.2: 26263404-26263404 on Build GRCh38
rs768738452 Chr.2: 26263454-26263454 on Build GRCh38
rs772220363 Chr.2: 26263444-26263444 on Build GRCh38
rs369260367 Chr.2: 26263161-26263161 on Build GRCh38
rs568097891 Chr.2: 26263144-26263144 on Build GRCh38
rs867374777 Chr.2: 26263458-26263458 on Build GRCh38
rs1018506295 Chr.2: 26263011-26263011 on Build GRCh38
rs750072104 Chr.2: 26263353-26263353 on Build GRCh38
rs139107013 Chr.2: 26263299-26263299 on Build GRCh38
rs746076418 Chr.2: 26263443-26263443 on Build GRCh38
rs908000054 Chr.2: 26263251-26263251 on Build GRCh38
rs751072011 Chr.2: 26263395-26263395 on Build GRCh38
rs966428594 Chr.2: 26263202-26263202 on Build GRCh38
rs574464531 Chr.2: 26263405-26263405 on Build GRCh38
rs909977699 Chr.2: 26263427-26263427 on Build GRCh38
rs762888164 Chr.2: 26263197-26263197 on Build GRCh38
rs755967503 Chr.2: 26263424-26263424 on Build GRCh38
rs759380176 Chr.2: 26262997-26262997 on Build GRCh38
rs536860962 Chr.2: 26263239-26263239 on Build GRCh38
rs372192901 Chr.2: 26263345-26263345 on Build GRCh38
rs182541227 Chr.2: 26263164-26263164 on Build GRCh38
rs765893367 Chr.2: 26263386-26263386 on Build GRCh38
rs548074813 Chr.2: 26263226-26263226 on Build GRCh38
rs75496350 Chr.2: 26263312-26263315 on Build GRCh38

View Full Product Details