Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26278413-26278925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HADHB
Entrez Gene ID
3032
Gene Name
hydroxyacyl-CoA dehydrogenase/3-ketoacyl-CoA thiolase/enoyl-CoA hydratase (trifunctional protein), beta subunit
Gene Aliases
ECHB, MSTP029, MTPB, TP-BETA
UniGene
Hs.515848
Gene Chromosome Location
Chr.2: 26244748-26290465 on Build GRCh38
Transcripts
NM_000183.2 NM_001281512.1 NM_001281513.1 XM_011532803.1 XM_017003939.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6677476 c.508G>A p.D170N Chr.2: 26278679-26278679 on Build GRCh38
1019324 c.480C>T p.I160I Chr.2: 26278651-26278651 on Build GRCh38
6577985 c.539G>C p.R180T Chr.2: 26278710-26278710 on Build GRCh38
5099406 c.542G>A p.R181Q Chr.2: 26278779-26278779 on Build GRCh38
5144856 c.536A>C p.K179T Chr.2: 26278773-26278773 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757949243 Chr.2: 26278789-26278789 on Build GRCh38
rs767827358 Chr.2: 26278576-26278576 on Build GRCh38
rs778346714 Chr.2: 26278680-26278680 on Build GRCh38
rs948390021 Chr.2: 26278461-26278461 on Build GRCh38
rs756770407 Chr.2: 26278610-26278610 on Build GRCh38
rs17600515 Chr.2: 26278521-26278521 on Build GRCh38
rs546252491 Chr.2: 26278841-26278841 on Build GRCh38
rs778077779 Chr.2: 26278730-26278730 on Build GRCh38
rs765246893 Chr.2: 26278826-26278826 on Build GRCh38
rs17851200 Chr.2: 26278796-26278796 on Build GRCh38
rs372006332 Chr.2: 26278739-26278739 on Build GRCh38
rs775855041 Chr.2: 26278838-26278838 on Build GRCh38
rs746755261 Chr.2: 26278720-26278720 on Build GRCh38
rs756489045 Chr.2: 26278672-26278672 on Build GRCh38
rs575370098 Chr.2: 26278506-26278506 on Build GRCh38
rs35274385 Chr.2: 26278679-26278679 on Build GRCh38
rs755803677 Chr.2: 26278594-26278594 on Build GRCh38
rs201297849 Chr.2: 26278644-26278644 on Build GRCh38
rs752963725 Chr.2: 26278670-26278670 on Build GRCh38
rs889174248 Chr.2: 26278845-26278845 on Build GRCh38
rs763674788 Chr.2: 26278673-26278673 on Build GRCh38
rs534616210 Chr.2: 26278778-26278778 on Build GRCh38
rs771016054 Chr.2: 26278810-26278810 on Build GRCh38
rs540213260 Chr.2: 26278548-26278548 on Build GRCh38
rs768588630 Chr.2: 26278721-26278721 on Build GRCh38
rs760009170 Chr.2: 26278668-26278668 on Build GRCh38
rs752021591 Chr.2: 26278564-26278564 on Build GRCh38
rs780072612 Chr.2: 26278691-26278691 on Build GRCh38
rs999181743 Chr.2: 26278490-26278490 on Build GRCh38
rs751550829 Chr.2: 26278792-26278792 on Build GRCh38
rs775200563 Chr.2: 26278651-26278651 on Build GRCh38
rs572908123 Chr.2: 26278528-26278528 on Build GRCh38
rs181097368 Chr.2: 26278514-26278514 on Build GRCh38
rs749861331 Chr.2: 26278779-26278779 on Build GRCh38
rs772196306 Chr.2: 26278822-26278822 on Build GRCh38
rs753474001 Chr.2: 26278759-26278759 on Build GRCh38
rs1017633093 Chr.2: 26278718-26278718 on Build GRCh38
rs933023555 Chr.2: 26278781-26278781 on Build GRCh38
rs557241139 Chr.2: 26278479-26278479 on Build GRCh38
rs745857676 Chr.2: 26278628-26278628 on Build GRCh38
rs1008052766 Chr.2: 26278867-26278867 on Build GRCh38
rs552292698 Chr.2: 26278754-26278754 on Build GRCh38
rs781502172 Chr.2: 26278600-26278600 on Build GRCh38
rs770105592 Chr.2: 26278654-26278654 on Build GRCh38
rs935016371 Chr.2: 26278829-26278829 on Build GRCh38
rs1011444793 Chr.2: 26278557-26278557 on Build GRCh38
rs893624843 Chr.2: 26278527-26278527 on Build GRCh38
rs776077327 Chr.2: 26278749-26278749 on Build GRCh38
rs966108514 Chr.2: 26278645-26278645 on Build GRCh38
rs951478194 Chr.2: 26278698-26278698 on Build GRCh38
rs771889730 Chr.2: 26278646-26278646 on Build GRCh38
rs781179195 Chr.2: 26278797-26278797 on Build GRCh38
rs568752809 Chr.2: 26278898-26278898 on Build GRCh38
rs779214608 Chr.2: 26278817-26278817 on Build GRCh38
rs892966891 Chr.2: 26278546-26278546 on Build GRCh38
rs202174966 Chr.2: 26278660-26278660 on Build GRCh38
rs777434443 Chr.2: 26278603-26278603 on Build GRCh38
rs752914522 Chr.2: 26278657-26278657 on Build GRCh38
rs776385012 Chr.2: 26278561-26278561 on Build GRCh38
rs374852324 Chr.2: 26278681-26278681 on Build GRCh38
rs763070593 Chr.2: 26278663-26278663 on Build GRCh38
rs367891116 Chr.2: 26278685-26278685 on Build GRCh38
rs754819096 Chr.2: 26278794-26278794 on Build GRCh38
rs984371001 Chr.2: 26278704-26278704 on Build GRCh38
rs764507324 Chr.2: 26278755-26278755 on Build GRCh38
rs745878042 Chr.2: 26278820-26278820 on Build GRCh38
rs143683481 Chr.2: 26278736-26278736 on Build GRCh38
rs777022830 Chr.2: 26278847-26278847 on Build GRCh38
rs138696018 Chr.2: 26278731-26278731 on Build GRCh38
rs772405727 Chr.2: 26278745-26278745 on Build GRCh38
rs752371387 Chr.2: 26278585-26278585 on Build GRCh38
rs1022372780 Chr.2: 26278562-26278562 on Build GRCh38
rs755305131 Chr.2: 26278565-26278565 on Build GRCh38
rs774905703 Chr.2: 26278665-26278665 on Build GRCh38
rs756299205 Chr.2: 26278718-26278718 on Build GRCh38
rs568062748 Chr.2: 26278482-26278482 on Build GRCh38
rs766573917 Chr.2: 26278664-26278664 on Build GRCh38
rs749486969 Chr.2: 26278806-26278806 on Build GRCh38
rs752524069 Chr.2: 26278872-26278872 on Build GRCh38
rs371100088 Chr.2: 26278652-26278652 on Build GRCh38
rs753492483 Chr.2: 26278678-26278678 on Build GRCh38
rs142475516 Chr.2: 26278760-26278760 on Build GRCh38
rs984941671 Chr.2: 26278723-26278723 on Build GRCh38
rs548272672 Chr.2: 26278448-26278448 on Build GRCh38
rs779801534 Chr.2: 26278790-26278790 on Build GRCh38
rs778752845 Chr.2: 26278622-26278622 on Build GRCh38
rs371257973 Chr.2: 26278604-26278604 on Build GRCh38
rs778744548 Chr.2: 26278800-26278804 on Build GRCh38

View Full Product Details