Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27022253-27022802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAPRE3
Entrez Gene ID
22924
Gene Name
microtubule associated protein RP/EB family member 3
Gene Aliases
EB3, EBF3, EBF3-S, RP3
UniGene
Hs.515860
Gene Chromosome Location
Chr.2: 26970371-27027219 on Build GRCh38
Transcripts
NM_001303050.1 NM_012326.3 XM_006711967.3 XM_011532700.1 XM_017003597.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5735982 c.111G>T p.Q37H Chr.2: 27022329-27022329 on Build GRCh38
359409 c.121+2T>C p.? Chr.2: 27022341-27022341 on Build GRCh38
6450321 c.120A>C p.S40S Chr.2: 27022338-27022338 on Build GRCh38
720678 c.45G>T p.L15L Chr.2: 27022263-27022263 on Build GRCh38
6995830 c.50G>A p.R17H Chr.2: 27022268-27022268 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs932429301 Chr.2: 27022380-27022380 on Build GRCh38
rs912332970 Chr.2: 27022754-27022754 on Build GRCh38
rs545842967 Chr.2: 27022479-27022479 on Build GRCh38
rs995174481 Chr.2: 27022768-27022768 on Build GRCh38
rs760913125 Chr.2: 27022375-27022375 on Build GRCh38
rs561665976 Chr.2: 27022545-27022545 on Build GRCh38
rs973932387 Chr.2: 27022683-27022683 on Build GRCh38
rs151223078 Chr.2: 27022310-27022310 on Build GRCh38
rs374386550 Chr.2: 27022664-27022664 on Build GRCh38
rs1036945076 Chr.2: 27022759-27022759 on Build GRCh38
rs990068501 Chr.2: 27022579-27022579 on Build GRCh38
rs750022436 Chr.2: 27022522-27022522 on Build GRCh38
rs998523796 Chr.2: 27022395-27022395 on Build GRCh38
rs898162943 Chr.2: 27022452-27022452 on Build GRCh38
rs148013056 Chr.2: 27022583-27022583 on Build GRCh38
rs185199967 Chr.2: 27022367-27022367 on Build GRCh38
rs530948772 Chr.2: 27022598-27022598 on Build GRCh38
rs908444612 Chr.2: 27022589-27022589 on Build GRCh38
rs529126170 Chr.2: 27022567-27022567 on Build GRCh38
rs141366427 Chr.2: 27022478-27022478 on Build GRCh38
rs1031898819 Chr.2: 27022396-27022396 on Build GRCh38
rs556967512 Chr.2: 27022372-27022372 on Build GRCh38
rs1004118246 Chr.2: 27022449-27022449 on Build GRCh38
rs878897779 Chr.2: 27022406-27022406 on Build GRCh38
rs1050792882 Chr.2: 27022732-27022732 on Build GRCh38
rs572989315 Chr.2: 27022530-27022530 on Build GRCh38
rs1037131831 Chr.2: 27022451-27022451 on Build GRCh38
rs199621083 Chr.2: 27022501-27022502 on Build GRCh38
rs1051395686 Chr.2: 27022687-27022687 on Build GRCh38
rs540183746 Chr.2: 27022532-27022532 on Build GRCh38
rs932481555 Chr.2: 27022711-27022711 on Build GRCh38
rs768144243 Chr.2: 27022294-27022294 on Build GRCh38
rs1054460707 Chr.2: 27022351-27022351 on Build GRCh38
rs891047964 Chr.2: 27022416-27022416 on Build GRCh38
rs368278253 Chr.2: 27022347-27022347 on Build GRCh38
rs141664201 Chr.2: 27022761-27022761 on Build GRCh38
rs769194250 Chr.2: 27022543-27022543 on Build GRCh38
rs535456601 Chr.2: 27022371-27022371 on Build GRCh38
rs564754711 Chr.2: 27022547-27022547 on Build GRCh38
rs936126887 Chr.2: 27022322-27022322 on Build GRCh38
rs924648419 Chr.2: 27022555-27022555 on Build GRCh38
rs771693518 Chr.2: 27022374-27022395 on Build GRCh38
rs912381455 Chr.2: 27022697-27022697 on Build GRCh38
rs939974038 Chr.2: 27022758-27022758 on Build GRCh38
rs548184170 Chr.2: 27022569-27022569 on Build GRCh38
rs921132967 Chr.2: 27022684-27022684 on Build GRCh38
rs149090215 Chr.2: 27022293-27022293 on Build GRCh38
rs753961112 Chr.2: 27022390-27022390 on Build GRCh38
rs977698953 Chr.2: 27022551-27022551 on Build GRCh38
rs747545412 Chr.2: 27022313-27022313 on Build GRCh38
rs934313677 Chr.2: 27022756-27022756 on Build GRCh38
rs959072982 Chr.2: 27022493-27022493 on Build GRCh38
rs7355650 Chr.2: 27022607-27022607 on Build GRCh38
rs773083055 Chr.2: 27022349-27022349 on Build GRCh38
rs146977621 Chr.2: 27022507-27022507 on Build GRCh38
rs764167314 Chr.2: 27022389-27022389 on Build GRCh38

View Full Product Details